Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81066083-81066589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf46
Entrez Gene ID
123775
Gene Name
chromosome 16 open reading frame 46
Gene Aliases
-
UniGene
Hs.658684
Gene Chromosome Location
Chr.16: 81053497-81077267 on Build GRCh38
Transcripts
NM_001100873.1 NM_152337.2 XM_005255792.2 XM_011522848.2 XM_011522849.2 XM_017022916.1 XM_017022917.1

SNP Detail

SNP ID Chromosome Location
rs1005299115 Chr.16: 81066231-81066231 on Build GRCh38
rs536140642 Chr.16: 81066272-81066272 on Build GRCh38
rs983957839 Chr.16: 81066539-81066539 on Build GRCh38
rs561683996 Chr.16: 81066114-81066114 on Build GRCh38
rs986690303 Chr.16: 81066314-81066314 on Build GRCh38
rs115790866 Chr.16: 81066297-81066297 on Build GRCh38
rs1011786043 Chr.16: 81066444-81066444 on Build GRCh38
rs766790788 Chr.16: 81066391-81066391 on Build GRCh38
rs1019752376 Chr.16: 81066463-81066463 on Build GRCh38
rs975009190 Chr.16: 81066506-81066507 on Build GRCh38
rs995340162 Chr.16: 81066497-81066497 on Build GRCh38
rs774348810 Chr.16: 81066318-81066318 on Build GRCh38
rs949160097 Chr.16: 81066397-81066397 on Build GRCh38
rs9937594 Chr.16: 81066375-81066375 on Build GRCh38
rs1014986607 Chr.16: 81066486-81066486 on Build GRCh38
rs908351394 Chr.16: 81066540-81066540 on Build GRCh38
rs887826700 Chr.16: 81066221-81066221 on Build GRCh38
rs961209246 Chr.16: 81066349-81066349 on Build GRCh38
rs745697936 Chr.16: 81066243-81066243 on Build GRCh38
rs937655855 Chr.16: 81066198-81066198 on Build GRCh38
rs902640381 Chr.16: 81066247-81066247 on Build GRCh38
rs145763276 Chr.16: 81066496-81066496 on Build GRCh38
rs867811430 Chr.16: 81066518-81066518 on Build GRCh38
rs181613800 Chr.16: 81066269-81066269 on Build GRCh38
rs148884203 Chr.16: 81066162-81066162 on Build GRCh38
rs189318872 Chr.16: 81066215-81066215 on Build GRCh38
rs974415630 Chr.16: 81066370-81066370 on Build GRCh38
rs548342879 Chr.16: 81066152-81066152 on Build GRCh38
rs747482668 Chr.16: 81066392-81066392 on Build GRCh38
rs552581683 Chr.16: 81066507-81066507 on Build GRCh38
rs550811107 Chr.16: 81066471-81066471 on Build GRCh38
rs804895 Chr.16: 81066108-81066108 on Build GRCh38
rs975314536 Chr.16: 81066356-81066358 on Build GRCh38
rs182880290 Chr.16: 81066446-81066446 on Build GRCh38
rs1001531706 Chr.16: 81066252-81066252 on Build GRCh38
rs571486748 Chr.16: 81066380-81066380 on Build GRCh38
rs1030449049 Chr.16: 81066282-81066282 on Build GRCh38
rs777460758 Chr.16: 81066442-81066442 on Build GRCh38
rs575134541 Chr.16: 81066371-81066371 on Build GRCh38
rs186388512 Chr.16: 81066513-81066513 on Build GRCh38
rs563549601 Chr.16: 81066485-81066485 on Build GRCh38
rs1005390960 Chr.16: 81066464-81066464 on Build GRCh38
rs145557611 Chr.16: 81066156-81066156 on Build GRCh38
rs879617458 Chr.16: 81066251-81066251 on Build GRCh38
rs568590593 Chr.16: 81066566-81066566 on Build GRCh38
rs970389004 Chr.16: 81066557-81066557 on Build GRCh38
rs750920481 Chr.16: 81066379-81066379 on Build GRCh38
rs11865620 Chr.16: 81066246-81066246 on Build GRCh38
rs939823370 Chr.16: 81066564-81066564 on Build GRCh38
rs186494872 Chr.16: 81066122-81066122 on Build GRCh38
rs9937014 Chr.16: 81066435-81066435 on Build GRCh38
rs189765451 Chr.16: 81066514-81066514 on Build GRCh38
rs191552389 Chr.16: 81066417-81066417 on Build GRCh38
rs890428870 Chr.16: 81066452-81066452 on Build GRCh38
rs530497496 Chr.16: 81066510-81066510 on Build GRCh38
rs902691608 Chr.16: 81066414-81066414 on Build GRCh38
rs896229808 Chr.16: 81066239-81066239 on Build GRCh38
rs987024330 Chr.16: 81066150-81066150 on Build GRCh38
rs868653862 Chr.16: 81066520-81066520 on Build GRCh38
rs919607018 Chr.16: 81066171-81066171 on Build GRCh38
rs535258387 Chr.16: 81066241-81066241 on Build GRCh38
rs939936436 Chr.16: 81066161-81066161 on Build GRCh38
rs540080806 Chr.16: 81066427-81066427 on Build GRCh38
rs143484143 Chr.16: 81066187-81066187 on Build GRCh38
rs186752885 Chr.16: 81066378-81066378 on Build GRCh38
rs998601826 Chr.16: 81066460-81066460 on Build GRCh38
rs531687153 Chr.16: 81066319-81066319 on Build GRCh38
rs917589072 Chr.16: 81066535-81066535 on Build GRCh38
rs1042103721 Chr.16: 81066401-81066401 on Build GRCh38
rs763015902 Chr.16: 81066276-81066276 on Build GRCh38

View Full Product Details