Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84121837-84122362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSDL1
Entrez Gene ID
83693
Gene Name
hydroxysteroid dehydrogenase like 1
Gene Aliases
SDR12C3
UniGene
Hs.555992
Gene Chromosome Location
Chr.16: 84122139-84145195 on Build GRCh38
Transcripts
NM_001146051.1 NM_031463.4 XM_005256189.2

SNP Detail

SNP ID Chromosome Location
rs774716858 Chr.16: 84122052-84122052 on Build GRCh38
rs1045796489 Chr.16: 84122323-84122323 on Build GRCh38
rs1007996096 Chr.16: 84122202-84122202 on Build GRCh38
rs189704956 Chr.16: 84122331-84122331 on Build GRCh38
rs1053453906 Chr.16: 84122118-84122118 on Build GRCh38
rs765814387 Chr.16: 84122265-84122265 on Build GRCh38
rs184063793 Chr.16: 84122296-84122296 on Build GRCh38
rs755946680 Chr.16: 84122044-84122044 on Build GRCh38
rs1049192146 Chr.16: 84122056-84122056 on Build GRCh38
rs953764515 Chr.16: 84122161-84122161 on Build GRCh38
rs1011679581 Chr.16: 84122217-84122218 on Build GRCh38
rs750278670 Chr.16: 84121982-84121982 on Build GRCh38
rs919032818 Chr.16: 84122035-84122035 on Build GRCh38
rs550710280 Chr.16: 84122154-84122154 on Build GRCh38
rs915976376 Chr.16: 84121956-84121956 on Build GRCh38
rs374319864 Chr.16: 84122251-84122265 on Build GRCh38
rs191129238 Chr.16: 84122269-84122269 on Build GRCh38
rs1045679045 Chr.16: 84122073-84122073 on Build GRCh38
rs370615866 Chr.16: 84122083-84122084 on Build GRCh38
rs998744041 Chr.16: 84122098-84122098 on Build GRCh38
rs111273677 Chr.16: 84122332-84122332 on Build GRCh38
rs956511790 Chr.16: 84121910-84121910 on Build GRCh38
rs919396880 Chr.16: 84121880-84121880 on Build GRCh38
rs887955438 Chr.16: 84122102-84122102 on Build GRCh38
rs1013131757 Chr.16: 84122123-84122123 on Build GRCh38
rs987994795 Chr.16: 84121919-84121919 on Build GRCh38
rs138164369 Chr.16: 84122103-84122103 on Build GRCh38
rs972457987 Chr.16: 84122252-84122252 on Build GRCh38
rs554069616 Chr.16: 84122238-84122238 on Build GRCh38
rs773699812 Chr.16: 84122229-84122229 on Build GRCh38
rs891859828 Chr.16: 84122201-84122201 on Build GRCh38
rs780836078 Chr.16: 84121901-84121901 on Build GRCh38
rs1032208516 Chr.16: 84121903-84121903 on Build GRCh38
rs948782296 Chr.16: 84122309-84122309 on Build GRCh38
rs149584051 Chr.16: 84122289-84122289 on Build GRCh38
rs916092371 Chr.16: 84122293-84122293 on Build GRCh38
rs543083932 Chr.16: 84122237-84122251 on Build GRCh38
rs776168957 Chr.16: 84122320-84122320 on Build GRCh38
rs186800236 Chr.16: 84122237-84122237 on Build GRCh38
rs529173608 Chr.16: 84122008-84122008 on Build GRCh38
rs1022101373 Chr.16: 84122218-84122218 on Build GRCh38
rs878881380 Chr.16: 84122317-84122317 on Build GRCh38
rs893088560 Chr.16: 84122119-84122119 on Build GRCh38
rs948834475 Chr.16: 84122043-84122043 on Build GRCh38
rs530805237 Chr.16: 84121992-84121992 on Build GRCh38
rs116561945 Chr.16: 84121873-84121873 on Build GRCh38
rs1045084067 Chr.16: 84122128-84122128 on Build GRCh38
rs967926690 Chr.16: 84121946-84121946 on Build GRCh38
rs930797262 Chr.16: 84121894-84121894 on Build GRCh38
rs984509125 Chr.16: 84122330-84122330 on Build GRCh38
rs780191507 Chr.16: 84122200-84122200 on Build GRCh38
rs1049267125 Chr.16: 84121951-84121951 on Build GRCh38
rs944408379 Chr.16: 84121966-84121966 on Build GRCh38
rs746776072 Chr.16: 84121929-84121929 on Build GRCh38
rs1031741196 Chr.16: 84122176-84122176 on Build GRCh38
rs907230884 Chr.16: 84122092-84122092 on Build GRCh38
rs1011823201 Chr.16: 84122153-84122153 on Build GRCh38

View Full Product Details