Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85918326-85918827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF8
Entrez Gene ID
3394
Gene Name
interferon regulatory factor 8
Gene Aliases
H-ICSBP, ICSBP, ICSBP1, IMD32A, IMD32B, IRF-8
UniGene
Hs.137427
Gene Chromosome Location
Chr.16: 85899168-85922606 on Build GRCh38
Transcripts
NM_002163.2 XM_017023199.1 NR_106832.1

Gene Symbol
MIR6774
Entrez Gene ID
102466732
Gene Name
microRNA 6774
Gene Aliases
hsa-mir-6774
UniGene
Gene Chromosome Location
Chr.16: 85918347-85918416 on Build GRCh38
Transcripts
NM_002163.2 XM_017023199.1 NR_106832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574128 c.739C>A p.L247M Chr.16: 85918554-85918554 on Build GRCh38
6555967 c.762G>T p.P254P Chr.16: 85918577-85918577 on Build GRCh38
4579347 c.828C>T p.R276R Chr.16: 85918643-85918643 on Build GRCh38
6555965 c.646G>T p.V216L Chr.16: 85918461-85918461 on Build GRCh38
4579345 c.664A>C p.T222P Chr.16: 85918479-85918479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995903402 Chr.16: 85918652-85918652 on Build GRCh38
rs754529909 Chr.16: 85918617-85918617 on Build GRCh38
rs575823888 Chr.16: 85918782-85918782 on Build GRCh38
rs774890679 Chr.16: 85918390-85918390 on Build GRCh38
rs747618015 Chr.16: 85918622-85918622 on Build GRCh38
rs749472608 Chr.16: 85918785-85918785 on Build GRCh38
rs764103061 Chr.16: 85918583-85918583 on Build GRCh38
rs185056029 Chr.16: 85918398-85918398 on Build GRCh38
rs372510203 Chr.16: 85918679-85918679 on Build GRCh38
rs781716749 Chr.16: 85918781-85918781 on Build GRCh38
rs1011079712 Chr.16: 85918685-85918685 on Build GRCh38
rs772016377 Chr.16: 85918743-85918743 on Build GRCh38
rs753421098 Chr.16: 85918759-85918759 on Build GRCh38
rs770113596 Chr.16: 85918643-85918643 on Build GRCh38
rs202107230 Chr.16: 85918680-85918680 on Build GRCh38
rs751576714 Chr.16: 85918584-85918584 on Build GRCh38
rs754894066 Chr.16: 85918361-85918361 on Build GRCh38
rs759805702 Chr.16: 85918580-85918580 on Build GRCh38
rs749986122 Chr.16: 85918612-85918612 on Build GRCh38
rs755787508 Chr.16: 85918614-85918614 on Build GRCh38
rs750578118 Chr.16: 85918392-85918392 on Build GRCh38
rs777712868 Chr.16: 85918454-85918454 on Build GRCh38
rs144550295 Chr.16: 85918667-85918667 on Build GRCh38
rs760447497 Chr.16: 85918748-85918748 on Build GRCh38
rs775730870 Chr.16: 85918581-85918581 on Build GRCh38
rs902811291 Chr.16: 85918359-85918359 on Build GRCh38
rs746599540 Chr.16: 85918460-85918460 on Build GRCh38
rs374144564 Chr.16: 85918742-85918742 on Build GRCh38
rs35288009 Chr.16: 85918492-85918492 on Build GRCh38
rs755125266 Chr.16: 85918418-85918418 on Build GRCh38
rs199975901 Chr.16: 85918380-85918380 on Build GRCh38
rs777551539 Chr.16: 85918372-85918372 on Build GRCh38
rs61995933 Chr.16: 85918709-85918709 on Build GRCh38
rs748765030 Chr.16: 85918616-85918616 on Build GRCh38
rs766228782 Chr.16: 85918754-85918754 on Build GRCh38
rs763945332 Chr.16: 85918527-85918527 on Build GRCh38
rs1003823910 Chr.16: 85918710-85918710 on Build GRCh38
rs767707170 Chr.16: 85918353-85918353 on Build GRCh38
rs142267779 Chr.16: 85918539-85918539 on Build GRCh38
rs368274996 Chr.16: 85918745-85918745 on Build GRCh38
rs746720474 Chr.16: 85918378-85918378 on Build GRCh38
rs375583961 Chr.16: 85918382-85918382 on Build GRCh38
rs767169156 Chr.16: 85918666-85918666 on Build GRCh38
rs772597014 Chr.16: 85918560-85918560 on Build GRCh38
rs751949620 Chr.16: 85918690-85918690 on Build GRCh38
rs774613883 Chr.16: 85918453-85918453 on Build GRCh38
rs148746145 Chr.16: 85918497-85918497 on Build GRCh38
rs746036448 Chr.16: 85918642-85918642 on Build GRCh38
rs147892295 Chr.16: 85918383-85918383 on Build GRCh38
rs749529672 Chr.16: 85918724-85918724 on Build GRCh38
rs771384589 Chr.16: 85918576-85918576 on Build GRCh38
rs759196494 Chr.16: 85918767-85918767 on Build GRCh38
rs780940426 Chr.16: 85918469-85918469 on Build GRCh38
rs752611388 Chr.16: 85918522-85918522 on Build GRCh38
rs930540226 Chr.16: 85918351-85918351 on Build GRCh38
rs535845826 Chr.16: 85918712-85918712 on Build GRCh38
rs780424115 Chr.16: 85918722-85918722 on Build GRCh38
rs780579711 Chr.16: 85918784-85918784 on Build GRCh38
rs758259189 Chr.16: 85918432-85918432 on Build GRCh38
rs371253181 Chr.16: 85918725-85918725 on Build GRCh38
rs28368113 Chr.16: 85918547-85918547 on Build GRCh38
rs1042585766 Chr.16: 85918718-85918718 on Build GRCh38
rs755554608 Chr.16: 85918546-85918546 on Build GRCh38
rs200999470 Chr.16: 85918411-85918411 on Build GRCh38
rs747919479 Chr.16: 85918369-85918369 on Build GRCh38
rs752281339 Chr.16: 85918776-85918776 on Build GRCh38
rs368754394 Chr.16: 85918385-85918385 on Build GRCh38
rs372079655 Chr.16: 85918553-85918553 on Build GRCh38
rs900241674 Chr.16: 85918384-85918384 on Build GRCh38
rs770342843 Chr.16: 85918721-85918721 on Build GRCh38
rs774512853 Chr.16: 85918738-85918738 on Build GRCh38
rs201720336 Chr.16: 85918673-85918673 on Build GRCh38
rs200917903 Chr.16: 85918526-85918526 on Build GRCh38
rs774034337 Chr.16: 85918615-85918615 on Build GRCh38
rs138551718 Chr.16: 85918640-85918640 on Build GRCh38
rs774835569 Chr.16: 85918486-85918486 on Build GRCh38
rs750704275 Chr.16: 85918354-85918354 on Build GRCh38
rs745351465 Chr.16: 85918475-85918475 on Build GRCh38
rs745602508 Chr.16: 85918545-85918545 on Build GRCh38
rs757883676 Chr.16: 85918778-85918778 on Build GRCh38
rs202046728 Chr.16: 85918797-85918797 on Build GRCh38
rs764013030 Chr.16: 85918469-85918470 on Build GRCh38
rs778971066 Chr.16: 85918419-85918419 on Build GRCh38
rs747312747 Chr.16: 85918574-85918574 on Build GRCh38
rs907782498 Chr.16: 85918619-85918619 on Build GRCh38
rs144424711 Chr.16: 85918417-85918417 on Build GRCh38
rs376692640 Chr.16: 85918595-85918595 on Build GRCh38
rs770523853 Chr.16: 85918463-85918463 on Build GRCh38
rs757802318 Chr.16: 85918703-85918703 on Build GRCh38
rs147080359 Chr.16: 85918628-85918628 on Build GRCh38
rs945535357 Chr.16: 85918716-85918716 on Build GRCh38
rs375458441 Chr.16: 85918531-85918531 on Build GRCh38
rs546899655 Chr.16: 85918355-85918355 on Build GRCh38
rs141752045 Chr.16: 85918391-85918391 on Build GRCh38
rs752736280 Chr.16: 85918431-85918431 on Build GRCh38
rs772041171 Chr.16: 85918370-85918370 on Build GRCh38
rs146986764 Chr.16: 85918715-85918715 on Build GRCh38
rs778729286 Chr.16: 85918362-85918362 on Build GRCh38
rs760515767 Chr.16: 85918366-85918367 on Build GRCh38
rs778129733 Chr.16: 85918618-85918618 on Build GRCh38
rs764598769 Chr.16: 85918774-85918774 on Build GRCh38
rs757001229 Chr.16: 85918585-85918585 on Build GRCh38
rs768927767 Chr.16: 85918659-85918659 on Build GRCh38
rs996522491 Chr.16: 85918787-85918787 on Build GRCh38
rs765476821 Chr.16: 85918519-85918519 on Build GRCh38
rs776961095 Chr.16: 85918513-85918513 on Build GRCh38
rs773747033 Chr.16: 85918566-85918566 on Build GRCh38
rs760910506 Chr.16: 85918488-85918488 on Build GRCh38
rs371565651 Chr.16: 85918387-85918387 on Build GRCh38
rs763115763 Chr.16: 85918655-85918655 on Build GRCh38
rs370022572 Chr.16: 85918665-85918665 on Build GRCh38
rs775906478 Chr.16: 85918650-85918650 on Build GRCh38
rs768955460 Chr.16: 85918792-85918792 on Build GRCh38
rs759520672 Chr.16: 85918514-85918514 on Build GRCh38
rs776710272 Chr.16: 85918577-85918577 on Build GRCh38
rs778486190 Chr.16: 85918688-85918688 on Build GRCh38
rs766477043 Chr.16: 85918510-85918510 on Build GRCh38
rs974140945 Chr.16: 85918676-85918676 on Build GRCh38
rs57770209 Chr.16: 85918487-85918487 on Build GRCh38
rs867075236 Chr.16: 85918480-85918480 on Build GRCh38
rs769407228 Chr.16: 85918485-85918485 on Build GRCh38
rs28368114 Chr.16: 85918674-85918674 on Build GRCh38
rs756483517 Chr.16: 85918783-85918783 on Build GRCh38

View Full Product Details