Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88698821-88699314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF166
Entrez Gene ID
115992
Gene Name
ring finger protein 166
Gene Aliases
-
UniGene
Hs.513804
Gene Chromosome Location
Chr.16: 88696495-88706421 on Build GRCh38
Transcripts
NM_001171815.1 NM_001171816.1 NM_178841.3 XM_011522845.2 XM_011522846.1 XM_011522847.1 XM_017022910.1 XM_017022913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6874816 c.484C>A p.Q162K Chr.16: 88699027-88699027 on Build GRCh38
6373294 c.515G>T p.S172I Chr.16: 88698996-88698996 on Build GRCh38
3737115 c.506_507delGT p.V170fs*51 Chr.16: 88699004-88699005 on Build GRCh38
7285289 c.473G>A p.R158H Chr.16: 88699038-88699038 on Build GRCh38
3988717 c.498G>T p.V166V Chr.16: 88699013-88699013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777315475 Chr.16: 88699004-88699004 on Build GRCh38
rs148962408 Chr.16: 88699016-88699016 on Build GRCh38
rs750585340 Chr.16: 88699047-88699047 on Build GRCh38
rs765208075 Chr.16: 88699128-88699128 on Build GRCh38
rs914107216 Chr.16: 88699279-88699279 on Build GRCh38
rs776661601 Chr.16: 88699124-88699124 on Build GRCh38
rs756717516 Chr.16: 88699095-88699096 on Build GRCh38
rs151028878 Chr.16: 88699039-88699039 on Build GRCh38
rs556555074 Chr.16: 88698900-88698900 on Build GRCh38
rs945618597 Chr.16: 88699281-88699281 on Build GRCh38
rs372521601 Chr.16: 88698954-88698954 on Build GRCh38
rs759441167 Chr.16: 88699126-88699126 on Build GRCh38
rs372194241 Chr.16: 88698946-88698946 on Build GRCh38
rs531790794 Chr.16: 88699200-88699200 on Build GRCh38
rs1034633384 Chr.16: 88698871-88698871 on Build GRCh38
rs752982647 Chr.16: 88699275-88699275 on Build GRCh38
rs750497512 Chr.16: 88698976-88698976 on Build GRCh38
rs753732709 Chr.16: 88698989-88698989 on Build GRCh38
rs190718096 Chr.16: 88699131-88699131 on Build GRCh38
rs936561296 Chr.16: 88699123-88699123 on Build GRCh38
rs558365512 Chr.16: 88698985-88698985 on Build GRCh38
rs1019753654 Chr.16: 88699223-88699223 on Build GRCh38
rs779935611 Chr.16: 88698932-88698932 on Build GRCh38
rs747659559 Chr.16: 88698948-88698948 on Build GRCh38
rs1026354539 Chr.16: 88699220-88699220 on Build GRCh38
rs756337259 Chr.16: 88699290-88699290 on Build GRCh38
rs763393981 Chr.16: 88699038-88699038 on Build GRCh38
rs560871419 Chr.16: 88699144-88699144 on Build GRCh38
rs527880741 Chr.16: 88699159-88699159 on Build GRCh38
rs970647863 Chr.16: 88699291-88699291 on Build GRCh38
rs768411827 Chr.16: 88698945-88698945 on Build GRCh38
rs969078850 Chr.16: 88698844-88698844 on Build GRCh38
rs777399729 Chr.16: 88699092-88699092 on Build GRCh38
rs911313941 Chr.16: 88699215-88699215 on Build GRCh38
rs555440381 Chr.16: 88699216-88699217 on Build GRCh38
rs749721608 Chr.16: 88699100-88699100 on Build GRCh38
rs528400560 Chr.16: 88699163-88699163 on Build GRCh38
rs1054812936 Chr.16: 88699145-88699145 on Build GRCh38
rs770292556 Chr.16: 88699096-88699096 on Build GRCh38
rs574110532 Chr.16: 88699130-88699130 on Build GRCh38
rs1032309001 Chr.16: 88699268-88699268 on Build GRCh38
rs758107212 Chr.16: 88699091-88699091 on Build GRCh38
rs988059228 Chr.16: 88699289-88699289 on Build GRCh38
rs766321027 Chr.16: 88699118-88699118 on Build GRCh38
rs898835126 Chr.16: 88699184-88699184 on Build GRCh38
rs950627958 Chr.16: 88699213-88699213 on Build GRCh38
rs1026130917 Chr.16: 88699181-88699181 on Build GRCh38
rs370524434 Chr.16: 88698975-88698975 on Build GRCh38
rs368459374 Chr.16: 88698939-88698939 on Build GRCh38
rs764385570 Chr.16: 88698963-88698963 on Build GRCh38
rs140937282 Chr.16: 88699041-88699041 on Build GRCh38
rs772900256 Chr.16: 88698973-88698973 on Build GRCh38
rs780637828 Chr.16: 88699099-88699099 on Build GRCh38
rs576910734 Chr.16: 88699025-88699025 on Build GRCh38
rs778639362 Chr.16: 88698991-88698991 on Build GRCh38
rs550317894 Chr.16: 88699222-88699222 on Build GRCh38
rs185661741 Chr.16: 88698974-88698974 on Build GRCh38
rs34809978 Chr.16: 88699089-88699089 on Build GRCh38
rs1051149011 Chr.16: 88699101-88699101 on Build GRCh38
rs751677312 Chr.16: 88698965-88698965 on Build GRCh38
rs7201334 Chr.16: 88698891-88698891 on Build GRCh38
rs747572101 Chr.16: 88698882-88698882 on Build GRCh38
rs538236038 Chr.16: 88698893-88698893 on Build GRCh38
rs778279236 Chr.16: 88699102-88699103 on Build GRCh38
rs376509894 Chr.16: 88699052-88699052 on Build GRCh38
rs964071573 Chr.16: 88699234-88699234 on Build GRCh38
rs532751219 Chr.16: 88698861-88698861 on Build GRCh38
rs200350722 Chr.16: 88699044-88699044 on Build GRCh38
rs912554676 Chr.16: 88698905-88698905 on Build GRCh38
rs763226144 Chr.16: 88698961-88698961 on Build GRCh38
rs951429341 Chr.16: 88698899-88698899 on Build GRCh38
rs762251826 Chr.16: 88699107-88699107 on Build GRCh38
rs570974751 Chr.16: 88698892-88698892 on Build GRCh38
rs747748900 Chr.16: 88698992-88698992 on Build GRCh38
rs138161076 Chr.16: 88699061-88699061 on Build GRCh38
rs760077454 Chr.16: 88698953-88698953 on Build GRCh38
rs375514729 Chr.16: 88698950-88698950 on Build GRCh38
rs771683988 Chr.16: 88699001-88699001 on Build GRCh38
rs745748335 Chr.16: 88698925-88698925 on Build GRCh38
rs756002055 Chr.16: 88698929-88698929 on Build GRCh38
rs1054048528 Chr.16: 88698896-88698896 on Build GRCh38
rs564536417 Chr.16: 88699167-88699167 on Build GRCh38
rs375210021 Chr.16: 88699135-88699135 on Build GRCh38
rs760820740 Chr.16: 88699117-88699117 on Build GRCh38
rs887258044 Chr.16: 88699221-88699221 on Build GRCh38
rs904916982 Chr.16: 88699157-88699157 on Build GRCh38
rs1040266641 Chr.16: 88698890-88698890 on Build GRCh38
rs912617537 Chr.16: 88698885-88698885 on Build GRCh38
rs373775236 Chr.16: 88698986-88698986 on Build GRCh38
rs140521926 Chr.16: 88699042-88699042 on Build GRCh38
rs942741925 Chr.16: 88698928-88698928 on Build GRCh38
rs752467880 Chr.16: 88699090-88699090 on Build GRCh38
rs773500233 Chr.16: 88699116-88699116 on Build GRCh38
rs774484513 Chr.16: 88699251-88699252 on Build GRCh38
rs374108814 Chr.16: 88699103-88699103 on Build GRCh38
rs368291481 Chr.16: 88699115-88699115 on Build GRCh38
rs944163541 Chr.16: 88698887-88698887 on Build GRCh38
rs964000003 Chr.16: 88699102-88699102 on Build GRCh38
rs115969046 Chr.16: 88698955-88698955 on Build GRCh38
rs963192957 Chr.16: 88698877-88698877 on Build GRCh38
rs767590252 Chr.16: 88699253-88699253 on Build GRCh38
rs1033929488 Chr.16: 88698968-88698968 on Build GRCh38
rs141002164 Chr.16: 88699073-88699073 on Build GRCh38
rs764043009 Chr.16: 88698932-88698932 on Build GRCh38
rs113652548 Chr.16: 88699162-88699162 on Build GRCh38
rs192694044 Chr.16: 88698949-88698949 on Build GRCh38
rs145516855 Chr.16: 88699060-88699060 on Build GRCh38
rs746529321 Chr.16: 88699094-88699094 on Build GRCh38
rs753731506 Chr.16: 88698936-88698937 on Build GRCh38
rs975874836 Chr.16: 88699250-88699250 on Build GRCh38
rs958312321 Chr.16: 88699274-88699274 on Build GRCh38
rs377640480 Chr.16: 88698990-88698990 on Build GRCh38
rs995766881 Chr.16: 88699187-88699187 on Build GRCh38

View Full Product Details