Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88715330-88715838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTU2
Entrez Gene ID
348180
Gene Name
cytosolic thiouridylase subunit 2
Gene Aliases
C16orf84, NCS2, UPF0432
UniGene
Hs.592074
Gene Chromosome Location
Chr.16: 88706463-88715386 on Build GRCh38
Transcripts
NM_001012759.2 NM_001012762.2 NM_001318507.1 NM_001318513.1 XM_017023210.1 NM_001142864.3

Gene Symbol
PIEZO1
Entrez Gene ID
9780
Gene Name
piezo type mechanosensitive ion channel component 1
Gene Aliases
DHS, FAM38A, LMPH3, Mib
UniGene
Hs.377001
Gene Chromosome Location
Chr.16: 88715338-88785220 on Build GRCh38
Transcripts
NM_001012759.2 NM_001012762.2 NM_001318507.1 NM_001318513.1 XM_017023210.1 NM_001142864.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4656224 c.920G>T p.R307L Chr.16: 88715817-88715817 on Build GRCh38
2922431 c.7427G>A p.R2476H Chr.16: 88715744-88715744 on Build GRCh38
5583937 c.7560G>A p.K2520K Chr.16: 88715611-88715611 on Build GRCh38
6290236 c.7380C>T p.S2460S Chr.16: 88715791-88715791 on Build GRCh38
4595689 c.7518C>G p.L2506L Chr.16: 88715653-88715653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192853485 Chr.16: 88715434-88715434 on Build GRCh38
rs767650265 Chr.16: 88715611-88715613 on Build GRCh38
rs752052449 Chr.16: 88715757-88715757 on Build GRCh38
rs900953168 Chr.16: 88715523-88715523 on Build GRCh38
rs1057028185 Chr.16: 88715477-88715477 on Build GRCh38
rs531058403 Chr.16: 88715817-88715817 on Build GRCh38
rs963098971 Chr.16: 88715555-88715555 on Build GRCh38
rs964611699 Chr.16: 88715775-88715775 on Build GRCh38
rs979651269 Chr.16: 88715362-88715365 on Build GRCh38
rs112202523 Chr.16: 88715420-88715420 on Build GRCh38
rs562297402 Chr.16: 88715406-88715406 on Build GRCh38
rs34830861 Chr.16: 88715666-88715666 on Build GRCh38
rs111564291 Chr.16: 88715803-88715803 on Build GRCh38
rs769439251 Chr.16: 88715409-88715409 on Build GRCh38
rs556227621 Chr.16: 88715527-88715527 on Build GRCh38
rs568355773 Chr.16: 88715531-88715531 on Build GRCh38
rs538142617 Chr.16: 88715507-88715507 on Build GRCh38
rs945494220 Chr.16: 88715693-88715693 on Build GRCh38
rs1061228 Chr.16: 88715671-88715671 on Build GRCh38
rs1061238 Chr.16: 88715442-88715442 on Build GRCh38
rs530762101 Chr.16: 88715647-88715647 on Build GRCh38
rs747286972 Chr.16: 88715586-88715586 on Build GRCh38
rs924760381 Chr.16: 88715447-88715447 on Build GRCh38
rs772748505 Chr.16: 88715740-88715740 on Build GRCh38
rs545941884 Chr.16: 88715587-88715587 on Build GRCh38
rs953710841 Chr.16: 88715529-88715529 on Build GRCh38
rs1028089124 Chr.16: 88715603-88715603 on Build GRCh38
rs768686405 Chr.16: 88715720-88715720 on Build GRCh38
rs572190306 Chr.16: 88715363-88715363 on Build GRCh38
rs1000925279 Chr.16: 88715509-88715509 on Build GRCh38
rs973148096 Chr.16: 88715625-88715625 on Build GRCh38
rs762374984 Chr.16: 88715818-88715818 on Build GRCh38
rs768807373 Chr.16: 88715788-88715788 on Build GRCh38
rs587776992 Chr.16: 88715692-88715693 on Build GRCh38
rs376785885 Chr.16: 88715500-88715500 on Build GRCh38
rs941879758 Chr.16: 88715390-88715394 on Build GRCh38
rs764668729 Chr.16: 88715812-88715812 on Build GRCh38
rs897773962 Chr.16: 88715391-88715391 on Build GRCh38
rs1037374997 Chr.16: 88715669-88715669 on Build GRCh38
rs1032488142 Chr.16: 88715426-88715426 on Build GRCh38
rs532748737 Chr.16: 88715808-88715808 on Build GRCh38
rs533232864 Chr.16: 88715633-88715633 on Build GRCh38
rs370579491 Chr.16: 88715508-88715508 on Build GRCh38
rs557783308 Chr.16: 88715590-88715590 on Build GRCh38
rs975061667 Chr.16: 88715777-88715777 on Build GRCh38
rs1023051361 Chr.16: 88715811-88715811 on Build GRCh38
rs758885868 Chr.16: 88715643-88715643 on Build GRCh38
rs552016965 Chr.16: 88715584-88715584 on Build GRCh38
rs774391363 Chr.16: 88715722-88715722 on Build GRCh38
rs993426050 Chr.16: 88715392-88715392 on Build GRCh38
rs938218192 Chr.16: 88715800-88715800 on Build GRCh38
rs956362052 Chr.16: 88715366-88715367 on Build GRCh38
rs202127176 Chr.16: 88715797-88715797 on Build GRCh38
rs200243384 Chr.16: 88715751-88715751 on Build GRCh38
rs780669235 Chr.16: 88715683-88715688 on Build GRCh38
rs1033364623 Chr.16: 88715543-88715543 on Build GRCh38
rs1018815323 Chr.16: 88715414-88715414 on Build GRCh38
rs749413029 Chr.16: 88715786-88715786 on Build GRCh38
rs201746476 Chr.16: 88715700-88715700 on Build GRCh38
rs61745086 Chr.16: 88715642-88715642 on Build GRCh38
rs911827789 Chr.16: 88715570-88715570 on Build GRCh38
rs997942023 Chr.16: 88715526-88715526 on Build GRCh38
rs948748493 Chr.16: 88715572-88715572 on Build GRCh38
rs35736353 Chr.16: 88715676-88715676 on Build GRCh38
rs201104063 Chr.16: 88715683-88715683 on Build GRCh38
rs747385238 Chr.16: 88715802-88715802 on Build GRCh38
rs565924717 Chr.16: 88715744-88715744 on Build GRCh38
rs1048278841 Chr.16: 88715574-88715574 on Build GRCh38
rs780322188 Chr.16: 88715784-88715784 on Build GRCh38
rs574227772 Chr.16: 88715785-88715785 on Build GRCh38
rs371391349 Chr.16: 88715790-88715790 on Build GRCh38
rs1026737466 Chr.16: 88715402-88715402 on Build GRCh38
rs914014470 Chr.16: 88715690-88715690 on Build GRCh38
rs774268616 Chr.16: 88715618-88715618 on Build GRCh38
rs980874158 Chr.16: 88715539-88715539 on Build GRCh38
rs543807946 Chr.16: 88715399-88715399 on Build GRCh38
rs374964860 Chr.16: 88715709-88715709 on Build GRCh38
rs587776988 Chr.16: 88715804-88715804 on Build GRCh38
rs922550263 Chr.16: 88715771-88715771 on Build GRCh38
rs977653163 Chr.16: 88715542-88715542 on Build GRCh38
rs763489080 Chr.16: 88715755-88715755 on Build GRCh38
rs987739634 Chr.16: 88715369-88715370 on Build GRCh38
rs764435951 Chr.16: 88715674-88715674 on Build GRCh38
rs922432809 Chr.16: 88715670-88715670 on Build GRCh38
rs943315603 Chr.16: 88715573-88715573 on Build GRCh38
rs750927939 Chr.16: 88715756-88715756 on Build GRCh38
rs973203620 Chr.16: 88715461-88715461 on Build GRCh38
rs576096834 Chr.16: 88715778-88715778 on Build GRCh38
rs113882095 Chr.16: 88715435-88715435 on Build GRCh38
rs570744198 Chr.16: 88715613-88715613 on Build GRCh38
rs182627221 Chr.16: 88715534-88715534 on Build GRCh38
rs547008930 Chr.16: 88715619-88715619 on Build GRCh38
rs35514587 Chr.16: 88715513-88715514 on Build GRCh38
rs760119794 Chr.16: 88715626-88715626 on Build GRCh38
rs535588603 Chr.16: 88715504-88715504 on Build GRCh38
rs926467594 Chr.16: 88715387-88715388 on Build GRCh38
rs547972506 Chr.16: 88715428-88715428 on Build GRCh38
rs755226463 Chr.16: 88715693-88715694 on Build GRCh38
rs72190263 Chr.16: 88715361-88715362 on Build GRCh38
rs1042176965 Chr.16: 88715443-88715443 on Build GRCh38
rs761535896 Chr.16: 88715729-88715729 on Build GRCh38
rs753320868 Chr.16: 88715752-88715752 on Build GRCh38
rs780275226 Chr.16: 88715702-88715702 on Build GRCh38
rs1032435811 Chr.16: 88715510-88715510 on Build GRCh38
rs992857659 Chr.16: 88715634-88715634 on Build GRCh38
rs906703800 Chr.16: 88715712-88715712 on Build GRCh38
rs902194588 Chr.16: 88715716-88715716 on Build GRCh38
rs532009479 Chr.16: 88715393-88715393 on Build GRCh38
rs992893593 Chr.16: 88715568-88715568 on Build GRCh38
rs201019404 Chr.16: 88715806-88715806 on Build GRCh38
rs1004070979 Chr.16: 88715514-88715514 on Build GRCh38
rs559557496 Chr.16: 88715621-88715621 on Build GRCh38
rs919085550 Chr.16: 88715566-88715566 on Build GRCh38
rs889573368 Chr.16: 88715679-88715679 on Build GRCh38
rs766184064 Chr.16: 88715556-88715556 on Build GRCh38
rs747793811 Chr.16: 88715408-88715408 on Build GRCh38
rs184706181 Chr.16: 88715783-88715783 on Build GRCh38
rs976482599 Chr.16: 88715418-88715418 on Build GRCh38
rs749288233 Chr.16: 88715708-88715708 on Build GRCh38
rs969290603 Chr.16: 88715541-88715541 on Build GRCh38
rs115423293 Chr.16: 88715641-88715641 on Build GRCh38
rs73262683 Chr.16: 88715610-88715610 on Build GRCh38
rs948617984 Chr.16: 88715491-88715491 on Build GRCh38
rs1019161006 Chr.16: 88715767-88715767 on Build GRCh38
rs960494512 Chr.16: 88715629-88715629 on Build GRCh38
rs1061247 Chr.16: 88715383-88715383 on Build GRCh38
rs545779147 Chr.16: 88715374-88715374 on Build GRCh38
rs35544968 Chr.16: 88715809-88715809 on Build GRCh38
rs113700874 Chr.16: 88715377-88715377 on Build GRCh38
rs965396436 Chr.16: 88715415-88715415 on Build GRCh38
rs751834996 Chr.16: 88715649-88715649 on Build GRCh38
rs142386387 Chr.16: 88715601-88715601 on Build GRCh38
rs889436765 Chr.16: 88715604-88715604 on Build GRCh38
rs1015755916 Chr.16: 88715520-88715520 on Build GRCh38
rs1010667257 Chr.16: 88715699-88715699 on Build GRCh38
rs988356689 Chr.16: 88715430-88715430 on Build GRCh38
rs1037603751 Chr.16: 88715391-88715397 on Build GRCh38
rs867324252 Chr.16: 88715794-88715794 on Build GRCh38
rs748599881 Chr.16: 88715571-88715571 on Build GRCh38
rs536609140 Chr.16: 88715782-88715782 on Build GRCh38
rs972662761 Chr.16: 88715763-88715763 on Build GRCh38
rs768652167 Chr.16: 88715624-88715625 on Build GRCh38
rs944061069 Chr.16: 88715479-88715479 on Build GRCh38
rs370827778 Chr.16: 88715400-88715400 on Build GRCh38
rs554022189 Chr.16: 88715569-88715569 on Build GRCh38
rs938675724 Chr.16: 88715472-88715472 on Build GRCh38
rs894485572 Chr.16: 88715482-88715482 on Build GRCh38
rs577860843 Chr.16: 88715799-88715799 on Build GRCh38
rs974697358 Chr.16: 88715656-88715656 on Build GRCh38
rs187723809 Chr.16: 88715591-88715591 on Build GRCh38
rs144035770 Chr.16: 88715745-88715745 on Build GRCh38
rs925564859 Chr.16: 88715362-88715363 on Build GRCh38
rs914719660 Chr.16: 88715437-88715437 on Build GRCh38
rs1045222014 Chr.16: 88715386-88715387 on Build GRCh38
rs547600679 Chr.16: 88715737-88715737 on Build GRCh38
rs529613614 Chr.16: 88715611-88715611 on Build GRCh38
rs1039780514 Chr.16: 88715499-88715499 on Build GRCh38
rs368497431 Chr.16: 88715766-88715766 on Build GRCh38
rs1032366684 Chr.16: 88715725-88715725 on Build GRCh38
rs937824210 Chr.16: 88715478-88715478 on Build GRCh38
rs779314125 Chr.16: 88715563-88715563 on Build GRCh38

View Full Product Details