Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89563230-89563770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL13
Entrez Gene ID
6137
Gene Name
ribosomal protein L13
Gene Aliases
BBC1, D16S444E, D16S44E, L13
UniGene
Hs.410817
Gene Chromosome Location
Chr.16: 89560657-89566829 on Build GRCh38
Transcripts
NM_000977.3 NM_001243130.1 NM_001243131.1 NM_033251.2

SNP Detail

SNP ID Chromosome Location
rs892022570 Chr.16: 89563582-89563587 on Build GRCh38
rs571344480 Chr.16: 89563339-89563339 on Build GRCh38
rs760065687 Chr.16: 89563452-89563452 on Build GRCh38
rs13380730 Chr.16: 89563684-89563684 on Build GRCh38
rs918005967 Chr.16: 89563708-89563708 on Build GRCh38
rs1053782245 Chr.16: 89563574-89563579 on Build GRCh38
rs1038254540 Chr.16: 89563739-89563739 on Build GRCh38
rs925260296 Chr.16: 89563271-89563271 on Build GRCh38
rs967014321 Chr.16: 89563527-89563527 on Build GRCh38
rs532506593 Chr.16: 89563584-89563587 on Build GRCh38
rs1008378228 Chr.16: 89563647-89563647 on Build GRCh38
rs894345861 Chr.16: 89563353-89563353 on Build GRCh38
rs72037989 Chr.16: 89563574-89563575 on Build GRCh38
rs917967517 Chr.16: 89563546-89563546 on Build GRCh38
rs925015909 Chr.16: 89563695-89563695 on Build GRCh38
rs528376805 Chr.16: 89563335-89563335 on Build GRCh38
rs949521406 Chr.16: 89563711-89563711 on Build GRCh38
rs111439133 Chr.16: 89563706-89563706 on Build GRCh38
rs911091590 Chr.16: 89563651-89563651 on Build GRCh38
rs970101332 Chr.16: 89563387-89563387 on Build GRCh38
rs1050921342 Chr.16: 89563473-89563473 on Build GRCh38
rs558559345 Chr.16: 89563657-89563657 on Build GRCh38
rs201427329 Chr.16: 89563339-89563341 on Build GRCh38
rs955732185 Chr.16: 89563484-89563484 on Build GRCh38
rs937983653 Chr.16: 89563536-89563536 on Build GRCh38
rs375445545 Chr.16: 89563585-89563588 on Build GRCh38
rs955123140 Chr.16: 89563646-89563646 on Build GRCh38
rs3211567 Chr.16: 89563328-89563328 on Build GRCh38
rs57775530 Chr.16: 89563520-89563520 on Build GRCh38
rs560715880 Chr.16: 89563697-89563697 on Build GRCh38
rs779597918 Chr.16: 89563655-89563655 on Build GRCh38
rs527712224 Chr.16: 89563585-89563586 on Build GRCh38
rs3211569 Chr.16: 89563411-89563411 on Build GRCh38
rs756200283 Chr.16: 89563518-89563518 on Build GRCh38
rs563363785 Chr.16: 89563338-89563338 on Build GRCh38
rs796088763 Chr.16: 89563573-89563574 on Build GRCh38
rs567700415 Chr.16: 89563534-89563534 on Build GRCh38
rs577796870 Chr.16: 89563588-89563588 on Build GRCh38
rs773366804 Chr.16: 89563286-89563286 on Build GRCh38
rs757871429 Chr.16: 89563643-89563643 on Build GRCh38
rs577696405 Chr.16: 89563332-89563333 on Build GRCh38
rs1001653395 Chr.16: 89563589-89563589 on Build GRCh38
rs950369741 Chr.16: 89563729-89563729 on Build GRCh38
rs887005541 Chr.16: 89563405-89563405 on Build GRCh38
rs1016580711 Chr.16: 89563428-89563428 on Build GRCh38
rs548892314 Chr.16: 89563395-89563395 on Build GRCh38
rs188754482 Chr.16: 89563260-89563260 on Build GRCh38
rs967277025 Chr.16: 89563659-89563659 on Build GRCh38
rs556602259 Chr.16: 89563629-89563629 on Build GRCh38
rs1027994660 Chr.16: 89563454-89563454 on Build GRCh38
rs139037193 Chr.16: 89563489-89563489 on Build GRCh38
rs949940802 Chr.16: 89563587-89563587 on Build GRCh38
rs942528148 Chr.16: 89563661-89563661 on Build GRCh38
rs151033862 Chr.16: 89563280-89563280 on Build GRCh38
rs570180216 Chr.16: 89563442-89563442 on Build GRCh38
rs1043400041 Chr.16: 89563303-89563303 on Build GRCh38
rs906039713 Chr.16: 89563737-89563737 on Build GRCh38
rs757025175 Chr.16: 89563616-89563616 on Build GRCh38
rs1024506966 Chr.16: 89563588-89563591 on Build GRCh38
rs879775934 Chr.16: 89563680-89563680 on Build GRCh38
rs886924513 Chr.16: 89563600-89563600 on Build GRCh38
rs764992234 Chr.16: 89563630-89563630 on Build GRCh38
rs999164225 Chr.16: 89563574-89563578 on Build GRCh38
rs3211568 Chr.16: 89563372-89563372 on Build GRCh38
rs962353289 Chr.16: 89563435-89563435 on Build GRCh38
rs145764650 Chr.16: 89563524-89563524 on Build GRCh38
rs540139256 Chr.16: 89563731-89563731 on Build GRCh38
rs1016997467 Chr.16: 89563626-89563626 on Build GRCh38
rs889511984 Chr.16: 89563293-89563293 on Build GRCh38
rs1046061846 Chr.16: 89563382-89563382 on Build GRCh38
rs538872205 Chr.16: 89563601-89563601 on Build GRCh38
rs908408535 Chr.16: 89563720-89563720 on Build GRCh38
rs979661887 Chr.16: 89563530-89563530 on Build GRCh38
rs939823931 Chr.16: 89563733-89563733 on Build GRCh38
rs144981608 Chr.16: 89563309-89563309 on Build GRCh38
rs1031913401 Chr.16: 89563350-89563350 on Build GRCh38
rs546920024 Chr.16: 89563336-89563336 on Build GRCh38
rs918514479 Chr.16: 89563636-89563636 on Build GRCh38
rs1014384778 Chr.16: 89563367-89563367 on Build GRCh38
rs11546506 Chr.16: 89563269-89563269 on Build GRCh38
rs983941914 Chr.16: 89563717-89563717 on Build GRCh38
rs896126535 Chr.16: 89563713-89563713 on Build GRCh38
rs561295351 Chr.16: 89563425-89563428 on Build GRCh38
rs750054727 Chr.16: 89563635-89563635 on Build GRCh38
rs868384233 Chr.16: 89563479-89563479 on Build GRCh38
rs1028048645 Chr.16: 89563645-89563645 on Build GRCh38
rs553665657 Chr.16: 89563542-89563542 on Build GRCh38
rs371003318 Chr.16: 89563639-89563639 on Build GRCh38
rs1045600197 Chr.16: 89563598-89563598 on Build GRCh38
rs996586877 Chr.16: 89563438-89563438 on Build GRCh38
rs372710293 Chr.16: 89563300-89563300 on Build GRCh38
rs534999270 Chr.16: 89563540-89563540 on Build GRCh38

View Full Product Details