Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89644610-89645116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP1A
Entrez Gene ID
5119
Gene Name
charged multivesicular body protein 1A
Gene Aliases
CHMP1, PCH8, PCOLN3, PRSM1, VPS46-1, VPS46A
UniGene
Hs.589427
Gene Chromosome Location
Chr.16: 89644431-89657785 on Build GRCh38
Transcripts
NM_001083314.3 NM_002768.4 NR_046418.2

SNP Detail

SNP ID Chromosome Location
rs528954844 Chr.16: 89644756-89644756 on Build GRCh38
rs768142054 Chr.16: 89644717-89644717 on Build GRCh38
rs539830727 Chr.16: 89645079-89645079 on Build GRCh38
rs926360609 Chr.16: 89644713-89644713 on Build GRCh38
rs1059960 Chr.16: 89644976-89644976 on Build GRCh38
rs543955146 Chr.16: 89644703-89644703 on Build GRCh38
rs1059958 Chr.16: 89645083-89645083 on Build GRCh38
rs956622185 Chr.16: 89644673-89644673 on Build GRCh38
rs973102892 Chr.16: 89644838-89644838 on Build GRCh38
rs750136723 Chr.16: 89644773-89644773 on Build GRCh38
rs1000871047 Chr.16: 89644661-89644661 on Build GRCh38
rs114424524 Chr.16: 89644851-89644851 on Build GRCh38
rs1059995 Chr.16: 89644651-89644651 on Build GRCh38
rs3200812 Chr.16: 89645013-89645013 on Build GRCh38
rs551010574 Chr.16: 89644822-89644822 on Build GRCh38
rs77979076 Chr.16: 89644829-89644829 on Build GRCh38
rs945368969 Chr.16: 89645080-89645080 on Build GRCh38
rs1021369148 Chr.16: 89644986-89644986 on Build GRCh38
rs146960250 Chr.16: 89644761-89644761 on Build GRCh38
rs6860 Chr.16: 89644712-89644712 on Build GRCh38
rs963056808 Chr.16: 89644714-89644714 on Build GRCh38
rs980298683 Chr.16: 89645031-89645031 on Build GRCh38
rs1007912443 Chr.16: 89644636-89644636 on Build GRCh38
rs11556763 Chr.16: 89644845-89644845 on Build GRCh38
rs970956704 Chr.16: 89644685-89644685 on Build GRCh38
rs781613304 Chr.16: 89644864-89644864 on Build GRCh38
rs748705531 Chr.16: 89644921-89644921 on Build GRCh38
rs141704543 Chr.16: 89644668-89644668 on Build GRCh38
rs973049107 Chr.16: 89644716-89644716 on Build GRCh38
rs930245694 Chr.16: 89644743-89644743 on Build GRCh38
rs989867725 Chr.16: 89644667-89644667 on Build GRCh38
rs567374319 Chr.16: 89644860-89644860 on Build GRCh38
rs367714771 Chr.16: 89644836-89644836 on Build GRCh38
rs899926974 Chr.16: 89644791-89644791 on Build GRCh38
rs892506372 Chr.16: 89645019-89645019 on Build GRCh38
rs554283279 Chr.16: 89644905-89644905 on Build GRCh38
rs59496184 Chr.16: 89644938-89644938 on Build GRCh38
rs75779036 Chr.16: 89644677-89644677 on Build GRCh38
rs997399125 Chr.16: 89644915-89644915 on Build GRCh38
rs1048446070 Chr.16: 89644827-89644827 on Build GRCh38
rs867395369 Chr.16: 89645093-89645093 on Build GRCh38
rs572453661 Chr.16: 89644650-89644650 on Build GRCh38
rs533524083 Chr.16: 89644641-89644641 on Build GRCh38
rs7405433 Chr.16: 89644820-89644820 on Build GRCh38
rs936763583 Chr.16: 89644949-89644949 on Build GRCh38
rs1034809436 Chr.16: 89644797-89644816 on Build GRCh38
rs983088836 Chr.16: 89644848-89644848 on Build GRCh38
rs955780759 Chr.16: 89644657-89644657 on Build GRCh38
rs534845331 Chr.16: 89644861-89644861 on Build GRCh38
rs186815550 Chr.16: 89644934-89644934 on Build GRCh38
rs867619706 Chr.16: 89644740-89644740 on Build GRCh38
rs532746940 Chr.16: 89644792-89644792 on Build GRCh38
rs1064732 Chr.16: 89645081-89645081 on Build GRCh38
rs547104404 Chr.16: 89644760-89644760 on Build GRCh38
rs572617326 Chr.16: 89644925-89644925 on Build GRCh38
rs867060947 Chr.16: 89645032-89645032 on Build GRCh38
rs1039834332 Chr.16: 89644775-89644775 on Build GRCh38
rs918559738 Chr.16: 89644818-89644818 on Build GRCh38
rs1047742452 Chr.16: 89644745-89644745 on Build GRCh38
rs1033482619 Chr.16: 89644837-89644837 on Build GRCh38
rs1008567917 Chr.16: 89644783-89644783 on Build GRCh38
rs997181196 Chr.16: 89644649-89644649 on Build GRCh38
rs980798752 Chr.16: 89644698-89644698 on Build GRCh38
rs564764880 Chr.16: 89644676-89644676 on Build GRCh38
rs1059961 Chr.16: 89644974-89644974 on Build GRCh38
rs554183007 Chr.16: 89644640-89644640 on Build GRCh38
rs1053806686 Chr.16: 89644809-89644809 on Build GRCh38
rs955665688 Chr.16: 89644975-89644975 on Build GRCh38
rs963109219 Chr.16: 89644837-89644837 on Build GRCh38
rs970672781 Chr.16: 89645022-89645022 on Build GRCh38
rs542348478 Chr.16: 89644630-89644630 on Build GRCh38

View Full Product Details