Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1534007-1534509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PITPNA
Entrez Gene ID
5306
Gene Name
phosphatidylinositol transfer protein alpha
Gene Aliases
HEL-S-36, PI-TPalpha, PITPN, VIB1A
UniGene
Hs.429819
Gene Chromosome Location
Chr.17: 1517989-1562816 on Build GRCh38
Transcripts
NM_006224.3 XM_011523939.1 XM_011523940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6746793 c.749C>T p.T250M Chr.17: 1534118-1534118 on Build GRCh38
3680082 c.737T>G p.M246R Chr.17: 1534130-1534130 on Build GRCh38
4341572 c.724G>A p.D242N Chr.17: 1534143-1534143 on Build GRCh38
4863899 c.700A>G p.K234E Chr.17: 1534167-1534167 on Build GRCh38
3680083 c.700A>T p.K234* Chr.17: 1534167-1534167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892398342 Chr.17: 1534387-1534387 on Build GRCh38
rs776841050 Chr.17: 1534058-1534061 on Build GRCh38
rs377298878 Chr.17: 1534072-1534072 on Build GRCh38
rs75018043 Chr.17: 1534397-1534397 on Build GRCh38
rs1000487794 Chr.17: 1534352-1534352 on Build GRCh38
rs980600347 Chr.17: 1534401-1534401 on Build GRCh38
rs974051498 Chr.17: 1534428-1534428 on Build GRCh38
rs748934198 Chr.17: 1534251-1534251 on Build GRCh38
rs756236948 Chr.17: 1534066-1534066 on Build GRCh38
rs572226347 Chr.17: 1534225-1534225 on Build GRCh38
rs930282275 Chr.17: 1534440-1534440 on Build GRCh38
rs1041508421 Chr.17: 1534342-1534342 on Build GRCh38
rs768809943 Chr.17: 1534050-1534051 on Build GRCh38
rs879173749 Chr.17: 1534099-1534099 on Build GRCh38
rs764210140 Chr.17: 1534183-1534183 on Build GRCh38
rs982932949 Chr.17: 1534224-1534224 on Build GRCh38
rs1007042480 Chr.17: 1534029-1534029 on Build GRCh38
rs756901696 Chr.17: 1534268-1534269 on Build GRCh38
rs371204666 Chr.17: 1534085-1534085 on Build GRCh38
rs758917340 Chr.17: 1534234-1534234 on Build GRCh38
rs1050171475 Chr.17: 1534292-1534292 on Build GRCh38
rs1017980027 Chr.17: 1534029-1534030 on Build GRCh38
rs767272403 Chr.17: 1534156-1534156 on Build GRCh38
rs2270229 Chr.17: 1534040-1534040 on Build GRCh38
rs140577171 Chr.17: 1534263-1534263 on Build GRCh38
rs761554915 Chr.17: 1534272-1534273 on Build GRCh38
rs746415782 Chr.17: 1534105-1534105 on Build GRCh38
rs770474302 Chr.17: 1534138-1534138 on Build GRCh38
rs574892612 Chr.17: 1534341-1534341 on Build GRCh38
rs780706551 Chr.17: 1534236-1534236 on Build GRCh38
rs181041763 Chr.17: 1534337-1534337 on Build GRCh38
rs903104572 Chr.17: 1534351-1534351 on Build GRCh38
rs777266858 Chr.17: 1534248-1534248 on Build GRCh38
rs769260246 Chr.17: 1534126-1534126 on Build GRCh38
rs1000815201 Chr.17: 1534046-1534046 on Build GRCh38
rs772734963 Chr.17: 1534114-1534114 on Build GRCh38
rs962729002 Chr.17: 1534420-1534420 on Build GRCh38
rs375499014 Chr.17: 1534132-1534132 on Build GRCh38
rs1034412723 Chr.17: 1534412-1534412 on Build GRCh38
rs568501564 Chr.17: 1534232-1534232 on Build GRCh38
rs775556237 Chr.17: 1534269-1534269 on Build GRCh38
rs917338059 Chr.17: 1534161-1534161 on Build GRCh38
rs978805183 Chr.17: 1534117-1534117 on Build GRCh38
rs186465010 Chr.17: 1534398-1534398 on Build GRCh38
rs767459668 Chr.17: 1534148-1534148 on Build GRCh38
rs2230464 Chr.17: 1534141-1534141 on Build GRCh38
rs753858906 Chr.17: 1534077-1534077 on Build GRCh38
rs754139606 Chr.17: 1534184-1534184 on Build GRCh38
rs867207548 Chr.17: 1534430-1534430 on Build GRCh38
rs372520299 Chr.17: 1534166-1534166 on Build GRCh38
rs1032921923 Chr.17: 1534390-1534390 on Build GRCh38
rs183764382 Chr.17: 1534278-1534278 on Build GRCh38
rs200617729 Chr.17: 1534219-1534219 on Build GRCh38
rs374194242 Chr.17: 1534058-1534058 on Build GRCh38
rs755425650 Chr.17: 1534242-1534242 on Build GRCh38
rs757417607 Chr.17: 1534082-1534082 on Build GRCh38
rs531765844 Chr.17: 1534171-1534171 on Build GRCh38
rs891286446 Chr.17: 1534277-1534277 on Build GRCh38
rs757383601 Chr.17: 1534213-1534213 on Build GRCh38
rs2230464 Chr.17: 1534141-1534141 on Build GRCh38
rs113883904 Chr.17: 1534098-1534098 on Build GRCh38
rs971542612 Chr.17: 1534086-1534086 on Build GRCh38
rs967672668 Chr.17: 1534087-1534087 on Build GRCh38
rs537488853 Chr.17: 1534362-1534362 on Build GRCh38
rs188194986 Chr.17: 1534289-1534289 on Build GRCh38
rs369744436 Chr.17: 1534241-1534241 on Build GRCh38
rs745391605 Chr.17: 1534267-1534267 on Build GRCh38
rs371243344 Chr.17: 1534057-1534057 on Build GRCh38
rs191690916 Chr.17: 1534441-1534441 on Build GRCh38
rs890184641 Chr.17: 1534329-1534329 on Build GRCh38
rs539729488 Chr.17: 1534325-1534325 on Build GRCh38
rs367888223 Chr.17: 1534144-1534144 on Build GRCh38
rs1804173 Chr.17: 1534122-1534122 on Build GRCh38
rs775085618 Chr.17: 1534165-1534165 on Build GRCh38
rs982965526 Chr.17: 1534462-1534462 on Build GRCh38
rs780518060 Chr.17: 1534120-1534120 on Build GRCh38
rs200738801 Chr.17: 1534133-1534133 on Build GRCh38
rs920219681 Chr.17: 1534274-1534274 on Build GRCh38
rs1024426945 Chr.17: 1534434-1534434 on Build GRCh38

View Full Product Details