Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3811401-3811947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCBP3
Entrez Gene ID
55421
Gene Name
nuclear cap binding subunit 3
Gene Aliases
C17orf85, ELG, HSA277841
UniGene
Hs.120963
Gene Chromosome Location
Chr.17: 3806748-3846251 on Build GRCh38
Transcripts
NM_001114118.2 XM_005256717.2 XM_006721556.2 XM_011523954.2 XM_011523955.2 XM_017024826.1 XM_017024827.1 XM_017024828.1 XM_017024829.1

SNP Detail

SNP ID Chromosome Location
rs906841923 Chr.17: 3811650-3811650 on Build GRCh38
rs372759117 Chr.17: 3811437-3811437 on Build GRCh38
rs535663658 Chr.17: 3811895-3811895 on Build GRCh38
rs113222882 Chr.17: 3811707-3811707 on Build GRCh38
rs529260199 Chr.17: 3811763-3811763 on Build GRCh38
rs1042302844 Chr.17: 3811911-3811911 on Build GRCh38
rs541675890 Chr.17: 3811839-3811839 on Build GRCh38
rs973934537 Chr.17: 3811484-3811484 on Build GRCh38
rs149874757 Chr.17: 3811724-3811724 on Build GRCh38
rs971630747 Chr.17: 3811772-3811772 on Build GRCh38
rs908578784 Chr.17: 3811816-3811816 on Build GRCh38
rs924608622 Chr.17: 3811749-3811749 on Build GRCh38
rs17222977 Chr.17: 3811745-3811745 on Build GRCh38
rs996669828 Chr.17: 3811618-3811618 on Build GRCh38
rs926146851 Chr.17: 3811520-3811520 on Build GRCh38
rs35255448 Chr.17: 3811901-3811902 on Build GRCh38
rs921363819 Chr.17: 3811852-3811852 on Build GRCh38
rs556096313 Chr.17: 3811636-3811636 on Build GRCh38
rs569680890 Chr.17: 3811481-3811481 on Build GRCh38
rs890929572 Chr.17: 3811901-3811901 on Build GRCh38
rs941478682 Chr.17: 3811574-3811574 on Build GRCh38
rs1050888322 Chr.17: 3811655-3811655 on Build GRCh38
rs776070160 Chr.17: 3811702-3811702 on Build GRCh38
rs1047508948 Chr.17: 3811565-3811565 on Build GRCh38
rs938343607 Chr.17: 3811922-3811922 on Build GRCh38
rs934828820 Chr.17: 3811592-3811592 on Build GRCh38
rs985390296 Chr.17: 3811540-3811540 on Build GRCh38
rs948943849 Chr.17: 3811634-3811634 on Build GRCh38
rs1044668260 Chr.17: 3811649-3811649 on Build GRCh38
rs941353295 Chr.17: 3811831-3811831 on Build GRCh38
rs568233154 Chr.17: 3811555-3811555 on Build GRCh38
rs749449450 Chr.17: 3811429-3811429 on Build GRCh38
rs983049339 Chr.17: 3811793-3811793 on Build GRCh38
rs910522661 Chr.17: 3811547-3811547 on Build GRCh38
rs932699014 Chr.17: 3811879-3811879 on Build GRCh38
rs146469598 Chr.17: 3811743-3811743 on Build GRCh38
rs1026792857 Chr.17: 3811706-3811706 on Build GRCh38
rs937505418 Chr.17: 3811524-3811524 on Build GRCh38
rs991629762 Chr.17: 3811744-3811744 on Build GRCh38
rs16953494 Chr.17: 3811593-3811593 on Build GRCh38
rs771372218 Chr.17: 3811552-3811552 on Build GRCh38
rs527474407 Chr.17: 3811875-3811875 on Build GRCh38
rs116568439 Chr.17: 3811666-3811666 on Build GRCh38
rs188996878 Chr.17: 3811718-3811718 on Build GRCh38
rs1009933531 Chr.17: 3811907-3811907 on Build GRCh38
rs746197678 Chr.17: 3811633-3811633 on Build GRCh38
rs772722462 Chr.17: 3811694-3811694 on Build GRCh38
rs2288685 Chr.17: 3811840-3811840 on Build GRCh38
rs1026824033 Chr.17: 3811511-3811511 on Build GRCh38
rs547313727 Chr.17: 3811892-3811892 on Build GRCh38
rs1015751011 Chr.17: 3811669-3811669 on Build GRCh38
rs908508384 Chr.17: 3811570-3811570 on Build GRCh38
rs539406435 Chr.17: 3811428-3811428 on Build GRCh38
rs1009446670 Chr.17: 3811691-3811691 on Build GRCh38
rs899691619 Chr.17: 3811613-3811613 on Build GRCh38
rs547999936 Chr.17: 3811774-3811774 on Build GRCh38
rs896871099 Chr.17: 3811671-3811671 on Build GRCh38
rs979170214 Chr.17: 3811514-3811514 on Build GRCh38
rs761339703 Chr.17: 3811917-3811917 on Build GRCh38
rs950280909 Chr.17: 3811560-3811560 on Build GRCh38
rs995265806 Chr.17: 3811673-3811673 on Build GRCh38
rs533443880 Chr.17: 3811851-3811851 on Build GRCh38

View Full Product Details