Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4718274-4718773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRB2
Entrez Gene ID
409
Gene Name
arrestin beta 2
Gene Aliases
ARB2, ARR2, BARR2
UniGene
Hs.435811
Gene Chromosome Location
Chr.17: 4710494-4721500 on Build GRCh38
Transcripts
NM_001257328.1 NM_001257329.1 NM_001257330.1 NM_001257331.1 NM_004313.3 NM_199004.1 NR_047516.1 XM_006721520.1 XM_011523858.2 XM_017024645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464943 c.740C>T p.T247I Chr.17: 4718645-4718645 on Build GRCh38
7341126 c.779A>T p.D260V Chr.17: 4718684-4718684 on Build GRCh38
6081061 c.708G>A p.V236V Chr.17: 4718613-4718613 on Build GRCh38
385135 c.681G>C p.K227N Chr.17: 4718320-4718320 on Build GRCh38
4663515 c.685G>A p.V229I Chr.17: 4718324-4718324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746977488 Chr.17: 4718496-4718496 on Build GRCh38
rs753429873 Chr.17: 4718719-4718719 on Build GRCh38
rs572262352 Chr.17: 4718444-4718444 on Build GRCh38
rs780961076 Chr.17: 4718300-4718300 on Build GRCh38
rs772243126 Chr.17: 4718647-4718647 on Build GRCh38
rs754430222 Chr.17: 4718299-4718299 on Build GRCh38
rs746331162 Chr.17: 4718646-4718646 on Build GRCh38
rs901933755 Chr.17: 4718652-4718652 on Build GRCh38
rs773144124 Chr.17: 4718568-4718568 on Build GRCh38
rs768642598 Chr.17: 4718676-4718676 on Build GRCh38
rs945868787 Chr.17: 4718535-4718535 on Build GRCh38
rs4522461 Chr.17: 4718478-4718478 on Build GRCh38
rs777343879 Chr.17: 4718598-4718598 on Build GRCh38
rs562865273 Chr.17: 4718588-4718588 on Build GRCh38
rs561991910 Chr.17: 4718732-4718732 on Build GRCh38
rs766395622 Chr.17: 4718573-4718573 on Build GRCh38
rs752157513 Chr.17: 4718595-4718595 on Build GRCh38
rs997495893 Chr.17: 4718671-4718671 on Build GRCh38
rs577204156 Chr.17: 4718540-4718540 on Build GRCh38
rs762870113 Chr.17: 4718683-4718683 on Build GRCh38
rs755799476 Chr.17: 4718597-4718597 on Build GRCh38
rs779963237 Chr.17: 4718487-4718487 on Build GRCh38
rs372780357 Chr.17: 4718694-4718694 on Build GRCh38
rs945887626 Chr.17: 4718331-4718331 on Build GRCh38
rs558012464 Chr.17: 4718486-4718486 on Build GRCh38
rs200044275 Chr.17: 4718523-4718524 on Build GRCh38
rs1016462136 Chr.17: 4718476-4718476 on Build GRCh38
rs764370768 Chr.17: 4718395-4718395 on Build GRCh38
rs775896165 Chr.17: 4718386-4718386 on Build GRCh38
rs370479564 Chr.17: 4718716-4718716 on Build GRCh38
rs376221990 Chr.17: 4718324-4718324 on Build GRCh38
rs1041792430 Chr.17: 4718649-4718649 on Build GRCh38
rs772042240 Chr.17: 4718385-4718385 on Build GRCh38
rs746019390 Chr.17: 4718384-4718384 on Build GRCh38
rs546145149 Chr.17: 4718558-4718558 on Build GRCh38
rs34496093 Chr.17: 4718408-4718408 on Build GRCh38
rs1020293286 Chr.17: 4718404-4718404 on Build GRCh38
rs767406643 Chr.17: 4718713-4718713 on Build GRCh38
rs370690870 Chr.17: 4718743-4718743 on Build GRCh38
rs769011737 Chr.17: 4718674-4718674 on Build GRCh38
rs963451269 Chr.17: 4718473-4718473 on Build GRCh38
rs977218321 Chr.17: 4718547-4718547 on Build GRCh38
rs370033247 Chr.17: 4718371-4718371 on Build GRCh38
rs774665003 Chr.17: 4718383-4718383 on Build GRCh38
rs756164545 Chr.17: 4718423-4718423 on Build GRCh38
rs3830452 Chr.17: 4718530-4718531 on Build GRCh38
rs775742956 Chr.17: 4718650-4718650 on Build GRCh38
rs113796886 Chr.17: 4718307-4718307 on Build GRCh38
rs749914147 Chr.17: 4718461-4718461 on Build GRCh38
rs368522371 Chr.17: 4718658-4718658 on Build GRCh38
rs911799994 Chr.17: 4718534-4718534 on Build GRCh38
rs760957342 Chr.17: 4718389-4718389 on Build GRCh38
rs761556531 Chr.17: 4718350-4718350 on Build GRCh38
rs1029211357 Chr.17: 4718439-4718439 on Build GRCh38
rs202087979 Chr.17: 4718717-4718717 on Build GRCh38
rs779125455 Chr.17: 4718730-4718730 on Build GRCh38
rs150965794 Chr.17: 4718376-4718376 on Build GRCh38
rs770188128 Chr.17: 4718375-4718375 on Build GRCh38
rs766053595 Chr.17: 4718692-4718692 on Build GRCh38
rs751444059 Chr.17: 4718574-4718574 on Build GRCh38
rs981090415 Chr.17: 4718529-4718529 on Build GRCh38
rs757550504 Chr.17: 4718625-4718625 on Build GRCh38
rs997528480 Chr.17: 4718438-4718438 on Build GRCh38
rs779542627 Chr.17: 4718626-4718626 on Build GRCh38
rs774392773 Chr.17: 4718693-4718693 on Build GRCh38
rs758979008 Chr.17: 4718736-4718736 on Build GRCh38
rs749679004 Chr.17: 4718602-4718602 on Build GRCh38
rs374363517 Chr.17: 4718580-4718580 on Build GRCh38
rs200781560 Chr.17: 4718726-4718726 on Build GRCh38
rs771476665 Chr.17: 4718382-4718382 on Build GRCh38

View Full Product Details