Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4932492-4932985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GP1BA
Entrez Gene ID
2811
Gene Name
glycoprotein Ib platelet alpha subunit
Gene Aliases
BDPLT1, BDPLT3, BSS, CD42B, CD42b-alpha, DBPLT3, GP1B, GPIbA, GPIbalpha, VWDP
UniGene
Hs.1472
Gene Chromosome Location
Chr.17: 4932275-4935030 on Build GRCh38
Transcripts
NM_000173.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
706964 c.361C>A p.L121M Chr.17: 4932965-4932965 on Build GRCh38
4067581 c.63G>T p.E21D Chr.17: 4932667-4932667 on Build GRCh38
6147325 c.108G>A p.R36R Chr.17: 4932712-4932712 on Build GRCh38
4067580 c.40T>G p.L14V Chr.17: 4932644-4932644 on Build GRCh38
4883350 c.310C>G p.Q104E Chr.17: 4932914-4932914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751416243 Chr.17: 4932671-4932671 on Build GRCh38
rs121908063 Chr.17: 4932821-4932821 on Build GRCh38
rs780863443 Chr.17: 4932681-4932681 on Build GRCh38
rs371712173 Chr.17: 4932793-4932793 on Build GRCh38
rs746692809 Chr.17: 4932584-4932584 on Build GRCh38
rs759849279 Chr.17: 4932636-4932636 on Build GRCh38
rs775619901 Chr.17: 4932724-4932724 on Build GRCh38
rs372373000 Chr.17: 4932641-4932641 on Build GRCh38
rs952083624 Chr.17: 4932649-4932649 on Build GRCh38
rs1015374822 Chr.17: 4932895-4932895 on Build GRCh38
rs999674592 Chr.17: 4932550-4932550 on Build GRCh38
rs183989314 Chr.17: 4932577-4932577 on Build GRCh38
rs371797414 Chr.17: 4932959-4932959 on Build GRCh38
rs752489274 Chr.17: 4932717-4932717 on Build GRCh38
rs773645699 Chr.17: 4932611-4932611 on Build GRCh38
rs758148266 Chr.17: 4932656-4932656 on Build GRCh38
rs778306339 Chr.17: 4932709-4932709 on Build GRCh38
rs974535671 Chr.17: 4932640-4932640 on Build GRCh38
rs776888187 Chr.17: 4932925-4932925 on Build GRCh38
rs201827537 Chr.17: 4932696-4932696 on Build GRCh38
rs554661306 Chr.17: 4932544-4932544 on Build GRCh38
rs373756376 Chr.17: 4932938-4932938 on Build GRCh38
rs761923069 Chr.17: 4932844-4932844 on Build GRCh38
rs760556141 Chr.17: 4932941-4932941 on Build GRCh38
rs746407497 Chr.17: 4932891-4932891 on Build GRCh38
rs770303624 Chr.17: 4932603-4932603 on Build GRCh38
rs992419285 Chr.17: 4932704-4932704 on Build GRCh38
rs767153882 Chr.17: 4932785-4932785 on Build GRCh38
rs760041588 Chr.17: 4932718-4932718 on Build GRCh38
rs766966718 Chr.17: 4932954-4932954 on Build GRCh38
rs768722388 Chr.17: 4932840-4932840 on Build GRCh38
rs544642889 Chr.17: 4932528-4932528 on Build GRCh38
rs774841072 Chr.17: 4932716-4932716 on Build GRCh38
rs768098161 Chr.17: 4932585-4932585 on Build GRCh38
rs561538792 Chr.17: 4932877-4932877 on Build GRCh38
rs754622081 Chr.17: 4932672-4932672 on Build GRCh38
rs767906261 Chr.17: 4932638-4932638 on Build GRCh38
rs751849232 Chr.17: 4932964-4932964 on Build GRCh38
rs756056031 Chr.17: 4932801-4932801 on Build GRCh38
rs748862472 Chr.17: 4932796-4932796 on Build GRCh38
rs763331024 Chr.17: 4932624-4932624 on Build GRCh38
rs755711021 Chr.17: 4932868-4932868 on Build GRCh38
rs752327740 Chr.17: 4932786-4932786 on Build GRCh38
rs763628591 Chr.17: 4932945-4932945 on Build GRCh38
rs757202817 Chr.17: 4932571-4932571 on Build GRCh38
rs757831948 Chr.17: 4932801-4932801 on Build GRCh38
rs560610176 Chr.17: 4932591-4932591 on Build GRCh38
rs369323536 Chr.17: 4932935-4932935 on Build GRCh38
rs377601326 Chr.17: 4932961-4932961 on Build GRCh38
rs779826628 Chr.17: 4932669-4932669 on Build GRCh38
rs760705175 Chr.17: 4932653-4932653 on Build GRCh38
rs751326171 Chr.17: 4932776-4932776 on Build GRCh38
rs759290392 Chr.17: 4932850-4932850 on Build GRCh38
rs761366532 Chr.17: 4932953-4932953 on Build GRCh38
rs13306411 Chr.17: 4932860-4932860 on Build GRCh38
rs778779735 Chr.17: 4932575-4932575 on Build GRCh38
rs748178518 Chr.17: 4932928-4932928 on Build GRCh38
rs767711763 Chr.17: 4932561-4932561 on Build GRCh38
rs776571145 Chr.17: 4932841-4932841 on Build GRCh38
rs747475095 Chr.17: 4932908-4932908 on Build GRCh38
rs774650985 Chr.17: 4932634-4932634 on Build GRCh38
rs982160551 Chr.17: 4932865-4932865 on Build GRCh38
rs753658079 Chr.17: 4932569-4932569 on Build GRCh38
rs753563601 Chr.17: 4932950-4932950 on Build GRCh38
rs772389629 Chr.17: 4932720-4932720 on Build GRCh38
rs780652188 Chr.17: 4932962-4932962 on Build GRCh38
rs781032554 Chr.17: 4932611-4932613 on Build GRCh38
rs1043841849 Chr.17: 4932748-4932748 on Build GRCh38
rs111292798 Chr.17: 4932710-4932710 on Build GRCh38
rs81663 Chr.17: 4932557-4932557 on Build GRCh38
rs6068 Chr.17: 4932819-4932819 on Build GRCh38
rs1018866562 Chr.17: 4932833-4932833 on Build GRCh38
rs767224320 Chr.17: 4932851-4932851 on Build GRCh38
rs138825640 Chr.17: 4932810-4932810 on Build GRCh38
rs763871261 Chr.17: 4932875-4932875 on Build GRCh38
rs755156892 Chr.17: 4932571-4932572 on Build GRCh38
rs753365146 Chr.17: 4932876-4932876 on Build GRCh38
rs866570250 Chr.17: 4932548-4932548 on Build GRCh38
rs542182223 Chr.17: 4932802-4932802 on Build GRCh38
rs751721207 Chr.17: 4932566-4932567 on Build GRCh38
rs777896666 Chr.17: 4932812-4932813 on Build GRCh38
rs911283855 Chr.17: 4932542-4932542 on Build GRCh38
rs375350354 Chr.17: 4932892-4932892 on Build GRCh38
rs958143327 Chr.17: 4932574-4932574 on Build GRCh38
rs916849846 Chr.17: 4932907-4932907 on Build GRCh38
rs780294091 Chr.17: 4932905-4932905 on Build GRCh38
rs960740250 Chr.17: 4932901-4932901 on Build GRCh38
rs935734274 Chr.17: 4932764-4932764 on Build GRCh38
rs781541857 Chr.17: 4932845-4932845 on Build GRCh38
rs750305979 Chr.17: 4932742-4932742 on Build GRCh38
rs1023639222 Chr.17: 4932823-4932823 on Build GRCh38
rs760759446 Chr.17: 4932741-4932741 on Build GRCh38
rs755588034 Chr.17: 4932792-4932792 on Build GRCh38
rs376091002 Chr.17: 4932578-4932578 on Build GRCh38
rs201512242 Chr.17: 4932848-4932848 on Build GRCh38
rs2243093 Chr.17: 4932600-4932600 on Build GRCh38
rs376767591 Chr.17: 4932934-4932934 on Build GRCh38
rs766371816 Chr.17: 4932765-4932765 on Build GRCh38
rs373524550 Chr.17: 4932919-4932919 on Build GRCh38
rs754816223 Chr.17: 4932778-4932778 on Build GRCh38
rs747673131 Chr.17: 4932694-4932694 on Build GRCh38
rs965941179 Chr.17: 4932523-4932523 on Build GRCh38
rs771456681 Chr.17: 4932631-4932631 on Build GRCh38
rs772587498 Chr.17: 4932803-4932803 on Build GRCh38
rs765198146 Chr.17: 4932655-4932655 on Build GRCh38
rs900384352 Chr.17: 4932847-4932847 on Build GRCh38
rs775140954 Chr.17: 4932940-4932940 on Build GRCh38
rs771366895 Chr.17: 4932711-4932711 on Build GRCh38

View Full Product Details