Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5474569-5475066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL2
Entrez Gene ID
51009
Gene Name
derlin 2
Gene Aliases
CGI-101, DERtrin-2, F-LAN-1, F-LANa, FLANa
UniGene
Hs.286131
Gene Chromosome Location
Chr.17: 5471251-5486230 on Build GRCh38
Transcripts
NM_001304777.1 NM_001304779.1 NM_016041.4 NR_130905.1 NR_130906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1709646 c.634C>T p.P212S Chr.17: 5474770-5474770 on Build GRCh38
7206929 c.635C>T p.P212L Chr.17: 5474769-5474769 on Build GRCh38
4677110 c.689C>T p.A230V Chr.17: 5474715-5474715 on Build GRCh38
1384627 c.674G>A p.R225Q Chr.17: 5474730-5474730 on Build GRCh38
5473918 c.632C>T p.T211I Chr.17: 5474772-5474772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925954385 Chr.17: 5474980-5474981 on Build GRCh38
rs1055752194 Chr.17: 5475035-5475035 on Build GRCh38
rs749900505 Chr.17: 5474667-5474667 on Build GRCh38
rs376886364 Chr.17: 5474644-5474644 on Build GRCh38
rs374575086 Chr.17: 5474768-5474768 on Build GRCh38
rs568093448 Chr.17: 5475032-5475032 on Build GRCh38
rs751446055 Chr.17: 5474732-5474732 on Build GRCh38
rs748768739 Chr.17: 5474785-5474785 on Build GRCh38
rs904379066 Chr.17: 5474687-5474687 on Build GRCh38
rs764352407 Chr.17: 5474837-5474837 on Build GRCh38
rs548751988 Chr.17: 5474984-5474984 on Build GRCh38
rs1023338744 Chr.17: 5475019-5475019 on Build GRCh38
rs774587285 Chr.17: 5474679-5474679 on Build GRCh38
rs529217125 Chr.17: 5474933-5474933 on Build GRCh38
rs944005426 Chr.17: 5474808-5474808 on Build GRCh38
rs760996782 Chr.17: 5474828-5474828 on Build GRCh38
rs969669315 Chr.17: 5474612-5474612 on Build GRCh38
rs760016431 Chr.17: 5474691-5474691 on Build GRCh38
rs1004692051 Chr.17: 5474912-5474912 on Build GRCh38
rs200000601 Chr.17: 5474741-5474741 on Build GRCh38
rs772601705 Chr.17: 5474821-5474821 on Build GRCh38
rs913101552 Chr.17: 5474928-5474928 on Build GRCh38
rs770347779 Chr.17: 5474805-5474805 on Build GRCh38
rs1013630324 Chr.17: 5474981-5474981 on Build GRCh38
rs368865081 Chr.17: 5474639-5474639 on Build GRCh38
rs373835634 Chr.17: 5474809-5474809 on Build GRCh38
rs954631605 Chr.17: 5474886-5474886 on Build GRCh38
rs760946814 Chr.17: 5474705-5474705 on Build GRCh38
rs371213424 Chr.17: 5474717-5474717 on Build GRCh38
rs780934634 Chr.17: 5474763-5474763 on Build GRCh38
rs931953470 Chr.17: 5474847-5474847 on Build GRCh38
rs775900239 Chr.17: 5474697-5474697 on Build GRCh38
rs35557217 Chr.17: 5474985-5474986 on Build GRCh38
rs749784803 Chr.17: 5474654-5474654 on Build GRCh38
rs563098081 Chr.17: 5474869-5474869 on Build GRCh38
rs754852260 Chr.17: 5474737-5474737 on Build GRCh38
rs967452988 Chr.17: 5474939-5474939 on Build GRCh38
rs896243883 Chr.17: 5474941-5474941 on Build GRCh38
rs887636324 Chr.17: 5474888-5474888 on Build GRCh38
rs764297327 Chr.17: 5474714-5474714 on Build GRCh38
rs988245118 Chr.17: 5474731-5474731 on Build GRCh38
rs963392902 Chr.17: 5474826-5474826 on Build GRCh38
rs746335418 Chr.17: 5474810-5474810 on Build GRCh38
rs747830827 Chr.17: 5474767-5474767 on Build GRCh38
rs200663470 Chr.17: 5474985-5474985 on Build GRCh38
rs979259873 Chr.17: 5474977-5474977 on Build GRCh38
rs771413501 Chr.17: 5474670-5474670 on Build GRCh38
rs1024272245 Chr.17: 5474608-5474608 on Build GRCh38
rs777282443 Chr.17: 5474772-5474772 on Build GRCh38
rs925474438 Chr.17: 5474665-5474665 on Build GRCh38
rs368073127 Chr.17: 5474824-5474824 on Build GRCh38
rs766273072 Chr.17: 5474726-5474726 on Build GRCh38
rs531767745 Chr.17: 5474943-5474943 on Build GRCh38
rs767757387 Chr.17: 5474695-5474695 on Build GRCh38

View Full Product Details