Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6650233-6650752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED31
Entrez Gene ID
51003
Gene Name
mediator complex subunit 31
Gene Aliases
3110004H13Rik, CGI-125, Soh1
UniGene
Hs.567493
Gene Chromosome Location
Chr.17: 6643313-6651804 on Build GRCh38
Transcripts
NM_016060.2 XM_017024710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
983261 c.30T>C p.D10D Chr.17: 6650432-6650432 on Build GRCh38
6393709 c.91C>T p.P31S Chr.17: 6650371-6650371 on Build GRCh38
6430818 c.34G>A p.A12T Chr.17: 6650428-6650428 on Build GRCh38
3370865 c.105T>C p.N35N Chr.17: 6650357-6650357 on Build GRCh38
460258 c.55C>T p.Q19* Chr.17: 6650407-6650407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536875583 Chr.17: 6650280-6650280 on Build GRCh38
rs1023450664 Chr.17: 6650687-6650687 on Build GRCh38
rs1036777234 Chr.17: 6650623-6650623 on Build GRCh38
rs758096065 Chr.17: 6650457-6650457 on Build GRCh38
rs781706588 Chr.17: 6650306-6650306 on Build GRCh38
rs574146260 Chr.17: 6650600-6650600 on Build GRCh38
rs1049331559 Chr.17: 6650562-6650562 on Build GRCh38
rs1037833394 Chr.17: 6650421-6650421 on Build GRCh38
rs376595857 Chr.17: 6650430-6650430 on Build GRCh38
rs766985027 Chr.17: 6650554-6650554 on Build GRCh38
rs899345338 Chr.17: 6650428-6650428 on Build GRCh38
rs772963194 Chr.17: 6650668-6650668 on Build GRCh38
rs766066805 Chr.17: 6650462-6650462 on Build GRCh38
rs750185288 Chr.17: 6650452-6650452 on Build GRCh38
rs770055375 Chr.17: 6650323-6650323 on Build GRCh38
rs976185917 Chr.17: 6650526-6650526 on Build GRCh38
rs774685951 Chr.17: 6650361-6650361 on Build GRCh38
rs1013689945 Chr.17: 6650684-6650684 on Build GRCh38
rs374641139 Chr.17: 6650317-6650317 on Build GRCh38
rs543402999 Chr.17: 6650515-6650515 on Build GRCh38
rs542173320 Chr.17: 6650579-6650579 on Build GRCh38
rs2301874 Chr.17: 6650263-6650263 on Build GRCh38
rs1013381587 Chr.17: 6650282-6650282 on Build GRCh38
rs1023481710 Chr.17: 6650305-6650305 on Build GRCh38
rs56978514 Chr.17: 6650581-6650581 on Build GRCh38
rs912353817 Chr.17: 6650496-6650496 on Build GRCh38
rs988287334 Chr.17: 6650491-6650491 on Build GRCh38
rs1008553263 Chr.17: 6650459-6650459 on Build GRCh38
rs932211269 Chr.17: 6650555-6650555 on Build GRCh38
rs773564415 Chr.17: 6650338-6650338 on Build GRCh38
rs1000372565 Chr.17: 6650580-6650580 on Build GRCh38
rs368920254 Chr.17: 6650355-6650355 on Build GRCh38
rs774337426 Chr.17: 6650277-6650277 on Build GRCh38
rs572374686 Chr.17: 6650593-6650593 on Build GRCh38
rs991403063 Chr.17: 6650626-6650626 on Build GRCh38
rs371954305 Chr.17: 6650404-6650404 on Build GRCh38
rs969265832 Chr.17: 6650326-6650326 on Build GRCh38
rs753894294 Chr.17: 6650431-6650431 on Build GRCh38
rs1000769204 Chr.17: 6650698-6650698 on Build GRCh38
rs748683122 Chr.17: 6650309-6650309 on Build GRCh38
rs982073360 Chr.17: 6650692-6650692 on Build GRCh38
rs1032292822 Chr.17: 6650708-6650708 on Build GRCh38
rs764957715 Chr.17: 6650438-6650438 on Build GRCh38
rs767695388 Chr.17: 6650419-6650419 on Build GRCh38
rs1024266537 Chr.17: 6650637-6650637 on Build GRCh38
rs557014719 Chr.17: 6650502-6650502 on Build GRCh38
rs760675542 Chr.17: 6650312-6650314 on Build GRCh38
rs896310264 Chr.17: 6650266-6650266 on Build GRCh38
rs564657294 Chr.17: 6650711-6650711 on Build GRCh38
rs541724751 Chr.17: 6650695-6650695 on Build GRCh38

View Full Product Details