Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15437493-15438014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT4
Entrez Gene ID
284040
Gene Name
CMT1A duplicated region transcript 4
Gene Aliases
-
UniGene
Hs.164595
Gene Chromosome Location
Chr.17: 15436015-15467611 on Build GRCh38
Transcripts
NM_001204477.1 NM_001204478.1 NR_037924.1

Gene Symbol
TVP23C-CDRT4
Entrez Gene ID
100533496
Gene Name
TVP23C-CDRT4 readthrough
Gene Aliases
FAM18B2-CDRT4
UniGene
Hs.164595
Gene Chromosome Location
Chr.17: 15436015-15563631 on Build GRCh38
Transcripts
NM_001204477.1 NM_001204478.1 NR_037924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4617722 c.323C>T p.A108V Chr.17: 15437906-15437906 on Build GRCh38
6002470 c.298A>T p.M100L Chr.17: 15437931-15437931 on Build GRCh38
6189902 c.399C>A p.N133K Chr.17: 15437830-15437830 on Build GRCh38
4623479 c.300G>A p.M100I Chr.17: 15437929-15437929 on Build GRCh38
6481983 c.373A>T p.R125* Chr.17: 15437856-15437856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923847412 Chr.17: 15437652-15437652 on Build GRCh38
rs989369077 Chr.17: 15437938-15437938 on Build GRCh38
rs775196750 Chr.17: 15437923-15437924 on Build GRCh38
rs778087419 Chr.17: 15437951-15437951 on Build GRCh38
rs774381341 Chr.17: 15437813-15437813 on Build GRCh38
rs763410708 Chr.17: 15437918-15437918 on Build GRCh38
rs371231050 Chr.17: 15437705-15437706 on Build GRCh38
rs758635780 Chr.17: 15437860-15437860 on Build GRCh38
rs566277630 Chr.17: 15437644-15437644 on Build GRCh38
rs142965522 Chr.17: 15437917-15437917 on Build GRCh38
rs759058630 Chr.17: 15437749-15437749 on Build GRCh38
rs1016372707 Chr.17: 15437862-15437862 on Build GRCh38
rs767174235 Chr.17: 15437815-15437815 on Build GRCh38
rs527757226 Chr.17: 15437897-15437897 on Build GRCh38
rs35353219 Chr.17: 15437844-15437844 on Build GRCh38
rs76787574 Chr.17: 15437706-15437706 on Build GRCh38
rs749567945 Chr.17: 15437534-15437534 on Build GRCh38
rs750738109 Chr.17: 15437762-15437762 on Build GRCh38
rs756426503 Chr.17: 15437874-15437874 on Build GRCh38
rs34381006 Chr.17: 15437923-15437923 on Build GRCh38
rs201152482 Chr.17: 15437911-15437911 on Build GRCh38
rs963439372 Chr.17: 15437551-15437551 on Build GRCh38
rs78593311 Chr.17: 15437712-15437712 on Build GRCh38
rs775130353 Chr.17: 15437902-15437902 on Build GRCh38
rs780374556 Chr.17: 15437528-15437528 on Build GRCh38
rs778817741 Chr.17: 15437879-15437879 on Build GRCh38
rs61742154 Chr.17: 15437864-15437864 on Build GRCh38
rs747109073 Chr.17: 15437788-15437788 on Build GRCh38
rs2954759 Chr.17: 15437866-15437866 on Build GRCh38
rs576928505 Chr.17: 15437639-15437639 on Build GRCh38
rs775114929 Chr.17: 15437829-15437829 on Build GRCh38
rs751192881 Chr.17: 15437718-15437718 on Build GRCh38
rs150459531 Chr.17: 15437638-15437638 on Build GRCh38
rs781426841 Chr.17: 15437792-15437792 on Build GRCh38
rs199557231 Chr.17: 15437752-15437752 on Build GRCh38
rs2954757 Chr.17: 15437705-15437705 on Build GRCh38
rs755005337 Chr.17: 15437791-15437791 on Build GRCh38
rs752827246 Chr.17: 15437937-15437937 on Build GRCh38
rs201168400 Chr.17: 15437704-15437705 on Build GRCh38
rs562020164 Chr.17: 15437878-15437878 on Build GRCh38
rs138248340 Chr.17: 15437683-15437683 on Build GRCh38
rs1039096062 Chr.17: 15437720-15437720 on Build GRCh38
rs61742151 Chr.17: 15437849-15437849 on Build GRCh38
rs145055448 Chr.17: 15437939-15437939 on Build GRCh38
rs760033711 Chr.17: 15437913-15437913 on Build GRCh38
rs745848525 Chr.17: 15437891-15437891 on Build GRCh38
rs549405492 Chr.17: 15437555-15437555 on Build GRCh38
rs755690353 Chr.17: 15437754-15437754 on Build GRCh38
rs371941860 Chr.17: 15437706-15437706 on Build GRCh38
rs748443857 Chr.17: 15437793-15437793 on Build GRCh38
rs766897737 Chr.17: 15437863-15437863 on Build GRCh38
rs982034130 Chr.17: 15437640-15437640 on Build GRCh38
rs146969230 Chr.17: 15437922-15437922 on Build GRCh38
rs1023351800 Chr.17: 15437873-15437873 on Build GRCh38
rs892643029 Chr.17: 15437759-15437759 on Build GRCh38
rs533614131 Chr.17: 15437805-15437805 on Build GRCh38
rs199516358 Chr.17: 15437814-15437814 on Build GRCh38
rs768007937 Chr.17: 15437931-15437931 on Build GRCh38
rs762998630 Chr.17: 15437738-15437738 on Build GRCh38
rs1004997954 Chr.17: 15437522-15437522 on Build GRCh38
rs149000474 Chr.17: 15437914-15437914 on Build GRCh38
rs759006768 Chr.17: 15437780-15437780 on Build GRCh38
rs1023734408 Chr.17: 15437579-15437579 on Build GRCh38
rs935192300 Chr.17: 15437663-15437663 on Build GRCh38
rs141346403 Chr.17: 15437924-15437924 on Build GRCh38
rs531401802 Chr.17: 15437707-15437707 on Build GRCh38
rs367911828 Chr.17: 15437953-15437953 on Build GRCh38
rs778905762 Chr.17: 15437987-15437987 on Build GRCh38
rs780468845 Chr.17: 15437781-15437781 on Build GRCh38
rs141229496 Chr.17: 15437747-15437747 on Build GRCh38
rs1016006019 Chr.17: 15437543-15437543 on Build GRCh38
rs75800378 Chr.17: 15437604-15437604 on Build GRCh38
rs371852022 Chr.17: 15437954-15437954 on Build GRCh38
rs990528840 Chr.17: 15437576-15437576 on Build GRCh38
rs769239378 Chr.17: 15437561-15437561 on Build GRCh38
rs1056517136 Chr.17: 15437753-15437753 on Build GRCh38
rs773118303 Chr.17: 15437801-15437801 on Build GRCh38
rs770914350 Chr.17: 15437812-15437812 on Build GRCh38
rs980793499 Chr.17: 15437895-15437895 on Build GRCh38
rs537731998 Chr.17: 15437708-15437708 on Build GRCh38
rs765680695 Chr.17: 15437756-15437756 on Build GRCh38
rs769887028 Chr.17: 15437795-15437795 on Build GRCh38
rs769595009 Chr.17: 15437733-15437733 on Build GRCh38
rs760431282 Chr.17: 15437834-15437834 on Build GRCh38
rs752708188 Chr.17: 15437868-15437868 on Build GRCh38
rs766213090 Chr.17: 15437740-15437740 on Build GRCh38
rs145874622 Chr.17: 15437876-15437876 on Build GRCh38
rs532260366 Chr.17: 15437627-15437627 on Build GRCh38
rs973996893 Chr.17: 15437625-15437625 on Build GRCh38
rs772852837 Chr.17: 15437736-15437736 on Build GRCh38
rs373614376 Chr.17: 15437804-15437804 on Build GRCh38
rs201573651 Chr.17: 15437892-15437892 on Build GRCh38

View Full Product Details