Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28863130-28863624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPRD1
Entrez Gene ID
4985
Gene Name
opioid receptor delta 1
Gene Aliases
OPRD
UniGene
Hs.372
Gene Chromosome Location
Chr.1: 28812142-28863696 on Build GRCh38
Transcripts
NM_000911.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116785 c.1032C>G p.S344R Chr.1: 28863196-28863196 on Build GRCh38
5623714 c.1004A>C p.K335T Chr.1: 28863168-28863168 on Build GRCh38
1259945 c.1094G>A p.G365D Chr.1: 28863258-28863258 on Build GRCh38
5444416 c.1048G>A p.E350K Chr.1: 28863212-28863212 on Build GRCh38
4799199 c.1005G>T p.K335N Chr.1: 28863169-28863169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992222061 Chr.1: 28863349-28863349 on Build GRCh38
rs1032188468 Chr.1: 28863423-28863423 on Build GRCh38
rs895842416 Chr.1: 28863486-28863486 on Build GRCh38
rs751125712 Chr.1: 28863246-28863246 on Build GRCh38
rs781771653 Chr.1: 28863258-28863258 on Build GRCh38
rs781740622 Chr.1: 28863165-28863165 on Build GRCh38
rs972936131 Chr.1: 28863289-28863289 on Build GRCh38
rs770174225 Chr.1: 28863263-28863263 on Build GRCh38
rs765128633 Chr.1: 28863210-28863210 on Build GRCh38
rs770240637 Chr.1: 28863178-28863178 on Build GRCh38
rs761939314 Chr.1: 28863229-28863238 on Build GRCh38
rs1023064027 Chr.1: 28863500-28863500 on Build GRCh38
rs753777054 Chr.1: 28863494-28863494 on Build GRCh38
rs893644779 Chr.1: 28863478-28863478 on Build GRCh38
rs572485582 Chr.1: 28863366-28863366 on Build GRCh38
rs999388913 Chr.1: 28863398-28863398 on Build GRCh38
rs990260976 Chr.1: 28863226-28863226 on Build GRCh38
rs759556278 Chr.1: 28863201-28863201 on Build GRCh38
rs1057381432 Chr.1: 28863402-28863402 on Build GRCh38
rs1035690657 Chr.1: 28863589-28863589 on Build GRCh38
rs573922084 Chr.1: 28863175-28863175 on Build GRCh38
rs879656886 Chr.1: 28863229-28863229 on Build GRCh38
rs970535328 Chr.1: 28863508-28863508 on Build GRCh38
rs369999443 Chr.1: 28863223-28863223 on Build GRCh38
rs1023277070 Chr.1: 28863354-28863354 on Build GRCh38
rs1040667582 Chr.1: 28863393-28863393 on Build GRCh38
rs778236672 Chr.1: 28863281-28863281 on Build GRCh38
rs1012471802 Chr.1: 28863492-28863492 on Build GRCh38
rs771202637 Chr.1: 28863318-28863318 on Build GRCh38
rs761620507 Chr.1: 28863205-28863205 on Build GRCh38
rs940238521 Chr.1: 28863279-28863279 on Build GRCh38
rs774551884 Chr.1: 28863191-28863191 on Build GRCh38
rs889503464 Chr.1: 28863361-28863361 on Build GRCh38
rs577425950 Chr.1: 28863545-28863545 on Build GRCh38
rs904614483 Chr.1: 28863545-28863545 on Build GRCh38
rs943745833 Chr.1: 28863365-28863365 on Build GRCh38
rs773326863 Chr.1: 28863179-28863179 on Build GRCh38
rs763280814 Chr.1: 28863180-28863180 on Build GRCh38
rs753737057 Chr.1: 28863204-28863204 on Build GRCh38
rs931433616 Chr.1: 28863324-28863324 on Build GRCh38
rs555836624 Chr.1: 28863288-28863288 on Build GRCh38
rs375950124 Chr.1: 28863589-28863590 on Build GRCh38
rs764009052 Chr.1: 28863262-28863262 on Build GRCh38
rs902475485 Chr.1: 28863394-28863394 on Build GRCh38
rs550851651 Chr.1: 28863152-28863152 on Build GRCh38
rs201916183 Chr.1: 28863182-28863182 on Build GRCh38
rs960426781 Chr.1: 28863345-28863345 on Build GRCh38
rs535844446 Chr.1: 28863266-28863266 on Build GRCh38
rs1049511839 Chr.1: 28863334-28863334 on Build GRCh38
rs988016403 Chr.1: 28863195-28863195 on Build GRCh38
rs779559537 Chr.1: 28863231-28863231 on Build GRCh38
rs975083278 Chr.1: 28863249-28863249 on Build GRCh38
rs981464821 Chr.1: 28863532-28863532 on Build GRCh38
rs999336782 Chr.1: 28863158-28863158 on Build GRCh38
rs758135542 Chr.1: 28863225-28863225 on Build GRCh38
rs748614920 Chr.1: 28863261-28863261 on Build GRCh38
rs567567465 Chr.1: 28863253-28863253 on Build GRCh38
rs887269154 Chr.1: 28863355-28863355 on Build GRCh38
rs764026265 Chr.1: 28863203-28863203 on Build GRCh38

View Full Product Details