Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31585567-31586066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TINAGL1
Entrez Gene ID
64129
Gene Name
tubulointerstitial nephritis antigen like 1
Gene Aliases
ARG1, LCN7, LIECG3, TINAGRP
UniGene
Hs.199368
Gene Chromosome Location
Chr.1: 31576384-31587686 on Build GRCh38
Transcripts
NM_001204414.1 NM_001204415.1 NM_022164.2 XM_005271106.2 XM_005271107.2 XM_011541946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6801946 c.1124T>C p.L375P Chr.1: 31585783-31585783 on Build GRCh38
3487981 c.1187G>A p.R396Q Chr.1: 31585846-31585846 on Build GRCh38
235484 c.1200C>T p.H400H Chr.1: 31585859-31585859 on Build GRCh38
3789874 c.1094-1G>A p.? Chr.1: 31585752-31585752 on Build GRCh38
6801947 c.1217+9C>T p.? Chr.1: 31585885-31585885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780235917 Chr.1: 31585822-31585822 on Build GRCh38
rs749184739 Chr.1: 31585842-31585842 on Build GRCh38
rs895767301 Chr.1: 31585670-31585670 on Build GRCh38
rs375984520 Chr.1: 31585843-31585843 on Build GRCh38
rs965544425 Chr.1: 31585614-31585614 on Build GRCh38
rs189923612 Chr.1: 31585769-31585769 on Build GRCh38
rs151277916 Chr.1: 31585994-31585994 on Build GRCh38
rs1009779636 Chr.1: 31585607-31585607 on Build GRCh38
rs532730668 Chr.1: 31585871-31585871 on Build GRCh38
rs371435956 Chr.1: 31585841-31585841 on Build GRCh38
rs576188446 Chr.1: 31585601-31585601 on Build GRCh38
rs200310989 Chr.1: 31585886-31585886 on Build GRCh38
rs886847875 Chr.1: 31585644-31585644 on Build GRCh38
rs879199474 Chr.1: 31585747-31585747 on Build GRCh38
rs548481850 Chr.1: 31586041-31586041 on Build GRCh38
rs773940511 Chr.1: 31585812-31585812 on Build GRCh38
rs778593912 Chr.1: 31585846-31585846 on Build GRCh38
rs200193842 Chr.1: 31585899-31585899 on Build GRCh38
rs375972013 Chr.1: 31585726-31585726 on Build GRCh38
rs879072950 Chr.1: 31585749-31585749 on Build GRCh38
rs1046657875 Chr.1: 31585966-31585966 on Build GRCh38
rs771800137 Chr.1: 31585709-31585709 on Build GRCh38
rs775188538 Chr.1: 31585716-31585716 on Build GRCh38
rs748266805 Chr.1: 31586008-31586008 on Build GRCh38
rs773502658 Chr.1: 31585902-31585902 on Build GRCh38
rs571792351 Chr.1: 31585636-31585636 on Build GRCh38
rs562991466 Chr.1: 31585948-31585948 on Build GRCh38
rs751700967 Chr.1: 31585832-31585832 on Build GRCh38
rs569995809 Chr.1: 31585901-31585901 on Build GRCh38
rs755955159 Chr.1: 31585965-31585965 on Build GRCh38
rs756686905 Chr.1: 31585708-31585708 on Build GRCh38
rs920929912 Chr.1: 31585612-31585612 on Build GRCh38
rs1057365333 Chr.1: 31585669-31585669 on Build GRCh38
rs879056807 Chr.1: 31585750-31585750 on Build GRCh38
rs370385551 Chr.1: 31585613-31585613 on Build GRCh38
rs879032102 Chr.1: 31585746-31585746 on Build GRCh38
rs1022843134 Chr.1: 31585748-31585748 on Build GRCh38
rs758386268 Chr.1: 31585605-31585605 on Build GRCh38
rs755033788 Chr.1: 31585834-31585834 on Build GRCh38
rs746577046 Chr.1: 31585885-31585885 on Build GRCh38
rs965533540 Chr.1: 31585918-31585918 on Build GRCh38
rs976087458 Chr.1: 31586036-31586036 on Build GRCh38
rs372930486 Chr.1: 31585848-31585848 on Build GRCh38
rs764765103 Chr.1: 31585734-31585734 on Build GRCh38
rs1048146733 Chr.1: 31585614-31585614 on Build GRCh38
rs561486262 Chr.1: 31585634-31585634 on Build GRCh38
rs148269987 Chr.1: 31585810-31585810 on Build GRCh38
rs150290691 Chr.1: 31585820-31585820 on Build GRCh38
rs111241208 Chr.1: 31585877-31585877 on Build GRCh38
rs1004027311 Chr.1: 31585665-31585665 on Build GRCh38
rs750762255 Chr.1: 31585811-31585811 on Build GRCh38
rs376164784 Chr.1: 31585613-31585614 on Build GRCh38
rs754287330 Chr.1: 31585740-31585740 on Build GRCh38
rs763277474 Chr.1: 31585907-31585907 on Build GRCh38
rs771890329 Chr.1: 31585849-31585849 on Build GRCh38

View Full Product Details