Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31798202-31798724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPOCD1
Entrez Gene ID
90853
Gene Name
SPOC domain containing 1
Gene Aliases
PPP1R146
UniGene
Hs.62604
Gene Chromosome Location
Chr.1: 31790422-31816051 on Build GRCh38
Transcripts
NM_001281987.1 NM_001281988.1 NM_144569.5 XM_011542413.2 XM_011542414.2 XM_017002779.1 XM_017002780.1 XM_017002781.1 XM_017002782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6191771 c.1925C>G p.A642G Chr.1: 31798545-31798545 on Build GRCh38
164570 c.1972C>T p.R658W Chr.1: 31798498-31798498 on Build GRCh38
414483 c.1944C>A p.L648L Chr.1: 31798526-31798526 on Build GRCh38
6063007 c.2074C>A p.L692M Chr.1: 31798278-31798278 on Build GRCh38
908214 c.2080C>T p.R694W Chr.1: 31798272-31798272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750781677 Chr.1: 31798677-31798677 on Build GRCh38
rs986368869 Chr.1: 31798395-31798395 on Build GRCh38
rs140554876 Chr.1: 31798590-31798590 on Build GRCh38
rs765864823 Chr.1: 31798248-31798248 on Build GRCh38
rs758896643 Chr.1: 31798684-31798684 on Build GRCh38
rs143924712 Chr.1: 31798375-31798375 on Build GRCh38
rs750415462 Chr.1: 31798370-31798370 on Build GRCh38
rs144387561 Chr.1: 31798282-31798282 on Build GRCh38
rs969147587 Chr.1: 31798270-31798270 on Build GRCh38
rs751604871 Chr.1: 31798283-31798283 on Build GRCh38
rs755115371 Chr.1: 31798286-31798286 on Build GRCh38
rs374813332 Chr.1: 31798359-31798359 on Build GRCh38
rs982623395 Chr.1: 31798392-31798392 on Build GRCh38
rs780646311 Chr.1: 31798605-31798605 on Build GRCh38
rs534016132 Chr.1: 31798320-31798320 on Build GRCh38
rs367626644 Chr.1: 31798697-31798697 on Build GRCh38
rs776087777 Chr.1: 31798611-31798611 on Build GRCh38
rs1039924664 Chr.1: 31798597-31798597 on Build GRCh38
rs965327054 Chr.1: 31798368-31798368 on Build GRCh38
rs770158498 Chr.1: 31798308-31798308 on Build GRCh38
rs575258605 Chr.1: 31798629-31798629 on Build GRCh38
rs757574494 Chr.1: 31798663-31798663 on Build GRCh38
rs746327804 Chr.1: 31798596-31798596 on Build GRCh38
rs763854723 Chr.1: 31798639-31798639 on Build GRCh38
rs760616184 Chr.1: 31798637-31798637 on Build GRCh38
rs773447239 Chr.1: 31798412-31798412 on Build GRCh38
rs774405449 Chr.1: 31798635-31798635 on Build GRCh38
rs140694403 Chr.1: 31798556-31798556 on Build GRCh38
rs771483105 Chr.1: 31798388-31798388 on Build GRCh38
rs146864047 Chr.1: 31798398-31798398 on Build GRCh38
rs760361459 Chr.1: 31798622-31798623 on Build GRCh38
rs771534395 Chr.1: 31798318-31798318 on Build GRCh38
rs143458300 Chr.1: 31798555-31798555 on Build GRCh38
rs773096538 Chr.1: 31798421-31798421 on Build GRCh38
rs757412948 Chr.1: 31798253-31798262 on Build GRCh38
rs756285560 Chr.1: 31798554-31798554 on Build GRCh38
rs560840320 Chr.1: 31798686-31798686 on Build GRCh38
rs774885380 Chr.1: 31798410-31798410 on Build GRCh38
rs771667382 Chr.1: 31798400-31798400 on Build GRCh38
rs111559582 Chr.1: 31798420-31798420 on Build GRCh38
rs377381369 Chr.1: 31798416-31798416 on Build GRCh38
rs202136039 Chr.1: 31798227-31798227 on Build GRCh38
rs774818084 Chr.1: 31798247-31798247 on Build GRCh38
rs779503315 Chr.1: 31798390-31798390 on Build GRCh38
rs760275131 Chr.1: 31798334-31798334 on Build GRCh38
rs904935025 Chr.1: 31798539-31798539 on Build GRCh38
rs758952336 Chr.1: 31798604-31798604 on Build GRCh38
rs544242508 Chr.1: 31798685-31798685 on Build GRCh38
rs529578656 Chr.1: 31798261-31798261 on Build GRCh38
rs1052660245 Chr.1: 31798544-31798544 on Build GRCh38
rs763900089 Chr.1: 31798348-31798348 on Build GRCh38
rs199645489 Chr.1: 31798271-31798271 on Build GRCh38
rs372884096 Chr.1: 31798226-31798226 on Build GRCh38
rs749612579 Chr.1: 31798385-31798385 on Build GRCh38
rs778097743 Chr.1: 31798377-31798377 on Build GRCh38
rs867871798 Chr.1: 31798301-31798301 on Build GRCh38
rs773978316 Chr.1: 31798258-31798258 on Build GRCh38
rs745910950 Chr.1: 31798356-31798356 on Build GRCh38
rs762421614 Chr.1: 31798351-31798351 on Build GRCh38
rs757160383 Chr.1: 31798281-31798281 on Build GRCh38
rs767146196 Chr.1: 31798588-31798605 on Build GRCh38
rs551577268 Chr.1: 31798337-31798337 on Build GRCh38
rs779450335 Chr.1: 31798592-31798592 on Build GRCh38
rs748326329 Chr.1: 31798289-31798289 on Build GRCh38
rs746492945 Chr.1: 31798606-31798606 on Build GRCh38
rs566256359 Chr.1: 31798326-31798326 on Build GRCh38
rs766223074 Chr.1: 31798632-31798632 on Build GRCh38
rs766391914 Chr.1: 31798498-31798498 on Build GRCh38
rs758980545 Chr.1: 31798346-31798346 on Build GRCh38
rs189437304 Chr.1: 31798331-31798331 on Build GRCh38
rs764236418 Chr.1: 31798653-31798653 on Build GRCh38
rs552781397 Chr.1: 31798545-31798545 on Build GRCh38
rs141184600 Chr.1: 31798373-31798373 on Build GRCh38
rs935329334 Chr.1: 31798458-31798458 on Build GRCh38
rs1046125201 Chr.1: 31798533-31798533 on Build GRCh38
rs773705475 Chr.1: 31798235-31798235 on Build GRCh38
rs760530160 Chr.1: 31798640-31798640 on Build GRCh38
rs925270519 Chr.1: 31798453-31798453 on Build GRCh38
rs762904016 Chr.1: 31798496-31798496 on Build GRCh38
rs760277922 Chr.1: 31798414-31798414 on Build GRCh38
rs766709111 Chr.1: 31798608-31798608 on Build GRCh38
rs769151411 Chr.1: 31798272-31798272 on Build GRCh38
rs769860002 Chr.1: 31798467-31798467 on Build GRCh38
rs752454097 Chr.1: 31798262-31798262 on Build GRCh38
rs753769375 Chr.1: 31798273-31798273 on Build GRCh38
rs775038735 Chr.1: 31798623-31798623 on Build GRCh38
rs772973689 Chr.1: 31798627-31798627 on Build GRCh38
rs949012925 Chr.1: 31798480-31798480 on Build GRCh38
rs765605637 Chr.1: 31798664-31798664 on Build GRCh38
rs773844459 Chr.1: 31798272-31798273 on Build GRCh38
rs74065946 Chr.1: 31798415-31798415 on Build GRCh38
rs201900130 Chr.1: 31798622-31798622 on Build GRCh38
rs796645743 Chr.1: 31798683-31798683 on Build GRCh38
rs770069982 Chr.1: 31798232-31798232 on Build GRCh38
rs763404858 Chr.1: 31798239-31798239 on Build GRCh38
rs200928438 Chr.1: 31798312-31798312 on Build GRCh38
rs771720168 Chr.1: 31798329-31798329 on Build GRCh38
rs976716102 Chr.1: 31798646-31798646 on Build GRCh38
rs958528971 Chr.1: 31798327-31798327 on Build GRCh38
rs79366319 Chr.1: 31798623-31798623 on Build GRCh38

View Full Product Details