Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29628932-29629437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSH2
Entrez Gene ID
85464
Gene Name
slingshot protein phosphatase 2
Gene Aliases
SSH-2, SSH-2L
UniGene
Hs.654754
Gene Chromosome Location
Chr.17: 29625938-29930239 on Build GRCh38
Transcripts
NM_001282129.1 NM_033389.3 XM_005258058.3 XM_005258059.2 XM_005258060.3 XM_006722149.3 XM_011525402.2 XM_011525404.2 XM_011525405.2 XM_011525406.2 XM_011525407.1 XM_017025251.1 XM_017025252.1

SNP Detail

SNP ID Chromosome Location
rs900055883 Chr.17: 29629409-29629409 on Build GRCh38
rs536711903 Chr.17: 29629057-29629057 on Build GRCh38
rs746769378 Chr.17: 29629086-29629086 on Build GRCh38
rs999608792 Chr.17: 29629298-29629298 on Build GRCh38
rs75383266 Chr.17: 29629084-29629084 on Build GRCh38
rs1041261876 Chr.17: 29629396-29629396 on Build GRCh38
rs772016620 Chr.17: 29628952-29628952 on Build GRCh38
rs769719540 Chr.17: 29629354-29629354 on Build GRCh38
rs771453158 Chr.17: 29629213-29629213 on Build GRCh38
rs890527895 Chr.17: 29629185-29629185 on Build GRCh38
rs190949140 Chr.17: 29629269-29629269 on Build GRCh38
rs769066904 Chr.17: 29629176-29629176 on Build GRCh38
rs929317748 Chr.17: 29629177-29629177 on Build GRCh38
rs948992291 Chr.17: 29629100-29629100 on Build GRCh38
rs761956316 Chr.17: 29629287-29629287 on Build GRCh38
rs1022636544 Chr.17: 29629246-29629246 on Build GRCh38
rs1034710634 Chr.17: 29629391-29629391 on Build GRCh38
rs1008398475 Chr.17: 29629363-29629363 on Build GRCh38
rs532083280 Chr.17: 29629040-29629040 on Build GRCh38
rs887286659 Chr.17: 29629026-29629026 on Build GRCh38
rs568530876 Chr.17: 29629208-29629208 on Build GRCh38
rs1021314574 Chr.17: 29629068-29629068 on Build GRCh38
rs76248435 Chr.17: 29629085-29629085 on Build GRCh38
rs948669760 Chr.17: 29629276-29629276 on Build GRCh38
rs931350392 Chr.17: 29629307-29629307 on Build GRCh38
rs139692577 Chr.17: 29629346-29629347 on Build GRCh38
rs1021953666 Chr.17: 29629022-29629022 on Build GRCh38
rs968849447 Chr.17: 29629113-29629113 on Build GRCh38
rs535817848 Chr.17: 29629241-29629241 on Build GRCh38
rs977199458 Chr.17: 29629172-29629172 on Build GRCh38
rs992658144 Chr.17: 29629199-29629199 on Build GRCh38
rs140585793 Chr.17: 29629032-29629032 on Build GRCh38
rs1038841553 Chr.17: 29629194-29629194 on Build GRCh38
rs1047776859 Chr.17: 29629184-29629184 on Build GRCh38
rs1047150788 Chr.17: 29629340-29629341 on Build GRCh38
rs974073964 Chr.17: 29629167-29629167 on Build GRCh38
rs944817707 Chr.17: 29629190-29629190 on Build GRCh38
rs186091359 Chr.17: 29629056-29629056 on Build GRCh38
rs547204622 Chr.17: 29628988-29628988 on Build GRCh38
rs916103545 Chr.17: 29629094-29629094 on Build GRCh38
rs1034784464 Chr.17: 29629043-29629043 on Build GRCh38
rs992981087 Chr.17: 29629058-29629058 on Build GRCh38

View Full Product Details