Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18245846-18246346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLII
Entrez Gene ID
2314
Gene Name
FLII, actin remodeling protein
Gene Aliases
FLI, FLIL, Fli1
UniGene
Hs.513984
Gene Chromosome Location
Chr.17: 18244815-18258916 on Build GRCh38
Transcripts
NM_001256264.1 NM_001256265.1 NM_002018.3 XM_005256555.3 XM_005256556.3 XM_005256558.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4543201 c.3334G>A p.D1112N Chr.17: 18245996-18245996 on Build GRCh38
436151 c.3337G>A p.E1113K Chr.17: 18245993-18245993 on Build GRCh38
7297077 c.3320G>A p.R1107Q Chr.17: 18246010-18246010 on Build GRCh38
2740890 c.3187G>A p.G1063S Chr.17: 18246327-18246327 on Build GRCh38
7278889 c.3396+10C>T p.? Chr.17: 18245924-18245924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756884511 Chr.17: 18245873-18245873 on Build GRCh38
rs965243241 Chr.17: 18246051-18246051 on Build GRCh38
rs766268755 Chr.17: 18246085-18246085 on Build GRCh38
rs202127618 Chr.17: 18245920-18245920 on Build GRCh38
rs968405712 Chr.17: 18245907-18245907 on Build GRCh38
rs778678110 Chr.17: 18245876-18245876 on Build GRCh38
rs773745535 Chr.17: 18245894-18245894 on Build GRCh38
rs776807517 Chr.17: 18246134-18246134 on Build GRCh38
rs778026726 Chr.17: 18246305-18246305 on Build GRCh38
rs747246238 Chr.17: 18246129-18246129 on Build GRCh38
rs777228781 Chr.17: 18245942-18245942 on Build GRCh38
rs368493265 Chr.17: 18246186-18246186 on Build GRCh38
rs373302992 Chr.17: 18245869-18245869 on Build GRCh38
rs765097929 Chr.17: 18246008-18246008 on Build GRCh38
rs996732165 Chr.17: 18246052-18246052 on Build GRCh38
rs147718630 Chr.17: 18246204-18246204 on Build GRCh38
rs530570572 Chr.17: 18246124-18246124 on Build GRCh38
rs368988400 Chr.17: 18246276-18246276 on Build GRCh38
rs763969004 Chr.17: 18245926-18245926 on Build GRCh38
rs781321061 Chr.17: 18246195-18246195 on Build GRCh38
rs777419957 Chr.17: 18245916-18245923 on Build GRCh38
rs759939609 Chr.17: 18246164-18246164 on Build GRCh38
rs878998412 Chr.17: 18246119-18246119 on Build GRCh38
rs754614930 Chr.17: 18246089-18246089 on Build GRCh38
rs769898644 Chr.17: 18246139-18246139 on Build GRCh38
rs770449087 Chr.17: 18245955-18245955 on Build GRCh38
rs758412355 Chr.17: 18246183-18246183 on Build GRCh38
rs201173622 Chr.17: 18245993-18245993 on Build GRCh38
rs1048370552 Chr.17: 18246256-18246256 on Build GRCh38
rs749811658 Chr.17: 18246042-18246042 on Build GRCh38
rs145565671 Chr.17: 18246013-18246013 on Build GRCh38
rs770947928 Chr.17: 18246245-18246245 on Build GRCh38
rs537633078 Chr.17: 18246167-18246167 on Build GRCh38
rs773892887 Chr.17: 18245956-18245956 on Build GRCh38
rs751223663 Chr.17: 18246019-18246019 on Build GRCh38
rs968545443 Chr.17: 18246070-18246070 on Build GRCh38
rs745480480 Chr.17: 18245878-18245878 on Build GRCh38
rs1036571647 Chr.17: 18246079-18246079 on Build GRCh38
rs766286847 Chr.17: 18246143-18246143 on Build GRCh38
rs370659231 Chr.17: 18245900-18245900 on Build GRCh38
rs536232194 Chr.17: 18246201-18246201 on Build GRCh38
rs539123081 Chr.17: 18246283-18246283 on Build GRCh38
rs145877855 Chr.17: 18245979-18245979 on Build GRCh38
rs746481063 Chr.17: 18245950-18245950 on Build GRCh38
rs745592481 Chr.17: 18245943-18245943 on Build GRCh38
rs770627473 Chr.17: 18245899-18245901 on Build GRCh38
rs905164512 Chr.17: 18246106-18246106 on Build GRCh38
rs200427238 Chr.17: 18245940-18245940 on Build GRCh38
rs773365321 Chr.17: 18246222-18246222 on Build GRCh38
rs748981447 Chr.17: 18245898-18245899 on Build GRCh38
rs765375716 Chr.17: 18246069-18246070 on Build GRCh38
rs746168467 Chr.17: 18246047-18246047 on Build GRCh38
rs552950931 Chr.17: 18246285-18246285 on Build GRCh38
rs771111506 Chr.17: 18246319-18246319 on Build GRCh38
rs773087341 Chr.17: 18246140-18246140 on Build GRCh38
rs368003319 Chr.17: 18245919-18245919 on Build GRCh38
rs373074842 Chr.17: 18246145-18246145 on Build GRCh38
rs760364052 Chr.17: 18246274-18246274 on Build GRCh38
rs374964844 Chr.17: 18246092-18246092 on Build GRCh38
rs774309595 Chr.17: 18246255-18246255 on Build GRCh38
rs758844825 Chr.17: 18246182-18246182 on Build GRCh38
rs368808829 Chr.17: 18246102-18246102 on Build GRCh38
rs758158947 Chr.17: 18246011-18246011 on Build GRCh38
rs374580964 Chr.17: 18246168-18246168 on Build GRCh38
rs1028235007 Chr.17: 18246322-18246322 on Build GRCh38
rs768803683 Chr.17: 18246133-18246133 on Build GRCh38
rs1008866754 Chr.17: 18246046-18246046 on Build GRCh38
rs768783137 Chr.17: 18246054-18246054 on Build GRCh38
rs750528799 Chr.17: 18246143-18246155 on Build GRCh38
rs899362605 Chr.17: 18245975-18245975 on Build GRCh38
rs200244752 Chr.17: 18245871-18245871 on Build GRCh38
rs779806523 Chr.17: 18246016-18246016 on Build GRCh38
rs369231938 Chr.17: 18246057-18246057 on Build GRCh38
rs377248019 Chr.17: 18246147-18246147 on Build GRCh38
rs1027689592 Chr.17: 18245902-18245902 on Build GRCh38
rs757934636 Chr.17: 18245947-18245947 on Build GRCh38
rs376249873 Chr.17: 18246037-18246037 on Build GRCh38
rs753698988 Chr.17: 18245931-18245931 on Build GRCh38
rs780754417 Chr.17: 18246091-18246091 on Build GRCh38
rs930834131 Chr.17: 18246206-18246206 on Build GRCh38
rs142476596 Chr.17: 18246318-18246318 on Build GRCh38
rs965687154 Chr.17: 18245881-18245881 on Build GRCh38
rs762161279 Chr.17: 18245959-18245961 on Build GRCh38
rs750256946 Chr.17: 18246068-18246068 on Build GRCh38
rs375540650 Chr.17: 18246228-18246228 on Build GRCh38
rs764129085 Chr.17: 18245999-18245999 on Build GRCh38
rs1014036905 Chr.17: 18246132-18246132 on Build GRCh38
rs772240221 Chr.17: 18245988-18245988 on Build GRCh38
rs372788424 Chr.17: 18245959-18245959 on Build GRCh38
rs200315491 Chr.17: 18245923-18245923 on Build GRCh38
rs552545157 Chr.17: 18246022-18246022 on Build GRCh38
rs117461683 Chr.17: 18245917-18245917 on Build GRCh38
rs746345073 Chr.17: 18246115-18246115 on Build GRCh38
rs753279393 Chr.17: 18246279-18246279 on Build GRCh38
rs748328577 Chr.17: 18246297-18246297 on Build GRCh38
rs752042607 Chr.17: 18246290-18246290 on Build GRCh38
rs11540886 Chr.17: 18245994-18245994 on Build GRCh38
rs756287955 Chr.17: 18246300-18246300 on Build GRCh38
rs377168924 Chr.17: 18245969-18245969 on Build GRCh38
rs765050095 Chr.17: 18246067-18246067 on Build GRCh38
rs372234480 Chr.17: 18245883-18245883 on Build GRCh38
rs779718707 Chr.17: 18245948-18245948 on Build GRCh38
rs879450744 Chr.17: 18246142-18246142 on Build GRCh38
rs371022314 Chr.17: 18245868-18245868 on Build GRCh38
rs762968922 Chr.17: 18246141-18246141 on Build GRCh38
rs1028099307 Chr.17: 18246062-18246062 on Build GRCh38
rs757117066 Chr.17: 18245932-18245932 on Build GRCh38
rs376365504 Chr.17: 18246138-18246138 on Build GRCh38
rs775131293 Chr.17: 18245884-18245884 on Build GRCh38
rs948940005 Chr.17: 18246161-18246161 on Build GRCh38
rs767027995 Chr.17: 18246165-18246165 on Build GRCh38
rs370490464 Chr.17: 18245924-18245924 on Build GRCh38
rs1004517524 Chr.17: 18245870-18245870 on Build GRCh38
rs772494602 Chr.17: 18246120-18246120 on Build GRCh38
rs780356301 Chr.17: 18246194-18246194 on Build GRCh38
rs762893389 Chr.17: 18246069-18246069 on Build GRCh38
rs766618137 Chr.17: 18245911-18245911 on Build GRCh38
rs760884592 Chr.17: 18246053-18246053 on Build GRCh38
rs779336446 Chr.17: 18246111-18246111 on Build GRCh38
rs759452097 Chr.17: 18246263-18246263 on Build GRCh38
rs773988104 Chr.17: 18245901-18245902 on Build GRCh38
rs199790278 Chr.17: 18246271-18246271 on Build GRCh38
rs780551347 Chr.17: 18246029-18246029 on Build GRCh38
rs57694884 Chr.17: 18245946-18245946 on Build GRCh38
rs376224540 Chr.17: 18245890-18245890 on Build GRCh38
rs771275856 Chr.17: 18246045-18246045 on Build GRCh38
rs748374225 Chr.17: 18246202-18246202 on Build GRCh38
rs868073977 Chr.17: 18246203-18246203 on Build GRCh38
rs897904178 Chr.17: 18246084-18246084 on Build GRCh38
rs200651341 Chr.17: 18246157-18246157 on Build GRCh38
rs886486799 Chr.17: 18246260-18246260 on Build GRCh38
rs900539410 Chr.17: 18246288-18246288 on Build GRCh38
rs751311736 Chr.17: 18246087-18246087 on Build GRCh38
rs199619176 Chr.17: 18246049-18246049 on Build GRCh38
rs376192838 Chr.17: 18246177-18246177 on Build GRCh38
rs367896113 Chr.17: 18246034-18246034 on Build GRCh38
rs774697171 Chr.17: 18245912-18245912 on Build GRCh38
rs772401250 Chr.17: 18246048-18246048 on Build GRCh38
rs781604192 Chr.17: 18246296-18246296 on Build GRCh38
rs761643815 Chr.17: 18246000-18246000 on Build GRCh38
rs1019556147 Chr.17: 18245879-18245879 on Build GRCh38
rs372274459 Chr.17: 18246010-18246010 on Build GRCh38
rs368339121 Chr.17: 18245986-18245986 on Build GRCh38
rs372477896 Chr.17: 18246293-18246293 on Build GRCh38
rs747337921 Chr.17: 18246193-18246193 on Build GRCh38
rs749331004 Chr.17: 18246316-18246316 on Build GRCh38
rs201849922 Chr.17: 18245907-18245914 on Build GRCh38
rs966316821 Chr.17: 18246207-18246207 on Build GRCh38
rs1019990965 Chr.17: 18245974-18245974 on Build GRCh38
rs374888370 Chr.17: 18246273-18246273 on Build GRCh38

View Full Product Details