Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21198262-21198772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM11
Entrez Gene ID
8834
Gene Name
transmembrane protein 11
Gene Aliases
C17orf35, PM1, PMI
UniGene
Hs.592945
Gene Chromosome Location
Chr.17: 21197950-21214595 on Build GRCh38
Transcripts
NM_003876.2 NR_024547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253294 c.396C>A p.C132* Chr.17: 21198507-21198507 on Build GRCh38
1679648 c.205G>A p.D69N Chr.17: 21198698-21198698 on Build GRCh38
247860 c.518C>T p.T173M Chr.17: 21198385-21198385 on Build GRCh38
6343434 c.255C>A p.A85A Chr.17: 21198648-21198648 on Build GRCh38
264076 c.562G>A p.E188K Chr.17: 21198341-21198341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765996667 Chr.17: 21198384-21198384 on Build GRCh38
rs769358201 Chr.17: 21198633-21198633 on Build GRCh38
rs142814599 Chr.17: 21198630-21198630 on Build GRCh38
rs541612728 Chr.17: 21198329-21198329 on Build GRCh38
rs150424039 Chr.17: 21198414-21198414 on Build GRCh38
rs779261029 Chr.17: 21198610-21198610 on Build GRCh38
rs771406829 Chr.17: 21198706-21198706 on Build GRCh38
rs760465328 Chr.17: 21198657-21198657 on Build GRCh38
rs748836900 Chr.17: 21198717-21198717 on Build GRCh38
rs752258502 Chr.17: 21198432-21198432 on Build GRCh38
rs150188248 Chr.17: 21198690-21198690 on Build GRCh38
rs953547825 Chr.17: 21198629-21198629 on Build GRCh38
rs9906188 Chr.17: 21198467-21198467 on Build GRCh38
rs899963166 Chr.17: 21198522-21198522 on Build GRCh38
rs371720143 Chr.17: 21198498-21198498 on Build GRCh38
rs368267363 Chr.17: 21198305-21198305 on Build GRCh38
rs767169112 Chr.17: 21198651-21198651 on Build GRCh38
rs1053752985 Chr.17: 21198675-21198675 on Build GRCh38
rs138107180 Chr.17: 21198699-21198699 on Build GRCh38
rs982399201 Chr.17: 21198472-21198472 on Build GRCh38
rs373207736 Chr.17: 21198615-21198615 on Build GRCh38
rs755322877 Chr.17: 21198609-21198609 on Build GRCh38
rs983735705 Chr.17: 21198481-21198481 on Build GRCh38
rs780978717 Chr.17: 21198453-21198453 on Build GRCh38
rs746010807 Chr.17: 21198459-21198459 on Build GRCh38
rs754121840 Chr.17: 21198602-21198602 on Build GRCh38
rs749474774 Chr.17: 21198471-21198471 on Build GRCh38
rs764987344 Chr.17: 21198431-21198431 on Build GRCh38
rs752591957 Chr.17: 21198300-21198300 on Build GRCh38
rs772688448 Chr.17: 21198486-21198486 on Build GRCh38
rs200292766 Chr.17: 21198485-21198485 on Build GRCh38
rs145501473 Chr.17: 21198665-21198666 on Build GRCh38
rs763880962 Chr.17: 21198557-21198557 on Build GRCh38
rs777725531 Chr.17: 21198320-21198320 on Build GRCh38
rs138310111 Chr.17: 21198601-21198601 on Build GRCh38
rs759123130 Chr.17: 21198489-21198489 on Build GRCh38
rs34336938 Chr.17: 21198346-21198346 on Build GRCh38
rs970114538 Chr.17: 21198405-21198405 on Build GRCh38
rs754034119 Chr.17: 21198674-21198674 on Build GRCh38
rs770113085 Chr.17: 21198334-21198334 on Build GRCh38
rs895841183 Chr.17: 21198621-21198621 on Build GRCh38
rs7212548 Chr.17: 21198290-21198290 on Build GRCh38
rs369634383 Chr.17: 21198639-21198639 on Build GRCh38
rs780357624 Chr.17: 21198469-21198469 on Build GRCh38
rs761700711 Chr.17: 21198648-21198648 on Build GRCh38
rs753450239 Chr.17: 21198287-21198287 on Build GRCh38
rs202195850 Chr.17: 21198632-21198632 on Build GRCh38
rs750289855 Chr.17: 21198593-21198593 on Build GRCh38
rs527761394 Chr.17: 21198371-21198371 on Build GRCh38
rs376685777 Chr.17: 21198422-21198422 on Build GRCh38
rs201549323 Chr.17: 21198723-21198723 on Build GRCh38
rs367895918 Chr.17: 21198289-21198289 on Build GRCh38
rs751510486 Chr.17: 21198441-21198441 on Build GRCh38
rs762532851 Chr.17: 21198533-21198533 on Build GRCh38
rs772289833 Chr.17: 21198624-21198624 on Build GRCh38
rs758718626 Chr.17: 21198630-21198630 on Build GRCh38
rs373174342 Chr.17: 21198652-21198652 on Build GRCh38
rs372525354 Chr.17: 21198330-21198330 on Build GRCh38
rs775552645 Chr.17: 21198341-21198341 on Build GRCh38
rs1023866840 Chr.17: 21198407-21198407 on Build GRCh38
rs140648624 Chr.17: 21198447-21198447 on Build GRCh38
rs564469579 Chr.17: 21198435-21198435 on Build GRCh38
rs900014117 Chr.17: 21198310-21198310 on Build GRCh38
rs755082002 Chr.17: 21198685-21198685 on Build GRCh38
rs376181647 Chr.17: 21198707-21198707 on Build GRCh38
rs750434978 Chr.17: 21198295-21198296 on Build GRCh38
rs773961880 Chr.17: 21198647-21198647 on Build GRCh38
rs1024426649 Chr.17: 21198385-21198385 on Build GRCh38
rs200494053 Chr.17: 21198696-21198696 on Build GRCh38
rs747554727 Chr.17: 21198701-21198701 on Build GRCh38
rs780021433 Chr.17: 21198688-21198688 on Build GRCh38
rs746907195 Chr.17: 21198327-21198327 on Build GRCh38
rs775144090 Chr.17: 21198509-21198509 on Build GRCh38
rs773566259 Chr.17: 21198488-21198488 on Build GRCh38
rs891918221 Chr.17: 21198364-21198364 on Build GRCh38
rs756242577 Chr.17: 21198598-21198598 on Build GRCh38
rs1012211259 Chr.17: 21198326-21198326 on Build GRCh38
rs761573656 Chr.17: 21198573-21198573 on Build GRCh38
rs757380574 Chr.17: 21198421-21198421 on Build GRCh38
rs969261920 Chr.17: 21198408-21198408 on Build GRCh38
rs766340376 Chr.17: 21198659-21198659 on Build GRCh38
rs944513458 Chr.17: 21198552-21198552 on Build GRCh38
rs200018147 Chr.17: 21198572-21198572 on Build GRCh38
rs745956055 Chr.17: 21198450-21198450 on Build GRCh38
rs185572860 Chr.17: 21198635-21198635 on Build GRCh38
rs762355957 Chr.17: 21198372-21198372 on Build GRCh38
rs758810977 Chr.17: 21198698-21198698 on Build GRCh38

View Full Product Details