Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44248810-44249326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC4A1
Entrez Gene ID
6521
Gene Name
solute carrier family 4 member 1 (Diego blood group)
Gene Aliases
AE1, BND3, CD233, CHC, DI, EMPB3, EPB3, FR, RTA1A, SAO, SPH4, SW, WD, WD1, WR
UniGene
Hs.210751
Gene Chromosome Location
Chr.17: 44248390-44268161 on Build GRCh38
Transcripts
NM_000342.3 XM_005257593.4 XM_011525129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4388131 c.86G>A p.G29E Chr.17: 44249119-44249119 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918321294 Chr.17: 44249260-44249260 on Build GRCh38
rs5033 Chr.17: 44248890-44248890 on Build GRCh38
rs903557040 Chr.17: 44248899-44248899 on Build GRCh38
rs891330487 Chr.17: 44249245-44249245 on Build GRCh38
rs886052991 Chr.17: 44249142-44249142 on Build GRCh38
rs1047987691 Chr.17: 44249043-44249043 on Build GRCh38
rs1010826661 Chr.17: 44249014-44249014 on Build GRCh38
rs1029872400 Chr.17: 44249045-44249045 on Build GRCh38
rs1029546540 Chr.17: 44248850-44248865 on Build GRCh38
rs12945748 Chr.17: 44248845-44248845 on Build GRCh38
rs1007173680 Chr.17: 44249053-44249053 on Build GRCh38
rs770415235 Chr.17: 44249122-44249122 on Build GRCh38
rs941148131 Chr.17: 44248864-44248864 on Build GRCh38
rs917772299 Chr.17: 44249124-44249124 on Build GRCh38
rs869183959 Chr.17: 44248877-44248878 on Build GRCh38
rs57466226 Chr.17: 44248877-44248877 on Build GRCh38
rs992050160 Chr.17: 44248857-44248857 on Build GRCh38
rs1033064799 Chr.17: 44249127-44249127 on Build GRCh38
rs974235517 Chr.17: 44249249-44249249 on Build GRCh38
rs948012092 Chr.17: 44248862-44248862 on Build GRCh38
rs749729080 Chr.17: 44249104-44249104 on Build GRCh38
rs1039193295 Chr.17: 44248849-44248849 on Build GRCh38
rs769140134 Chr.17: 44249185-44249185 on Build GRCh38
rs548849419 Chr.17: 44249180-44249180 on Build GRCh38
rs955205649 Chr.17: 44248854-44248854 on Build GRCh38
rs2857079 Chr.17: 44248840-44248840 on Build GRCh38
rs980667176 Chr.17: 44249146-44249146 on Build GRCh38
rs936404213 Chr.17: 44248929-44248929 on Build GRCh38
rs1052630095 Chr.17: 44248942-44248942 on Build GRCh38
rs973844054 Chr.17: 44248864-44248865 on Build GRCh38
rs1044979647 Chr.17: 44249093-44249093 on Build GRCh38
rs566138996 Chr.17: 44249153-44249153 on Build GRCh38
rs774513767 Chr.17: 44249221-44249221 on Build GRCh38
rs886052992 Chr.17: 44249260-44249260 on Build GRCh38
rs886052989 Chr.17: 44248850-44248850 on Build GRCh38
rs5032 Chr.17: 44249027-44249027 on Build GRCh38
rs891382961 Chr.17: 44249049-44249049 on Build GRCh38
rs891920821 Chr.17: 44248946-44248946 on Build GRCh38
rs1039998603 Chr.17: 44249265-44249265 on Build GRCh38
rs776118982 Chr.17: 44248976-44248976 on Build GRCh38
rs991812540 Chr.17: 44249279-44249279 on Build GRCh38
rs891803423 Chr.17: 44249145-44249145 on Build GRCh38
rs1007121116 Chr.17: 44249263-44249263 on Build GRCh38
rs139308660 Chr.17: 44249065-44249065 on Build GRCh38
rs962718832 Chr.17: 44249068-44249068 on Build GRCh38
rs142587809 Chr.17: 44249162-44249162 on Build GRCh38
rs936334860 Chr.17: 44249163-44249163 on Build GRCh38
rs141425539 Chr.17: 44249231-44249231 on Build GRCh38
rs929817457 Chr.17: 44248856-44248856 on Build GRCh38
rs1026153044 Chr.17: 44249114-44249114 on Build GRCh38
rs12945753 Chr.17: 44248850-44248850 on Build GRCh38
rs906028021 Chr.17: 44249101-44249101 on Build GRCh38
rs71361565 Chr.17: 44248849-44248850 on Build GRCh38
rs535702536 Chr.17: 44249261-44249261 on Build GRCh38
rs969263666 Chr.17: 44249142-44249142 on Build GRCh38
rs772978455 Chr.17: 44249195-44249195 on Build GRCh38
rs768606768 Chr.17: 44249185-44249185 on Build GRCh38
rs918256663 Chr.17: 44248919-44248919 on Build GRCh38
rs550914663 Chr.17: 44249076-44249076 on Build GRCh38
rs886052990 Chr.17: 44248862-44248862 on Build GRCh38
rs929607534 Chr.17: 44248947-44248947 on Build GRCh38
rs951529200 Chr.17: 44249117-44249117 on Build GRCh38
rs955299700 Chr.17: 44249214-44249214 on Build GRCh38
rs898456645 Chr.17: 44249278-44249278 on Build GRCh38
rs995921132 Chr.17: 44249301-44249301 on Build GRCh38
rs917824715 Chr.17: 44248860-44248860 on Build GRCh38
rs533631187 Chr.17: 44249156-44249156 on Build GRCh38

View Full Product Details