Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44353971-44354461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM171A2
Entrez Gene ID
284069
Gene Name
family with sequence similarity 171 member A2
Gene Aliases
-
UniGene
Hs.154001
Gene Chromosome Location
Chr.17: 44353215-44363867 on Build GRCh38
Transcripts
NM_198475.2 XM_017024490.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741013 c.1657A>C p.S553R Chr.17: 44354005-44354005 on Build GRCh38
7259052 c.1264delG p.A422fs*32 Chr.17: 44354398-44354398 on Build GRCh38
4792182 c.1368G>A p.K456K Chr.17: 44354294-44354294 on Build GRCh38
6025928 c.1496A>G p.D499G Chr.17: 44354166-44354166 on Build GRCh38
5015478 c.1456T>A p.Y486N Chr.17: 44354206-44354206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016901078 Chr.17: 44354438-44354438 on Build GRCh38
rs989638324 Chr.17: 44354086-44354086 on Build GRCh38
rs934086781 Chr.17: 44354348-44354348 on Build GRCh38
rs998245032 Chr.17: 44353997-44353997 on Build GRCh38
rs766063703 Chr.17: 44354283-44354283 on Build GRCh38
rs754863727 Chr.17: 44354301-44354301 on Build GRCh38
rs750274552 Chr.17: 44354174-44354174 on Build GRCh38
rs528777079 Chr.17: 44354279-44354279 on Build GRCh38
rs889764934 Chr.17: 44354043-44354043 on Build GRCh38
rs778539378 Chr.17: 44354342-44354342 on Build GRCh38
rs953769134 Chr.17: 44354046-44354047 on Build GRCh38
rs990546758 Chr.17: 44354099-44354099 on Build GRCh38
rs934886454 Chr.17: 44354263-44354263 on Build GRCh38
rs1005537151 Chr.17: 44354433-44354433 on Build GRCh38
rs752086752 Chr.17: 44354431-44354431 on Build GRCh38
rs758054480 Chr.17: 44354439-44354439 on Build GRCh38
rs1034809097 Chr.17: 44354075-44354083 on Build GRCh38
rs1049810183 Chr.17: 44354421-44354421 on Build GRCh38
rs140882895 Chr.17: 44354156-44354156 on Build GRCh38
rs985174879 Chr.17: 44354073-44354084 on Build GRCh38
rs1053690631 Chr.17: 44354331-44354331 on Build GRCh38
rs1028743424 Chr.17: 44354040-44354040 on Build GRCh38
rs889818638 Chr.17: 44354429-44354429 on Build GRCh38
rs1005483249 Chr.17: 44354046-44354046 on Build GRCh38
rs961223614 Chr.17: 44354057-44354058 on Build GRCh38
rs926192720 Chr.17: 44354200-44354200 on Build GRCh38
rs946357314 Chr.17: 44354183-44354183 on Build GRCh38
rs960745701 Chr.17: 44354082-44354082 on Build GRCh38
rs753852041 Chr.17: 44354296-44354296 on Build GRCh38
rs897066539 Chr.17: 44354442-44354442 on Build GRCh38
rs979013688 Chr.17: 44354185-44354185 on Build GRCh38

View Full Product Details