Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44803769-44804283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJC1
Entrez Gene ID
10052
Gene Name
gap junction protein gamma 1
Gene Aliases
CX45, GJA7
UniGene
Hs.712052
Gene Chromosome Location
Chr.17: 44794988-44830811 on Build GRCh38
Transcripts
NM_001080383.1 NM_005497.3 XM_005256920.1 XM_005256921.1

SNP Detail

SNP ID Chromosome Location
rs1017510560 Chr.17: 44803882-44803882 on Build GRCh38
rs1026781810 Chr.17: 44804178-44804178 on Build GRCh38
rs936944136 Chr.17: 44803951-44803951 on Build GRCh38
rs535660454 Chr.17: 44804249-44804249 on Build GRCh38
rs1050498292 Chr.17: 44804230-44804230 on Build GRCh38
rs968366789 Chr.17: 44804019-44804019 on Build GRCh38
rs1050066416 Chr.17: 44803835-44803835 on Build GRCh38
rs959233586 Chr.17: 44804197-44804202 on Build GRCh38
rs890023852 Chr.17: 44803865-44803865 on Build GRCh38
rs1012529307 Chr.17: 44804216-44804216 on Build GRCh38
rs531096708 Chr.17: 44803988-44803988 on Build GRCh38
rs56752018 Chr.17: 44804072-44804072 on Build GRCh38
rs961532409 Chr.17: 44803913-44803913 on Build GRCh38
rs562815597 Chr.17: 44804123-44804123 on Build GRCh38
rs568428612 Chr.17: 44804233-44804233 on Build GRCh38
rs61586130 Chr.17: 44803987-44803987 on Build GRCh38
rs778089708 Chr.17: 44804006-44804006 on Build GRCh38
rs754160132 Chr.17: 44803990-44803990 on Build GRCh38
rs1035077244 Chr.17: 44803978-44803978 on Build GRCh38
rs1005827022 Chr.17: 44803874-44803874 on Build GRCh38
rs184671332 Chr.17: 44804170-44804170 on Build GRCh38
rs979408463 Chr.17: 44804039-44804039 on Build GRCh38
rs924177637 Chr.17: 44803928-44803928 on Build GRCh38
rs61143193 Chr.17: 44804111-44804111 on Build GRCh38
rs935128515 Chr.17: 44804080-44804080 on Build GRCh38
rs990806153 Chr.17: 44803994-44803994 on Build GRCh38
rs59881417 Chr.17: 44803829-44803829 on Build GRCh38
rs542026580 Chr.17: 44803977-44803977 on Build GRCh38
rs544507518 Chr.17: 44804102-44804102 on Build GRCh38
rs975906877 Chr.17: 44804091-44804091 on Build GRCh38
rs901604388 Chr.17: 44803819-44803819 on Build GRCh38
rs752948905 Chr.17: 44803853-44803853 on Build GRCh38
rs570511154 Chr.17: 44804110-44804110 on Build GRCh38
rs1036716512 Chr.17: 44804055-44804055 on Build GRCh38
rs111340099 Chr.17: 44803927-44803927 on Build GRCh38
rs1007879817 Chr.17: 44804041-44804043 on Build GRCh38

View Full Product Details