Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32274914-32275405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCK
Entrez Gene ID
3932
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
IMD22, LSK, YT16, p56lck, pp58lck
UniGene
Hs.470627
Gene Chromosome Location
Chr.1: 32251239-32286167 on Build GRCh38
Transcripts
NM_001042771.2 NM_005356.4 XM_011541453.2 XM_011541454.2 XM_011541455.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1687340 c.334C>T p.P112S Chr.1: 32275376-32275376 on Build GRCh38
5984885 c.444C>G p.I148M Chr.1: 32275075-32275075 on Build GRCh38
4990562 c.427G>A p.G143R Chr.1: 32275058-32275058 on Build GRCh38
2077859 c.396C>T p.P132P Chr.1: 32275027-32275027 on Build GRCh38
908281 c.285C>T p.G95G Chr.1: 32275327-32275327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373949317 Chr.1: 32275270-32275270 on Build GRCh38
rs578188691 Chr.1: 32275228-32275228 on Build GRCh38
rs869302318 Chr.1: 32275062-32275063 on Build GRCh38
rs770884558 Chr.1: 32275285-32275285 on Build GRCh38
rs758959352 Chr.1: 32274948-32274948 on Build GRCh38
rs866634163 Chr.1: 32275152-32275152 on Build GRCh38
rs771045740 Chr.1: 32275017-32275017 on Build GRCh38
rs11576028 Chr.1: 32275200-32275200 on Build GRCh38
rs1003681059 Chr.1: 32275140-32275140 on Build GRCh38
rs377149314 Chr.1: 32275024-32275024 on Build GRCh38
rs142674647 Chr.1: 32275323-32275323 on Build GRCh38
rs763255214 Chr.1: 32275311-32275311 on Build GRCh38
rs769566620 Chr.1: 32274997-32274997 on Build GRCh38
rs752391858 Chr.1: 32275131-32275131 on Build GRCh38
rs985734576 Chr.1: 32275267-32275267 on Build GRCh38
rs780848515 Chr.1: 32274994-32274994 on Build GRCh38
rs778932130 Chr.1: 32275287-32275287 on Build GRCh38
rs765619775 Chr.1: 32275045-32275045 on Build GRCh38
rs776842939 Chr.1: 32275036-32275036 on Build GRCh38
rs754479058 Chr.1: 32274976-32274976 on Build GRCh38
rs775683851 Chr.1: 32275020-32275020 on Build GRCh38
rs898148096 Chr.1: 32275109-32275109 on Build GRCh38
rs537077999 Chr.1: 32275168-32275168 on Build GRCh38
rs762072590 Chr.1: 32275301-32275301 on Build GRCh38
rs868846795 Chr.1: 32274950-32274950 on Build GRCh38
rs948436853 Chr.1: 32274964-32274964 on Build GRCh38
rs777692288 Chr.1: 32275282-32275282 on Build GRCh38
rs768627081 Chr.1: 32275027-32275027 on Build GRCh38
rs752083850 Chr.1: 32275123-32275123 on Build GRCh38
rs911037520 Chr.1: 32275331-32275331 on Build GRCh38
rs533969475 Chr.1: 32275049-32275049 on Build GRCh38
rs1035188114 Chr.1: 32275144-32275144 on Build GRCh38
rs375477745 Chr.1: 32275317-32275317 on Build GRCh38
rs762005602 Chr.1: 32275038-32275038 on Build GRCh38
rs890599892 Chr.1: 32275186-32275186 on Build GRCh38
rs869295456 Chr.1: 32275245-32275246 on Build GRCh38
rs993933791 Chr.1: 32275110-32275110 on Build GRCh38
rs780574086 Chr.1: 32275128-32275128 on Build GRCh38
rs192341595 Chr.1: 32274963-32274963 on Build GRCh38
rs561703751 Chr.1: 32275357-32275357 on Build GRCh38
rs554553133 Chr.1: 32275305-32275305 on Build GRCh38
rs749139199 Chr.1: 32275283-32275283 on Build GRCh38
rs556498364 Chr.1: 32275221-32275221 on Build GRCh38
rs959930352 Chr.1: 32275187-32275187 on Build GRCh38
rs567952276 Chr.1: 32275241-32275241 on Build GRCh38
rs1053218137 Chr.1: 32275158-32275158 on Build GRCh38
rs745794507 Chr.1: 32275288-32275288 on Build GRCh38
rs866986533 Chr.1: 32274969-32274969 on Build GRCh38
rs768775326 Chr.1: 32275291-32275291 on Build GRCh38
rs370547625 Chr.1: 32275127-32275127 on Build GRCh38
rs545721651 Chr.1: 32275264-32275264 on Build GRCh38
rs567679161 Chr.1: 32275180-32275180 on Build GRCh38
rs377653414 Chr.1: 32275325-32275325 on Build GRCh38
rs555714485 Chr.1: 32275080-32275080 on Build GRCh38
rs767043658 Chr.1: 32275330-32275330 on Build GRCh38
rs992551046 Chr.1: 32274962-32274962 on Build GRCh38
rs777499240 Chr.1: 32275007-32275007 on Build GRCh38
rs779517104 Chr.1: 32274957-32274957 on Build GRCh38
rs11576029 Chr.1: 32275240-32275240 on Build GRCh38
rs140817239 Chr.1: 32275191-32275191 on Build GRCh38
rs752201210 Chr.1: 32275348-32275348 on Build GRCh38
rs1051027573 Chr.1: 32274998-32274998 on Build GRCh38
rs776630706 Chr.1: 32275298-32275298 on Build GRCh38
rs150810327 Chr.1: 32275051-32275051 on Build GRCh38
rs749226778 Chr.1: 32275009-32275009 on Build GRCh38
rs886611410 Chr.1: 32275137-32275137 on Build GRCh38
rs745911605 Chr.1: 32275324-32275324 on Build GRCh38
rs942638932 Chr.1: 32275375-32275375 on Build GRCh38
rs766830352 Chr.1: 32275112-32275112 on Build GRCh38
rs1039145806 Chr.1: 32275214-32275214 on Build GRCh38
rs867588722 Chr.1: 32275376-32275376 on Build GRCh38
rs201899413 Chr.1: 32275336-32275336 on Build GRCh38
rs112957416 Chr.1: 32275353-32275353 on Build GRCh38
rs763419572 Chr.1: 32275090-32275090 on Build GRCh38
rs1038465388 Chr.1: 32275058-32275058 on Build GRCh38
rs991069317 Chr.1: 32275198-32275198 on Build GRCh38
rs1022754875 Chr.1: 32275349-32275349 on Build GRCh38
rs909706504 Chr.1: 32275290-32275290 on Build GRCh38
rs967998211 Chr.1: 32275249-32275249 on Build GRCh38
rs900565718 Chr.1: 32275234-32275234 on Build GRCh38

View Full Product Details