Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32365418-32365910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSDC1
Entrez Gene ID
55108
Gene Name
BSD domain containing 1
Gene Aliases
-
UniGene
Hs.353454
Gene Chromosome Location
Chr.1: 32364633-32394461 on Build GRCh38
Transcripts
NM_001143888.2 NM_001143889.2 NM_001143890.2 NM_001300958.1 NM_018045.7 NR_125352.1

SNP Detail

SNP ID Chromosome Location
rs1039400966 Chr.1: 32365669-32365669 on Build GRCh38
rs540940091 Chr.1: 32365676-32365676 on Build GRCh38
rs908812157 Chr.1: 32365493-32365493 on Build GRCh38
rs370217955 Chr.1: 32365591-32365591 on Build GRCh38
rs913746216 Chr.1: 32365832-32365846 on Build GRCh38
rs977409440 Chr.1: 32365883-32365883 on Build GRCh38
rs900151871 Chr.1: 32365647-32365647 on Build GRCh38
rs564700872 Chr.1: 32365572-32365572 on Build GRCh38
rs57929051 Chr.1: 32365465-32365465 on Build GRCh38
rs980077964 Chr.1: 32365833-32365833 on Build GRCh38
rs779571531 Chr.1: 32365657-32365657 on Build GRCh38
rs770256981 Chr.1: 32365759-32365759 on Build GRCh38
rs554276173 Chr.1: 32365508-32365508 on Build GRCh38
rs558290605 Chr.1: 32365867-32365867 on Build GRCh38
rs1044487157 Chr.1: 32365537-32365537 on Build GRCh38
rs867432071 Chr.1: 32365468-32365468 on Build GRCh38
rs8430 Chr.1: 32365505-32365505 on Build GRCh38
rs979344418 Chr.1: 32365584-32365586 on Build GRCh38
rs41265845 Chr.1: 32365574-32365574 on Build GRCh38
rs1047770219 Chr.1: 32365748-32365748 on Build GRCh38
rs930942005 Chr.1: 32365736-32365736 on Build GRCh38
rs149266604 Chr.1: 32365515-32365515 on Build GRCh38
rs183523234 Chr.1: 32365777-32365777 on Build GRCh38
rs747000961 Chr.1: 32365702-32365702 on Build GRCh38
rs1021624004 Chr.1: 32365561-32365561 on Build GRCh38
rs759200204 Chr.1: 32365820-32365820 on Build GRCh38
rs995728978 Chr.1: 32365441-32365441 on Build GRCh38
rs771193993 Chr.1: 32365587-32365587 on Build GRCh38
rs7515234 Chr.1: 32365758-32365758 on Build GRCh38
rs936039384 Chr.1: 32365888-32365888 on Build GRCh38
rs191096171 Chr.1: 32365557-32365557 on Build GRCh38
rs1009224482 Chr.1: 32365593-32365593 on Build GRCh38
rs137907052 Chr.1: 32365735-32365735 on Build GRCh38
rs1025829296 Chr.1: 32365446-32365446 on Build GRCh38
rs1027377036 Chr.1: 32365663-32365663 on Build GRCh38
rs559712622 Chr.1: 32365726-32365726 on Build GRCh38
rs1040071844 Chr.1: 32365627-32365627 on Build GRCh38
rs571498361 Chr.1: 32365889-32365889 on Build GRCh38
rs887276596 Chr.1: 32365469-32365469 on Build GRCh38
rs138423392 Chr.1: 32365495-32365495 on Build GRCh38
rs143775028 Chr.1: 32365857-32365857 on Build GRCh38
rs948865874 Chr.1: 32365536-32365536 on Build GRCh38
rs917286162 Chr.1: 32365814-32365814 on Build GRCh38
rs1035845867 Chr.1: 32365765-32365765 on Build GRCh38
rs894594587 Chr.1: 32365800-32365800 on Build GRCh38
rs552813541 Chr.1: 32365860-32365860 on Build GRCh38
rs535956507 Chr.1: 32365507-32365507 on Build GRCh38
rs995904197 Chr.1: 32365651-32365651 on Build GRCh38

View Full Product Details