Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32946320-32946830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF19B
Entrez Gene ID
127544
Gene Name
ring finger protein 19B
Gene Aliases
IBRDC3, NKLAM
UniGene
Hs.591504
Gene Chromosome Location
Chr.1: 32932498-32965266 on Build GRCh38
Transcripts
NM_001127361.2 NM_001300826.1 NM_153341.3 XM_006710356.2 XM_006710357.3 XM_006710358.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6905898 c.984-8C>T p.? Chr.1: 32946572-32946572 on Build GRCh38
4433987 c.1133A>T p.Y378F Chr.1: 32946415-32946415 on Build GRCh38
5781460 c.1019G>C p.W340S Chr.1: 32946529-32946529 on Build GRCh38
6503658 c.1126C>T p.P376S Chr.1: 32946422-32946422 on Build GRCh38
4433988 c.563A>T p.Y188F Chr.1: 32946415-32946415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775597353 Chr.1: 32946357-32946357 on Build GRCh38
rs368182501 Chr.1: 32946758-32946758 on Build GRCh38
rs918947596 Chr.1: 32946798-32946798 on Build GRCh38
rs933115922 Chr.1: 32946720-32946720 on Build GRCh38
rs780719037 Chr.1: 32946679-32946679 on Build GRCh38
rs575228058 Chr.1: 32946620-32946620 on Build GRCh38
rs951474991 Chr.1: 32946588-32946588 on Build GRCh38
rs1036457079 Chr.1: 32946785-32946785 on Build GRCh38
rs941669937 Chr.1: 32946732-32946732 on Build GRCh38
rs759649860 Chr.1: 32946355-32946355 on Build GRCh38
rs758913107 Chr.1: 32946590-32946590 on Build GRCh38
rs997588838 Chr.1: 32946360-32946360 on Build GRCh38
rs754723472 Chr.1: 32946392-32946392 on Build GRCh38
rs141361817 Chr.1: 32946507-32946507 on Build GRCh38
rs780690670 Chr.1: 32946398-32946398 on Build GRCh38
rs993331033 Chr.1: 32946552-32946552 on Build GRCh38
rs376812829 Chr.1: 32946695-32946695 on Build GRCh38
rs767545315 Chr.1: 32946356-32946356 on Build GRCh38
rs760659205 Chr.1: 32946509-32946509 on Build GRCh38
rs1035717851 Chr.1: 32946604-32946604 on Build GRCh38
rs772058503 Chr.1: 32946493-32946493 on Build GRCh38
rs747984301 Chr.1: 32946414-32946414 on Build GRCh38
rs1037580895 Chr.1: 32946757-32946757 on Build GRCh38
rs374691161 Chr.1: 32946598-32946598 on Build GRCh38
rs201225036 Chr.1: 32946523-32946523 on Build GRCh38
rs746650142 Chr.1: 32946390-32946390 on Build GRCh38
rs965469826 Chr.1: 32946581-32946581 on Build GRCh38
rs773106988 Chr.1: 32946352-32946352 on Build GRCh38
rs201926593 Chr.1: 32946471-32946471 on Build GRCh38
rs572837952 Chr.1: 32946722-32946722 on Build GRCh38
rs780936067 Chr.1: 32946612-32946612 on Build GRCh38
rs750922969 Chr.1: 32946375-32946375 on Build GRCh38
rs374195039 Chr.1: 32946369-32946369 on Build GRCh38
rs780027556 Chr.1: 32946345-32946345 on Build GRCh38
rs779561180 Chr.1: 32946387-32946387 on Build GRCh38
rs151157398 Chr.1: 32946492-32946492 on Build GRCh38
rs760892264 Chr.1: 32946359-32946359 on Build GRCh38
rs539492342 Chr.1: 32946623-32946623 on Build GRCh38
rs538048289 Chr.1: 32946591-32946591 on Build GRCh38
rs765651717 Chr.1: 32946534-32946534 on Build GRCh38
rs760380487 Chr.1: 32946596-32946600 on Build GRCh38
rs769543687 Chr.1: 32946417-32946417 on Build GRCh38
rs770177839 Chr.1: 32946795-32946795 on Build GRCh38
rs1041415771 Chr.1: 32946347-32946347 on Build GRCh38
rs1021023270 Chr.1: 32946564-32946564 on Build GRCh38
rs192269346 Chr.1: 32946784-32946784 on Build GRCh38
rs888765343 Chr.1: 32946739-32946739 on Build GRCh38
rs958975177 Chr.1: 32946607-32946607 on Build GRCh38
rs764218663 Chr.1: 32946362-32946362 on Build GRCh38
rs758007937 Chr.1: 32946381-32946381 on Build GRCh38
rs754419951 Chr.1: 32946597-32946597 on Build GRCh38
rs1007501330 Chr.1: 32946403-32946403 on Build GRCh38
rs373827607 Chr.1: 32946555-32946555 on Build GRCh38
rs943044361 Chr.1: 32946753-32946753 on Build GRCh38
rs556754322 Chr.1: 32946614-32946614 on Build GRCh38
rs762300816 Chr.1: 32946524-32946524 on Build GRCh38
rs1051474407 Chr.1: 32946380-32946380 on Build GRCh38
rs754288661 Chr.1: 32946365-32946365 on Build GRCh38
rs765886239 Chr.1: 32946372-32946372 on Build GRCh38

View Full Product Details