Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33862774-33863269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000189.1 XM_017000190.1 XM_017000191.1 XM_017000192.1 XM_017000193.1 XM_017000194.1 NM_145205.5

Gene Symbol
HMGB4
Entrez Gene ID
127540
Gene Name
high mobility group box 4
Gene Aliases
dJ1007G16.5
UniGene
Hs.568628
Gene Chromosome Location
Chr.1: 33860475-33864791 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000189.1 XM_017000190.1 XM_017000191.1 XM_017000192.1 XM_017000193.1 XM_017000194.1 NM_145205.5

SNP Detail

SNP ID Chromosome Location
rs2295606 Chr.1: 33862951-33862951 on Build GRCh38
rs2295607 Chr.1: 33863047-33863047 on Build GRCh38
rs200669327 Chr.1: 33862954-33862954 on Build GRCh38
rs936447861 Chr.1: 33863214-33863214 on Build GRCh38
rs764078166 Chr.1: 33862955-33862955 on Build GRCh38
rs1053053537 Chr.1: 33863009-33863009 on Build GRCh38
rs932555041 Chr.1: 33863063-33863063 on Build GRCh38
rs374202941 Chr.1: 33863145-33863145 on Build GRCh38
rs112678128 Chr.1: 33863087-33863087 on Build GRCh38
rs896327009 Chr.1: 33862870-33862870 on Build GRCh38
rs1031120820 Chr.1: 33863044-33863044 on Build GRCh38
rs895526734 Chr.1: 33863177-33863177 on Build GRCh38
rs771999603 Chr.1: 33863168-33863168 on Build GRCh38
rs911942639 Chr.1: 33863027-33863027 on Build GRCh38
rs560184249 Chr.1: 33863229-33863230 on Build GRCh38
rs529754097 Chr.1: 33863140-33863140 on Build GRCh38
rs1048376380 Chr.1: 33863073-33863073 on Build GRCh38
rs934630244 Chr.1: 33862994-33862994 on Build GRCh38
rs1019171790 Chr.1: 33863067-33863067 on Build GRCh38
rs749712217 Chr.1: 33862854-33862854 on Build GRCh38
rs916092913 Chr.1: 33862943-33862943 on Build GRCh38
rs183369462 Chr.1: 33863048-33863048 on Build GRCh38
rs959818042 Chr.1: 33862901-33862901 on Build GRCh38
rs979003833 Chr.1: 33862962-33862962 on Build GRCh38
rs1003956108 Chr.1: 33862856-33862856 on Build GRCh38
rs947862453 Chr.1: 33862892-33862892 on Build GRCh38
rs984878599 Chr.1: 33863120-33863120 on Build GRCh38
rs771122070 Chr.1: 33862862-33862862 on Build GRCh38
rs974866291 Chr.1: 33863080-33863080 on Build GRCh38
rs1014031047 Chr.1: 33863208-33863208 on Build GRCh38
rs778105013 Chr.1: 33862811-33862811 on Build GRCh38
rs947894608 Chr.1: 33863182-33863182 on Build GRCh38
rs993381280 Chr.1: 33862904-33862904 on Build GRCh38
rs35600828 Chr.1: 33862944-33862945 on Build GRCh38
rs1023533353 Chr.1: 33863229-33863229 on Build GRCh38
rs925951418 Chr.1: 33862974-33862974 on Build GRCh38
rs903574951 Chr.1: 33862958-33862958 on Build GRCh38
rs992499062 Chr.1: 33863158-33863158 on Build GRCh38
rs144823421 Chr.1: 33863239-33863239 on Build GRCh38
rs796132642 Chr.1: 33863215-33863215 on Build GRCh38
rs1044916726 Chr.1: 33863185-33863185 on Build GRCh38
rs966206146 Chr.1: 33863071-33863071 on Build GRCh38
rs940678646 Chr.1: 33863146-33863146 on Build GRCh38
rs11808622 Chr.1: 33863108-33863108 on Build GRCh38
rs899688055 Chr.1: 33863057-33863057 on Build GRCh38
rs903689707 Chr.1: 33863198-33863198 on Build GRCh38

View Full Product Details