Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35006722-35007223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYM6
Entrez Gene ID
9204
Gene Name
zinc finger MYM-type containing 6
Gene Aliases
Buster2, MYM, ZBED7, ZNF198L4, ZNF258
UniGene
Hs.533986
Gene Chromosome Location
Chr.1: 34986165-35031968 on Build GRCh38
Transcripts
NM_007167.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
221229 c.1667T>A p.M556K Chr.1: 35007097-35007097 on Build GRCh38
6827587 c.1690C>T p.R564* Chr.1: 35007074-35007074 on Build GRCh38
2080012 c.1811A>C p.K604T Chr.1: 35006953-35006953 on Build GRCh38
6827586 c.1690C>T p.R564* Chr.1: 35007074-35007074 on Build GRCh38
2080017 c.1669G>A p.A557T Chr.1: 35007095-35007095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774159523 Chr.1: 35006970-35006970 on Build GRCh38
rs770588682 Chr.1: 35007079-35007079 on Build GRCh38
rs933134419 Chr.1: 35007177-35007177 on Build GRCh38
rs779199565 Chr.1: 35007008-35007008 on Build GRCh38
rs772037358 Chr.1: 35006776-35006776 on Build GRCh38
rs555947646 Chr.1: 35007163-35007163 on Build GRCh38
rs750923745 Chr.1: 35007062-35007073 on Build GRCh38
rs1031245534 Chr.1: 35006934-35006934 on Build GRCh38
rs780383655 Chr.1: 35007040-35007040 on Build GRCh38
rs372026265 Chr.1: 35007165-35007165 on Build GRCh38
rs763872677 Chr.1: 35007143-35007143 on Build GRCh38
rs977210813 Chr.1: 35007153-35007154 on Build GRCh38
rs750414559 Chr.1: 35007012-35007012 on Build GRCh38
rs921246865 Chr.1: 35006847-35006847 on Build GRCh38
rs767488208 Chr.1: 35006989-35006989 on Build GRCh38
rs755376193 Chr.1: 35007057-35007057 on Build GRCh38
rs200352168 Chr.1: 35007041-35007041 on Build GRCh38
rs940165256 Chr.1: 35006966-35006966 on Build GRCh38
rs148412755 Chr.1: 35007130-35007130 on Build GRCh38
rs77203730 Chr.1: 35007181-35007181 on Build GRCh38
rs766936383 Chr.1: 35007069-35007069 on Build GRCh38
rs757632787 Chr.1: 35006919-35006919 on Build GRCh38
rs369377351 Chr.1: 35007090-35007090 on Build GRCh38
rs747400838 Chr.1: 35007046-35007046 on Build GRCh38
rs201365292 Chr.1: 35007058-35007058 on Build GRCh38
rs747665896 Chr.1: 35006915-35006915 on Build GRCh38
rs199688464 Chr.1: 35007068-35007068 on Build GRCh38
rs748753112 Chr.1: 35007074-35007074 on Build GRCh38
rs752649137 Chr.1: 35007002-35007002 on Build GRCh38
rs977138698 Chr.1: 35006800-35006800 on Build GRCh38
rs765946298 Chr.1: 35006962-35006962 on Build GRCh38
rs1000269768 Chr.1: 35007118-35007118 on Build GRCh38
rs193216010 Chr.1: 35006856-35006856 on Build GRCh38
rs964083811 Chr.1: 35007123-35007123 on Build GRCh38
rs922704900 Chr.1: 35007162-35007162 on Build GRCh38
rs1032886562 Chr.1: 35007154-35007154 on Build GRCh38
rs905492245 Chr.1: 35007092-35007092 on Build GRCh38
rs538621431 Chr.1: 35006785-35006785 on Build GRCh38
rs1048600962 Chr.1: 35006901-35006901 on Build GRCh38
rs772780362 Chr.1: 35007073-35007073 on Build GRCh38
rs772809970 Chr.1: 35006944-35006944 on Build GRCh38
rs913980107 Chr.1: 35006772-35006772 on Build GRCh38
rs765344956 Chr.1: 35007010-35007010 on Build GRCh38
rs762582923 Chr.1: 35006950-35006950 on Build GRCh38
rs773518065 Chr.1: 35007025-35007025 on Build GRCh38
rs184937835 Chr.1: 35006984-35006984 on Build GRCh38
rs978274225 Chr.1: 35006929-35006929 on Build GRCh38
rs1037233611 Chr.1: 35007032-35007032 on Build GRCh38
rs373111034 Chr.1: 35007096-35007096 on Build GRCh38
rs777147341 Chr.1: 35006912-35006912 on Build GRCh38
rs757346834 Chr.1: 35007003-35007003 on Build GRCh38
rs769319480 Chr.1: 35006924-35006924 on Build GRCh38
rs573500134 Chr.1: 35007125-35007125 on Build GRCh38

View Full Product Details