Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35187039-35187451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFPQ
Entrez Gene ID
6421
Gene Name
splicing factor proline and glutamine rich
Gene Aliases
POMP100, PPP1R140, PSF
UniGene
Hs.355934
Gene Chromosome Location
Chr.1: 35176378-35193161 on Build GRCh38
Transcripts
NM_005066.2 NR_136702.1 NR_136703.1 XM_005271112.4 XM_005271113.4 XM_005271115.4 XM_011541950.2 XM_017002053.1 XM_017002054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3705808 c.1822A>T p.R608* Chr.1: 35187245-35187245 on Build GRCh38
4399391 c.1921G>C p.G641R Chr.1: 35187066-35187066 on Build GRCh38
5380792 c.1861G>A p.G621R Chr.1: 35187206-35187206 on Build GRCh38
908656 c.1905_1907delAGG p.G639delG Chr.1: 35187080-35187082 on Build GRCh38
6882036 c.1942G>A p.V648I Chr.1: 35187045-35187045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749368384 Chr.1: 35187224-35187224 on Build GRCh38
rs761293729 Chr.1: 35187254-35187254 on Build GRCh38
rs970606129 Chr.1: 35187402-35187402 on Build GRCh38
rs928753019 Chr.1: 35187230-35187230 on Build GRCh38
rs772954118 Chr.1: 35187284-35187285 on Build GRCh38
rs767708763 Chr.1: 35187167-35187167 on Build GRCh38
rs749566365 Chr.1: 35187155-35187155 on Build GRCh38
rs772510250 Chr.1: 35187249-35187249 on Build GRCh38
rs1036220349 Chr.1: 35187393-35187393 on Build GRCh38
rs748399931 Chr.1: 35187133-35187133 on Build GRCh38
rs780043878 Chr.1: 35187137-35187137 on Build GRCh38
rs775677201 Chr.1: 35187250-35187250 on Build GRCh38
rs189040885 Chr.1: 35187225-35187225 on Build GRCh38
rs765294564 Chr.1: 35187259-35187261 on Build GRCh38
rs113394628 Chr.1: 35187252-35187252 on Build GRCh38
rs191776739 Chr.1: 35187074-35187074 on Build GRCh38
rs183603648 Chr.1: 35187151-35187151 on Build GRCh38
rs764864655 Chr.1: 35187198-35187198 on Build GRCh38
rs750094587 Chr.1: 35187289-35187289 on Build GRCh38
rs753323073 Chr.1: 35187178-35187178 on Build GRCh38
rs3738697 Chr.1: 35187169-35187169 on Build GRCh38
rs1044968942 Chr.1: 35187420-35187420 on Build GRCh38
rs868647178 Chr.1: 35187240-35187240 on Build GRCh38
rs368160216 Chr.1: 35187189-35187189 on Build GRCh38
rs976067975 Chr.1: 35187328-35187328 on Build GRCh38
rs887159697 Chr.1: 35187118-35187118 on Build GRCh38
rs113612711 Chr.1: 35187334-35187334 on Build GRCh38
rs751294618 Chr.1: 35187303-35187303 on Build GRCh38
rs993841476 Chr.1: 35187383-35187383 on Build GRCh38
rs536164218 Chr.1: 35187376-35187376 on Build GRCh38
rs756872647 Chr.1: 35187082-35187082 on Build GRCh38
rs764570498 Chr.1: 35187258-35187258 on Build GRCh38
rs77260199 Chr.1: 35187311-35187311 on Build GRCh38
rs372398534 Chr.1: 35187158-35187158 on Build GRCh38
rs370767761 Chr.1: 35187284-35187284 on Build GRCh38
rs746993364 Chr.1: 35187209-35187209 on Build GRCh38
rs201299583 Chr.1: 35187156-35187156 on Build GRCh38
rs762469692 Chr.1: 35187291-35187291 on Build GRCh38
rs779662255 Chr.1: 35187205-35187205 on Build GRCh38
rs970512851 Chr.1: 35187196-35187196 on Build GRCh38
rs1026212496 Chr.1: 35187397-35187397 on Build GRCh38
rs761877695 Chr.1: 35187188-35187188 on Build GRCh38
rs370511688 Chr.1: 35187175-35187175 on Build GRCh38
rs778051389 Chr.1: 35187216-35187216 on Build GRCh38
rs745648598 Chr.1: 35187119-35187119 on Build GRCh38
rs529592254 Chr.1: 35187215-35187215 on Build GRCh38
rs750009834 Chr.1: 35187200-35187200 on Build GRCh38
rs908647211 Chr.1: 35187355-35187355 on Build GRCh38
rs749833241 Chr.1: 35187314-35187314 on Build GRCh38
rs758333137 Chr.1: 35187126-35187126 on Build GRCh38
rs1036582774 Chr.1: 35187144-35187144 on Build GRCh38
rs376421305 Chr.1: 35187062-35187062 on Build GRCh38
rs888896648 Chr.1: 35187330-35187330 on Build GRCh38
rs1051176496 Chr.1: 35187083-35187083 on Build GRCh38
rs763690702 Chr.1: 35187176-35187176 on Build GRCh38
rs776188339 Chr.1: 35187275-35187275 on Build GRCh38
rs1041961578 Chr.1: 35187301-35187301 on Build GRCh38
rs534000084 Chr.1: 35187350-35187350 on Build GRCh38
rs6702141 Chr.1: 35187340-35187340 on Build GRCh38
rs528161573 Chr.1: 35187154-35187154 on Build GRCh38
rs766232394 Chr.1: 35187298-35187298 on Build GRCh38
rs573969864 Chr.1: 35187410-35187410 on Build GRCh38
rs1051920238 Chr.1: 35187318-35187318 on Build GRCh38
rs191047231 Chr.1: 35187346-35187346 on Build GRCh38
rs746020776 Chr.1: 35187243-35187243 on Build GRCh38
rs758223526 Chr.1: 35187203-35187203 on Build GRCh38
rs769107656 Chr.1: 35187131-35187131 on Build GRCh38
rs773549769 Chr.1: 35187285-35187285 on Build GRCh38
rs112492610 Chr.1: 35187373-35187373 on Build GRCh38
rs774720675 Chr.1: 35187236-35187236 on Build GRCh38
rs778728322 Chr.1: 35187116-35187116 on Build GRCh38
rs764037727 Chr.1: 35187070-35187071 on Build GRCh38
rs940086026 Chr.1: 35187358-35187358 on Build GRCh38
rs753800338 Chr.1: 35187180-35187182 on Build GRCh38

View Full Product Details