Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58980288-58980793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPM1E
Entrez Gene ID
22843
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1E
Gene Aliases
CaMKP-N, POPX1, PP2CH, caMKN
UniGene
Hs.245044
Gene Chromosome Location
Chr.17: 58755869-58985617 on Build GRCh38
Transcripts
NM_014906.4 NR_048561.1 XM_005257169.4 XM_011524534.2 XM_017024359.1 XM_017024662.1 XM_017024663.1

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
UniGene
Hs.579079
Gene Chromosome Location
Chr.17: 58968068-59106921 on Build GRCh38
Transcripts
NM_014906.4 NR_048561.1 XM_005257169.4 XM_011524534.2 XM_017024359.1 XM_017024662.1 XM_017024663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6893098 c.1903A>G p.T635A Chr.17: 58980666-58980666 on Build GRCh38
981998 c.1634G>A p.R545H Chr.17: 58980397-58980397 on Build GRCh38
4917082 c.1705T>A p.Y569N Chr.17: 58980468-58980468 on Build GRCh38
5964500 c.1716C>T p.L572L Chr.17: 58980479-58980479 on Build GRCh38
6839490 c.1599G>C p.Q533H Chr.17: 58980362-58980362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766408513 Chr.17: 58980372-58980372 on Build GRCh38
rs199649187 Chr.17: 58980681-58980681 on Build GRCh38
rs147003260 Chr.17: 58980321-58980321 on Build GRCh38
rs937435070 Chr.17: 58980355-58980355 on Build GRCh38
rs35177815 Chr.17: 58980672-58980673 on Build GRCh38
rs754234636 Chr.17: 58980513-58980513 on Build GRCh38
rs755928815 Chr.17: 58980588-58980588 on Build GRCh38
rs749465016 Chr.17: 58980463-58980463 on Build GRCh38
rs2055780 Chr.17: 58980757-58980757 on Build GRCh38
rs750053884 Chr.17: 58980407-58980407 on Build GRCh38
rs61739451 Chr.17: 58980743-58980743 on Build GRCh38
rs113639836 Chr.17: 58980516-58980516 on Build GRCh38
rs577558425 Chr.17: 58980462-58980462 on Build GRCh38
rs369815595 Chr.17: 58980312-58980312 on Build GRCh38
rs777220400 Chr.17: 58980765-58980765 on Build GRCh38
rs377209500 Chr.17: 58980672-58980672 on Build GRCh38
rs564602902 Chr.17: 58980608-58980608 on Build GRCh38
rs770059329 Chr.17: 58980334-58980334 on Build GRCh38
rs760128692 Chr.17: 58980378-58980378 on Build GRCh38
rs374085623 Chr.17: 58980604-58980604 on Build GRCh38
rs769256860 Chr.17: 58980633-58980633 on Build GRCh38
rs917406014 Chr.17: 58980432-58980432 on Build GRCh38
rs746918842 Chr.17: 58980319-58980319 on Build GRCh38
rs1001578068 Chr.17: 58980722-58980722 on Build GRCh38
rs781198246 Chr.17: 58980584-58980584 on Build GRCh38
rs774553877 Chr.17: 58980486-58980486 on Build GRCh38
rs762255757 Chr.17: 58980642-58980642 on Build GRCh38
rs777582558 Chr.17: 58980589-58980589 on Build GRCh38
rs776250946 Chr.17: 58980568-58980570 on Build GRCh38
rs765944508 Chr.17: 58980529-58980529 on Build GRCh38
rs111520252 Chr.17: 58980696-58980696 on Build GRCh38
rs756108762 Chr.17: 58980716-58980716 on Build GRCh38
rs775793603 Chr.17: 58980497-58980497 on Build GRCh38
rs748141026 Chr.17: 58980610-58980610 on Build GRCh38
rs149595183 Chr.17: 58980595-58980595 on Build GRCh38
rs2108433 Chr.17: 58980567-58980567 on Build GRCh38
rs138207802 Chr.17: 58980343-58980343 on Build GRCh38
rs756935591 Chr.17: 58980730-58980730 on Build GRCh38
rs747992276 Chr.17: 58980450-58980450 on Build GRCh38
rs368424199 Chr.17: 58980525-58980525 on Build GRCh38
rs753162691 Chr.17: 58980396-58980396 on Build GRCh38
rs145669199 Chr.17: 58980635-58980635 on Build GRCh38
rs148760609 Chr.17: 58980627-58980627 on Build GRCh38
rs369695755 Chr.17: 58980503-58980503 on Build GRCh38
rs754393205 Chr.17: 58980554-58980554 on Build GRCh38
rs747894727 Chr.17: 58980331-58980331 on Build GRCh38
rs144328670 Chr.17: 58980596-58980596 on Build GRCh38
rs779761727 Chr.17: 58980426-58980426 on Build GRCh38
rs1051547823 Chr.17: 58980547-58980547 on Build GRCh38
rs769344972 Chr.17: 58980763-58980763 on Build GRCh38
rs759121062 Chr.17: 58980671-58980671 on Build GRCh38
rs201271726 Chr.17: 58980662-58980662 on Build GRCh38
rs767905257 Chr.17: 58980383-58980383 on Build GRCh38
rs756485394 Chr.17: 58980397-58980397 on Build GRCh38
rs745964136 Chr.17: 58980600-58980600 on Build GRCh38
rs138211677 Chr.17: 58980581-58980581 on Build GRCh38
rs890283375 Chr.17: 58980564-58980564 on Build GRCh38
rs139860807 Chr.17: 58980537-58980537 on Build GRCh38
rs762529848 Chr.17: 58980710-58980710 on Build GRCh38
rs1011779488 Chr.17: 58980590-58980590 on Build GRCh38
rs374167393 Chr.17: 58980440-58980440 on Build GRCh38
rs769515069 Chr.17: 58980458-58980458 on Build GRCh38
rs769412144 Chr.17: 58980612-58980612 on Build GRCh38
rs1038678844 Chr.17: 58980526-58980526 on Build GRCh38
rs761247536 Chr.17: 58980700-58980702 on Build GRCh38
rs200537682 Chr.17: 58980441-58980441 on Build GRCh38
rs763026756 Chr.17: 58980347-58980347 on Build GRCh38
rs369185588 Chr.17: 58980734-58980734 on Build GRCh38
rs766068408 Chr.17: 58980653-58980653 on Build GRCh38
rs1010509355 Chr.17: 58980568-58980568 on Build GRCh38
rs183396449 Chr.17: 58980694-58980694 on Build GRCh38
rs541362319 Chr.17: 58980712-58980712 on Build GRCh38
rs71372861 Chr.17: 58980381-58980381 on Build GRCh38
rs768600680 Chr.17: 58980342-58980342 on Build GRCh38
rs764503398 Chr.17: 58980511-58980511 on Build GRCh38
rs752594592 Chr.17: 58980585-58980585 on Build GRCh38
rs759516881 Chr.17: 58980487-58980487 on Build GRCh38
rs774271250 Chr.17: 58980377-58980377 on Build GRCh38
rs377220140 Chr.17: 58980482-58980482 on Build GRCh38
rs764982053 Chr.17: 58980403-58980403 on Build GRCh38
rs960291670 Chr.17: 58980762-58980762 on Build GRCh38
rs751110645 Chr.17: 58980669-58980669 on Build GRCh38
rs747082201 Chr.17: 58980744-58980744 on Build GRCh38
rs267604972 Chr.17: 58980717-58980717 on Build GRCh38
rs758001755 Chr.17: 58980424-58980424 on Build GRCh38
rs768082592 Chr.17: 58980493-58980493 on Build GRCh38
rs780978666 Chr.17: 58980449-58980449 on Build GRCh38
rs772314556 Chr.17: 58980742-58980742 on Build GRCh38
rs142959142 Chr.17: 58980425-58980425 on Build GRCh38

View Full Product Details