Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61408319-61408809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX2
Entrez Gene ID
6909
Gene Name
T-box 2
Gene Aliases
-
UniGene
Hs.531085
Gene Chromosome Location
Chr.17: 61399896-61409466 on Build GRCh38
Transcripts
NM_005994.3 XM_011525159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3421754 c.2015G>A p.R672H Chr.17: 61408382-61408382 on Build GRCh38
4436023 c.2065C>G p.Q689E Chr.17: 61408432-61408432 on Build GRCh38
4617797 c.2016C>T p.R672R Chr.17: 61408383-61408383 on Build GRCh38
3520404 c.1988C>T p.A663V Chr.17: 61408355-61408355 on Build GRCh38
6426985 c.2051C>A p.A684E Chr.17: 61408418-61408418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753175622 Chr.17: 61408357-61408357 on Build GRCh38
rs549041111 Chr.17: 61408446-61408446 on Build GRCh38
rs770134275 Chr.17: 61408521-61408521 on Build GRCh38
rs78939123 Chr.17: 61408744-61408744 on Build GRCh38
rs573739062 Chr.17: 61408571-61408571 on Build GRCh38
rs889039841 Chr.17: 61408788-61408788 on Build GRCh38
rs34493156 Chr.17: 61408360-61408360 on Build GRCh38
rs923936854 Chr.17: 61408651-61408651 on Build GRCh38
rs949185977 Chr.17: 61408604-61408604 on Build GRCh38
rs965854531 Chr.17: 61408569-61408569 on Build GRCh38
rs940432521 Chr.17: 61408709-61408709 on Build GRCh38
rs61751978 Chr.17: 61408397-61408397 on Build GRCh38
rs1014995360 Chr.17: 61408471-61408471 on Build GRCh38
rs1009042266 Chr.17: 61408524-61408524 on Build GRCh38
rs557033648 Chr.17: 61408564-61408564 on Build GRCh38
rs545290637 Chr.17: 61408743-61408743 on Build GRCh38
rs1037683921 Chr.17: 61408431-61408431 on Build GRCh38
rs775935102 Chr.17: 61408490-61408490 on Build GRCh38
rs899163658 Chr.17: 61408472-61408472 on Build GRCh38
rs981937858 Chr.17: 61408630-61408630 on Build GRCh38
rs773230802 Chr.17: 61408396-61408396 on Build GRCh38
rs989659952 Chr.17: 61408669-61408669 on Build GRCh38
rs764671010 Chr.17: 61408361-61408361 on Build GRCh38
rs371416364 Chr.17: 61408347-61408347 on Build GRCh38
rs890619577 Chr.17: 61408504-61408504 on Build GRCh38
rs1046556033 Chr.17: 61408696-61408696 on Build GRCh38
rs779016245 Chr.17: 61408373-61408373 on Build GRCh38
rs373456109 Chr.17: 61408501-61408501 on Build GRCh38
rs553420511 Chr.17: 61408672-61408672 on Build GRCh38
rs780953070 Chr.17: 61408382-61408382 on Build GRCh38
rs767959344 Chr.17: 61408355-61408355 on Build GRCh38
rs1004517871 Chr.17: 61408456-61408456 on Build GRCh38
rs948234669 Chr.17: 61408384-61408384 on Build GRCh38
rs367963765 Chr.17: 61408372-61408372 on Build GRCh38
rs865851098 Chr.17: 61408557-61408557 on Build GRCh38
rs1009119179 Chr.17: 61408728-61408728 on Build GRCh38
rs1032540702 Chr.17: 61408782-61408782 on Build GRCh38
rs59382073 Chr.17: 61408724-61408724 on Build GRCh38
rs746060657 Chr.17: 61408379-61408379 on Build GRCh38
rs371734859 Chr.17: 61408528-61408528 on Build GRCh38
rs565941283 Chr.17: 61408497-61408497 on Build GRCh38
rs1057989 Chr.17: 61408433-61408433 on Build GRCh38
rs1019396980 Chr.17: 61408344-61408344 on Build GRCh38
rs1031967750 Chr.17: 61408548-61408548 on Build GRCh38
rs932021513 Chr.17: 61408717-61408717 on Build GRCh38
rs747949718 Chr.17: 61408383-61408383 on Build GRCh38
rs1028944131 Chr.17: 61408518-61408518 on Build GRCh38
rs571416681 Chr.17: 61408545-61408545 on Build GRCh38
rs753824335 Chr.17: 61408366-61408366 on Build GRCh38
rs948182337 Chr.17: 61408688-61408688 on Build GRCh38
rs990729246 Chr.17: 61408561-61408561 on Build GRCh38
rs1028891934 Chr.17: 61408766-61408766 on Build GRCh38
rs113424512 Chr.17: 61408381-61408381 on Build GRCh38
rs924184903 Chr.17: 61408346-61408346 on Build GRCh38
rs757537787 Chr.17: 61408359-61408359 on Build GRCh38
rs765906139 Chr.17: 61408339-61408339 on Build GRCh38
rs35789942 Chr.17: 61408407-61408407 on Build GRCh38
rs181013488 Chr.17: 61408765-61408765 on Build GRCh38
rs769551183 Chr.17: 61408395-61408395 on Build GRCh38
rs528883638 Chr.17: 61408425-61408425 on Build GRCh38
rs763099583 Chr.17: 61408658-61408658 on Build GRCh38
rs1050617790 Chr.17: 61408723-61408723 on Build GRCh38
rs906735179 Chr.17: 61408698-61408698 on Build GRCh38
rs537100522 Chr.17: 61408563-61408563 on Build GRCh38
rs953163538 Chr.17: 61408519-61408519 on Build GRCh38
rs536635857 Chr.17: 61408572-61408572 on Build GRCh38
rs936913573 Chr.17: 61408354-61408354 on Build GRCh38
rs966458923 Chr.17: 61408544-61408545 on Build GRCh38
rs748322106 Chr.17: 61408491-61408491 on Build GRCh38
rs903355791 Chr.17: 61408750-61408750 on Build GRCh38
rs188452348 Chr.17: 61408534-61408534 on Build GRCh38
rs761134753 Chr.17: 61408503-61408503 on Build GRCh38
rs759307454 Chr.17: 61408348-61408348 on Build GRCh38
rs771904517 Chr.17: 61408454-61408454 on Build GRCh38
rs916383563 Chr.17: 61408377-61408377 on Build GRCh38
rs773481704 Chr.17: 61408578-61408578 on Build GRCh38
rs907719064 Chr.17: 61408641-61408641 on Build GRCh38

View Full Product Details