Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61942496-61942997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED13
Entrez Gene ID
9969
Gene Name
mediator complex subunit 13
Gene Aliases
ARC250, DRIP250, HSPC221, THRAP1, TRAP240
UniGene
Hs.282678
Gene Chromosome Location
Chr.17: 61942605-62065983 on Build GRCh38
Transcripts
NM_005121.2 XM_011525551.2 XM_011525553.2

SNP Detail

SNP ID Chromosome Location
rs1049735085 Chr.17: 61942657-61942660 on Build GRCh38
rs973565534 Chr.17: 61942931-61942931 on Build GRCh38
rs143595519 Chr.17: 61942566-61942566 on Build GRCh38
rs1004334667 Chr.17: 61942892-61942892 on Build GRCh38
rs528492810 Chr.17: 61942898-61942898 on Build GRCh38
rs868271402 Chr.17: 61942573-61942573 on Build GRCh38
rs911916669 Chr.17: 61942668-61942668 on Build GRCh38
rs549491349 Chr.17: 61942917-61942917 on Build GRCh38
rs1014182999 Chr.17: 61942854-61942854 on Build GRCh38
rs543694077 Chr.17: 61942661-61942662 on Build GRCh38
rs115121869 Chr.17: 61942963-61942963 on Build GRCh38
rs904350758 Chr.17: 61942806-61942806 on Build GRCh38
rs922226108 Chr.17: 61942615-61942615 on Build GRCh38
rs893040825 Chr.17: 61942672-61942672 on Build GRCh38
rs963510717 Chr.17: 61942923-61942923 on Build GRCh38
rs147189111 Chr.17: 61942720-61942720 on Build GRCh38
rs1011277672 Chr.17: 61942682-61942682 on Build GRCh38
rs563522162 Chr.17: 61942937-61942937 on Build GRCh38
rs190038315 Chr.17: 61942662-61942662 on Build GRCh38
rs139441247 Chr.17: 61942777-61942779 on Build GRCh38
rs866938640 Chr.17: 61942893-61942893 on Build GRCh38
rs1041815047 Chr.17: 61942740-61942740 on Build GRCh38
rs202050543 Chr.17: 61942779-61942779 on Build GRCh38
rs764102397 Chr.17: 61942700-61942701 on Build GRCh38
rs951080230 Chr.17: 61942873-61942873 on Build GRCh38
rs377276211 Chr.17: 61942658-61942658 on Build GRCh38
rs985882065 Chr.17: 61942844-61942844 on Build GRCh38
rs908008816 Chr.17: 61942527-61942527 on Build GRCh38
rs963520629 Chr.17: 61942534-61942534 on Build GRCh38
rs920207346 Chr.17: 61942834-61942834 on Build GRCh38
rs750714479 Chr.17: 61942948-61942948 on Build GRCh38
rs373336007 Chr.17: 61942832-61942832 on Build GRCh38
rs756252963 Chr.17: 61942854-61942855 on Build GRCh38
rs556389600 Chr.17: 61942669-61942669 on Build GRCh38
rs546297762 Chr.17: 61942724-61942724 on Build GRCh38
rs762294611 Chr.17: 61942814-61942816 on Build GRCh38
rs928797591 Chr.17: 61942527-61942529 on Build GRCh38
rs532856958 Chr.17: 61942906-61942906 on Build GRCh38
rs758749766 Chr.17: 61942961-61942961 on Build GRCh38
rs940284719 Chr.17: 61942550-61942550 on Build GRCh38
rs1016672533 Chr.17: 61942918-61942918 on Build GRCh38
rs561852467 Chr.17: 61942731-61942731 on Build GRCh38
rs571701522 Chr.17: 61942670-61942670 on Build GRCh38
rs564170496 Chr.17: 61942830-61942830 on Build GRCh38
rs765536523 Chr.17: 61942866-61942866 on Build GRCh38
rs540675419 Chr.17: 61942788-61942788 on Build GRCh38
rs931535716 Chr.17: 61942841-61942841 on Build GRCh38
rs970232534 Chr.17: 61942742-61942742 on Build GRCh38
rs1052929864 Chr.17: 61942959-61942959 on Build GRCh38
rs762045094 Chr.17: 61942831-61942831 on Build GRCh38
rs972854782 Chr.17: 61942825-61942825 on Build GRCh38
rs889880054 Chr.17: 61942660-61942660 on Build GRCh38
rs75729250 Chr.17: 61942732-61942732 on Build GRCh38
rs578039563 Chr.17: 61942661-61942661 on Build GRCh38
rs931337510 Chr.17: 61942648-61942650 on Build GRCh38

View Full Product Details