Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62701620-62702120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARCH10
Entrez Gene ID
162333
Gene Name
membrane associated ring-CH-type finger 10
Gene Aliases
MARCH-X, RNF190
UniGene
Hs.665663
Gene Chromosome Location
Chr.17: 62701314-62808381 on Build GRCh38
Transcripts
NM_001100875.2 NM_001288779.1 NM_152598.3 XM_005257095.2 XM_005257096.2 XM_005257097.3 XM_005257098.2 XM_005257100.2 XM_005257102.2 XM_005257106.4 XM_011524435.1 XM_011524436.1 XM_011524437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
982488 c.2421C>T p.V807V Chr.17: 62701709-62701709 on Build GRCh38
3520566 c.2376G>A p.S792S Chr.17: 62701754-62701754 on Build GRCh38
6320598 c.2400C>A p.G800G Chr.17: 62701730-62701730 on Build GRCh38
3520565 c.2377G>A p.E793K Chr.17: 62701753-62701753 on Build GRCh38
2794457 c.2415C>T p.S805S Chr.17: 62701715-62701715 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766131670 Chr.17: 62701726-62701726 on Build GRCh38
rs113295934 Chr.17: 62702014-62702014 on Build GRCh38
rs560559583 Chr.17: 62701676-62701676 on Build GRCh38
rs775528889 Chr.17: 62701685-62701685 on Build GRCh38
rs749461005 Chr.17: 62701654-62701654 on Build GRCh38
rs115317261 Chr.17: 62702095-62702095 on Build GRCh38
rs926085517 Chr.17: 62701868-62701868 on Build GRCh38
rs553216647 Chr.17: 62701898-62701898 on Build GRCh38
rs201291034 Chr.17: 62701803-62701803 on Build GRCh38
rs547266316 Chr.17: 62701754-62701754 on Build GRCh38
rs369228177 Chr.17: 62701805-62701805 on Build GRCh38
rs140049028 Chr.17: 62701762-62701762 on Build GRCh38
rs747082381 Chr.17: 62701798-62701798 on Build GRCh38
rs755150781 Chr.17: 62701708-62701708 on Build GRCh38
rs749602678 Chr.17: 62701673-62701673 on Build GRCh38
rs750192370 Chr.17: 62701647-62701647 on Build GRCh38
rs761954945 Chr.17: 62701709-62701709 on Build GRCh38
rs78299543 Chr.17: 62701911-62701911 on Build GRCh38
rs936744275 Chr.17: 62701887-62701887 on Build GRCh38
rs560693689 Chr.17: 62702007-62702007 on Build GRCh38
rs543905583 Chr.17: 62702000-62702000 on Build GRCh38
rs771189562 Chr.17: 62701683-62701684 on Build GRCh38
rs1008677279 Chr.17: 62702026-62702026 on Build GRCh38
rs77551436 Chr.17: 62701820-62701820 on Build GRCh38
rs202231961 Chr.17: 62701773-62701773 on Build GRCh38
rs574083679 Chr.17: 62702008-62702008 on Build GRCh38
rs375530840 Chr.17: 62701825-62701825 on Build GRCh38
rs558487726 Chr.17: 62701930-62701930 on Build GRCh38
rs1028803136 Chr.17: 62701821-62701821 on Build GRCh38
rs372368836 Chr.17: 62701794-62701794 on Build GRCh38
rs1020185063 Chr.17: 62702033-62702033 on Build GRCh38
rs746270121 Chr.17: 62701790-62701790 on Build GRCh38
rs2251391 Chr.17: 62701664-62701664 on Build GRCh38
rs752249613 Chr.17: 62701751-62701751 on Build GRCh38
rs751697269 Chr.17: 62701679-62701679 on Build GRCh38
rs887046878 Chr.17: 62701953-62701953 on Build GRCh38
rs186586117 Chr.17: 62701827-62701827 on Build GRCh38
rs776241026 Chr.17: 62701672-62701672 on Build GRCh38
rs1038211572 Chr.17: 62702024-62702024 on Build GRCh38
rs574952381 Chr.17: 62701936-62701936 on Build GRCh38
rs771509574 Chr.17: 62701920-62701920 on Build GRCh38
rs936097367 Chr.17: 62701880-62701880 on Build GRCh38
rs760786649 Chr.17: 62701690-62701690 on Build GRCh38
rs575015725 Chr.17: 62701663-62701663 on Build GRCh38
rs771587977 Chr.17: 62702035-62702035 on Build GRCh38
rs538798176 Chr.17: 62701912-62701912 on Build GRCh38
rs986856069 Chr.17: 62701823-62701823 on Build GRCh38
rs145565856 Chr.17: 62702096-62702096 on Build GRCh38
rs530281485 Chr.17: 62701698-62701698 on Build GRCh38
rs780810554 Chr.17: 62701667-62701667 on Build GRCh38
rs776577721 Chr.17: 62701804-62701804 on Build GRCh38
rs754897213 Chr.17: 62701743-62701743 on Build GRCh38
rs898929985 Chr.17: 62701974-62701974 on Build GRCh38
rs539877760 Chr.17: 62702009-62702009 on Build GRCh38
rs1028069488 Chr.17: 62701815-62701815 on Build GRCh38
rs376408224 Chr.17: 62701670-62701670 on Build GRCh38
rs778306629 Chr.17: 62701826-62701826 on Build GRCh38
rs963148369 Chr.17: 62701819-62701819 on Build GRCh38
rs758284471 Chr.17: 62701651-62701651 on Build GRCh38
rs978513882 Chr.17: 62702076-62702076 on Build GRCh38
rs367733917 Chr.17: 62701806-62701806 on Build GRCh38
rs370591950 Chr.17: 62701931-62701931 on Build GRCh38
rs1046116353 Chr.17: 62701939-62701939 on Build GRCh38
rs368543883 Chr.17: 62701680-62701680 on Build GRCh38
rs369696727 Chr.17: 62701787-62701787 on Build GRCh38
rs949622229 Chr.17: 62701916-62701916 on Build GRCh38
rs986775286 Chr.17: 62701642-62701642 on Build GRCh38
rs751343965 Chr.17: 62701737-62701737 on Build GRCh38
rs886683532 Chr.17: 62701934-62701934 on Build GRCh38
rs752801399 Chr.17: 62701645-62701645 on Build GRCh38
rs977636858 Chr.17: 62701860-62701860 on Build GRCh38
rs747708851 Chr.17: 62701669-62701669 on Build GRCh38
rs767601418 Chr.17: 62701750-62701750 on Build GRCh38
rs913148520 Chr.17: 62701840-62701840 on Build GRCh38
rs779017407 Chr.17: 62701765-62701773 on Build GRCh38
rs542514893 Chr.17: 62701770-62701770 on Build GRCh38
rs533978335 Chr.17: 62701830-62701830 on Build GRCh38
rs376657500 Chr.17: 62701646-62701646 on Build GRCh38
rs772539507 Chr.17: 62701793-62701793 on Build GRCh38
rs772586720 Chr.17: 62701683-62701683 on Build GRCh38
rs868575556 Chr.17: 62701659-62701659 on Build GRCh38
rs546340515 Chr.17: 62702099-62702099 on Build GRCh38
rs538354576 Chr.17: 62701807-62701807 on Build GRCh38
rs954584199 Chr.17: 62702022-62702022 on Build GRCh38
rs958373620 Chr.17: 62702097-62702097 on Build GRCh38
rs966715979 Chr.17: 62701852-62701852 on Build GRCh38
rs774579140 Chr.17: 62701677-62701677 on Build GRCh38
rs968006041 Chr.17: 62701854-62701854 on Build GRCh38

View Full Product Details