Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45149556-45150063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXIM1
Entrez Gene ID
10614
Gene Name
hexamethylene bisacetamide inducible 1
Gene Aliases
CLP1, EDG1, HIS1, MAQ1
UniGene
Hs.586945
Gene Chromosome Location
Chr.17: 45147317-45152101 on Build GRCh38
Transcripts
NM_006460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691624 c.804G>A p.S268S Chr.17: 45149994-45149994 on Build GRCh38
6450969 c.819G>A p.R273R Chr.17: 45150009-45150009 on Build GRCh38
3932621 c.657T>C p.D219D Chr.17: 45149847-45149847 on Build GRCh38
6043096 c.477G>T p.K159N Chr.17: 45149667-45149667 on Build GRCh38
6052109 c.826A>G p.T276A Chr.17: 45150016-45150016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759584471 Chr.17: 45149709-45149709 on Build GRCh38
rs755618582 Chr.17: 45149635-45149635 on Build GRCh38
rs766533560 Chr.17: 45149889-45149891 on Build GRCh38
rs866763268 Chr.17: 45149786-45149786 on Build GRCh38
rs776890458 Chr.17: 45149703-45149703 on Build GRCh38
rs769841685 Chr.17: 45149856-45149856 on Build GRCh38
rs369159677 Chr.17: 45149850-45149850 on Build GRCh38
rs774379395 Chr.17: 45150021-45150021 on Build GRCh38
rs17845094 Chr.17: 45149845-45149845 on Build GRCh38
rs771328485 Chr.17: 45149836-45149836 on Build GRCh38
rs762800485 Chr.17: 45149723-45149723 on Build GRCh38
rs998890294 Chr.17: 45149665-45149665 on Build GRCh38
rs746307675 Chr.17: 45149955-45149955 on Build GRCh38
rs375900104 Chr.17: 45149679-45149679 on Build GRCh38
rs761857124 Chr.17: 45150030-45150030 on Build GRCh38
rs142819745 Chr.17: 45149768-45149768 on Build GRCh38
rs200660499 Chr.17: 45149841-45149841 on Build GRCh38
rs778315232 Chr.17: 45149798-45149798 on Build GRCh38
rs747440430 Chr.17: 45149696-45149696 on Build GRCh38
rs903131582 Chr.17: 45149985-45149985 on Build GRCh38
rs750041515 Chr.17: 45149925-45149925 on Build GRCh38
rs761658949 Chr.17: 45149910-45149910 on Build GRCh38
rs775423795 Chr.17: 45149859-45149859 on Build GRCh38
rs778225514 Chr.17: 45149942-45149942 on Build GRCh38
rs745348258 Chr.17: 45149632-45149632 on Build GRCh38
rs1028875778 Chr.17: 45149751-45149751 on Build GRCh38
rs781539082 Chr.17: 45149951-45149951 on Build GRCh38
rs552223740 Chr.17: 45149793-45149793 on Build GRCh38
rs374259515 Chr.17: 45149610-45149610 on Build GRCh38
rs536863589 Chr.17: 45149926-45149926 on Build GRCh38
rs908824824 Chr.17: 45149837-45149837 on Build GRCh38
rs763269543 Chr.17: 45149590-45149590 on Build GRCh38
rs369049036 Chr.17: 45149592-45149592 on Build GRCh38
rs556337511 Chr.17: 45150017-45150017 on Build GRCh38
rs573692319 Chr.17: 45149602-45149602 on Build GRCh38
rs987456156 Chr.17: 45149829-45149829 on Build GRCh38
rs773571332 Chr.17: 45149688-45149688 on Build GRCh38
rs925151349 Chr.17: 45149676-45149676 on Build GRCh38
rs766123446 Chr.17: 45149934-45149934 on Build GRCh38
rs751387013 Chr.17: 45149745-45149745 on Build GRCh38
rs1044977 Chr.17: 45149847-45149847 on Build GRCh38
rs758822022 Chr.17: 45149650-45149652 on Build GRCh38
rs991827235 Chr.17: 45150009-45150009 on Build GRCh38
rs779434293 Chr.17: 45149637-45149637 on Build GRCh38
rs750750456 Chr.17: 45149594-45149595 on Build GRCh38
rs751186893 Chr.17: 45149615-45149615 on Build GRCh38
rs757165785 Chr.17: 45149748-45149748 on Build GRCh38
rs751786730 Chr.17: 45149990-45149991 on Build GRCh38
rs199709995 Chr.17: 45149933-45149933 on Build GRCh38
rs769890801 Chr.17: 45149973-45149973 on Build GRCh38
rs930368194 Chr.17: 45149929-45149929 on Build GRCh38
rs768692691 Chr.17: 45149871-45149871 on Build GRCh38
rs774469216 Chr.17: 45149888-45149888 on Build GRCh38
rs566184202 Chr.17: 45149919-45149919 on Build GRCh38
rs752095784 Chr.17: 45149943-45149943 on Build GRCh38
rs755603917 Chr.17: 45149787-45149787 on Build GRCh38
rs997548764 Chr.17: 45149740-45149740 on Build GRCh38
rs1024230967 Chr.17: 45149636-45149636 on Build GRCh38
rs752620066 Chr.17: 45149578-45149578 on Build GRCh38
rs771132450 Chr.17: 45149698-45149698 on Build GRCh38
rs748626811 Chr.17: 45149658-45149658 on Build GRCh38
rs756961690 Chr.17: 45149627-45149627 on Build GRCh38
rs780372202 Chr.17: 45149991-45149991 on Build GRCh38
rs765202046 Chr.17: 45149577-45149577 on Build GRCh38
rs77347587 Chr.17: 45149956-45149956 on Build GRCh38
rs753274498 Chr.17: 45149940-45149940 on Build GRCh38
rs148496188 Chr.17: 45149712-45149712 on Build GRCh38
rs199723229 Chr.17: 45149736-45149736 on Build GRCh38
rs749295351 Chr.17: 45149995-45149995 on Build GRCh38
rs552016551 Chr.17: 45149857-45149857 on Build GRCh38
rs747356348 Chr.17: 45149808-45149808 on Build GRCh38
rs775437378 Chr.17: 45149715-45149715 on Build GRCh38
rs530470552 Chr.17: 45149770-45149770 on Build GRCh38
rs146567854 Chr.17: 45149796-45149796 on Build GRCh38
rs763806684 Chr.17: 45149597-45149597 on Build GRCh38
rs894323965 Chr.17: 45149585-45149585 on Build GRCh38

View Full Product Details