Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48112232-48112728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX11
Entrez Gene ID
29916
Gene Name
sorting nexin 11
Gene Aliases
-
UniGene
Hs.15827
Gene Chromosome Location
Chr.17: 48107533-48123064 on Build GRCh38
Transcripts
NM_013323.2 NM_152244.1 XM_005257260.3 XM_005257261.3 XM_005257262.3 XM_011524697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067329 c.68A>G p.D23G Chr.17: 48112599-48112599 on Build GRCh38
5484120 c.119T>A p.I40K Chr.17: 48112650-48112650 on Build GRCh38
4728785 c.124C>A p.L42I Chr.17: 48112655-48112655 on Build GRCh38
7391275 c.88G>T p.G30C Chr.17: 48112619-48112619 on Build GRCh38
4067330 c.73C>T p.R25* Chr.17: 48112604-48112604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976405579 Chr.17: 48112493-48112493 on Build GRCh38
rs770679863 Chr.17: 48112570-48112570 on Build GRCh38
rs984228239 Chr.17: 48112387-48112387 on Build GRCh38
rs746820189 Chr.17: 48112648-48112648 on Build GRCh38
rs780385503 Chr.17: 48112573-48112573 on Build GRCh38
rs376112408 Chr.17: 48112605-48112605 on Build GRCh38
rs756431818 Chr.17: 48112604-48112604 on Build GRCh38
rs764733952 Chr.17: 48112589-48112589 on Build GRCh38
rs151251392 Chr.17: 48112345-48112345 on Build GRCh38
rs762368357 Chr.17: 48112597-48112597 on Build GRCh38
rs771832003 Chr.17: 48112549-48112549 on Build GRCh38
rs748096620 Chr.17: 48112636-48112636 on Build GRCh38
rs200464637 Chr.17: 48112650-48112650 on Build GRCh38
rs370795647 Chr.17: 48112649-48112649 on Build GRCh38
rs993776674 Chr.17: 48112337-48112337 on Build GRCh38
rs755017995 Chr.17: 48112610-48112610 on Build GRCh38
rs373577504 Chr.17: 48112600-48112600 on Build GRCh38
rs775042497 Chr.17: 48112661-48112661 on Build GRCh38
rs541847225 Chr.17: 48112295-48112295 on Build GRCh38
rs757629994 Chr.17: 48112286-48112286 on Build GRCh38
rs754002023 Chr.17: 48112607-48112607 on Build GRCh38
rs776423204 Chr.17: 48112651-48112651 on Build GRCh38
rs966672180 Chr.17: 48112327-48112327 on Build GRCh38
rs781608188 Chr.17: 48112294-48112294 on Build GRCh38
rs768098972 Chr.17: 48112673-48112673 on Build GRCh38
rs373937737 Chr.17: 48112654-48112654 on Build GRCh38
rs927362732 Chr.17: 48112624-48112624 on Build GRCh38
rs374451851 Chr.17: 48112701-48112701 on Build GRCh38
rs748051529 Chr.17: 48112548-48112548 on Build GRCh38
rs199560203 Chr.17: 48112538-48112538 on Build GRCh38
rs201806847 Chr.17: 48112558-48112558 on Build GRCh38
rs770113801 Chr.17: 48112486-48112486 on Build GRCh38
rs1000404740 Chr.17: 48112478-48112478 on Build GRCh38
rs192959924 Chr.17: 48112517-48112517 on Build GRCh38
rs964142134 Chr.17: 48112489-48112489 on Build GRCh38
rs951060912 Chr.17: 48112362-48112362 on Build GRCh38
rs188297760 Chr.17: 48112389-48112389 on Build GRCh38
rs769244222 Chr.17: 48112660-48112660 on Build GRCh38
rs746710158 Chr.17: 48112568-48112568 on Build GRCh38
rs367893026 Chr.17: 48112695-48112695 on Build GRCh38
rs1052346115 Chr.17: 48112578-48112578 on Build GRCh38
rs759128338 Chr.17: 48112587-48112587 on Build GRCh38
rs776370236 Chr.17: 48112584-48112584 on Build GRCh38
rs1019123857 Chr.17: 48112322-48112322 on Build GRCh38
rs927816185 Chr.17: 48112510-48112510 on Build GRCh38
rs750753513 Chr.17: 48112603-48112603 on Build GRCh38
rs370606414 Chr.17: 48112667-48112667 on Build GRCh38
rs774783427 Chr.17: 48112590-48112590 on Build GRCh38
rs1020575023 Chr.17: 48112402-48112402 on Build GRCh38
rs779050065 Chr.17: 48112625-48112625 on Build GRCh38
rs200064194 Chr.17: 48112540-48112540 on Build GRCh38
rs969114120 Chr.17: 48112402-48112402 on Build GRCh38
rs754126821 Chr.17: 48112697-48112697 on Build GRCh38
rs761043956 Chr.17: 48112694-48112694 on Build GRCh38
rs777753905 Chr.17: 48112637-48112637 on Build GRCh38
rs183804905 Chr.17: 48112328-48112328 on Build GRCh38
rs374313217 Chr.17: 48112693-48112693 on Build GRCh38
rs746145318 Chr.17: 48112407-48112407 on Build GRCh38

View Full Product Details