Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48928062-48928570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2Z
Entrez Gene ID
65264
Gene Name
ubiquitin conjugating enzyme E2 Z
Gene Aliases
HOYS7, USE1
UniGene
Hs.514297
Gene Chromosome Location
Chr.17: 48908369-48929060 on Build GRCh38
Transcripts
NM_023079.4

SNP Detail

SNP ID Chromosome Location
rs538702827 Chr.17: 48928434-48928434 on Build GRCh38
rs567930192 Chr.17: 48928367-48928367 on Build GRCh38
rs867349933 Chr.17: 48928509-48928509 on Build GRCh38
rs55685312 Chr.17: 48928180-48928180 on Build GRCh38
rs201929916 Chr.17: 48928339-48928340 on Build GRCh38
rs7221215 Chr.17: 48928432-48928432 on Build GRCh38
rs1046366857 Chr.17: 48928443-48928443 on Build GRCh38
rs866183830 Chr.17: 48928518-48928518 on Build GRCh38
rs114544818 Chr.17: 48928090-48928090 on Build GRCh38
rs1023730283 Chr.17: 48928270-48928270 on Build GRCh38
rs999627356 Chr.17: 48928287-48928287 on Build GRCh38
rs1032429906 Chr.17: 48928483-48928483 on Build GRCh38
rs990080055 Chr.17: 48928093-48928093 on Build GRCh38
rs759111902 Chr.17: 48928088-48928088 on Build GRCh38
rs530352744 Chr.17: 48928396-48928396 on Build GRCh38
rs780199445 Chr.17: 48928167-48928167 on Build GRCh38
rs751349199 Chr.17: 48928334-48928334 on Build GRCh38
rs757060295 Chr.17: 48928358-48928358 on Build GRCh38
rs985725201 Chr.17: 48928409-48928409 on Build GRCh38
rs939795734 Chr.17: 48928171-48928171 on Build GRCh38
rs527304002 Chr.17: 48928248-48928248 on Build GRCh38
rs558651244 Chr.17: 48928464-48928464 on Build GRCh38
rs912307126 Chr.17: 48928150-48928150 on Build GRCh38
rs1028604104 Chr.17: 48928535-48928536 on Build GRCh38
rs550343444 Chr.17: 48928414-48928414 on Build GRCh38
rs28384242 Chr.17: 48928196-48928196 on Build GRCh38
rs1037144206 Chr.17: 48928178-48928178 on Build GRCh38
rs895101744 Chr.17: 48928227-48928227 on Build GRCh38
rs901328801 Chr.17: 48928459-48928459 on Build GRCh38
rs986471283 Chr.17: 48928340-48928343 on Build GRCh38
rs957200164 Chr.17: 48928298-48928298 on Build GRCh38
rs912238268 Chr.17: 48928340-48928340 on Build GRCh38
rs931538900 Chr.17: 48928212-48928212 on Build GRCh38
rs551250375 Chr.17: 48928120-48928122 on Build GRCh38
rs113236693 Chr.17: 48928417-48928417 on Build GRCh38
rs969467435 Chr.17: 48928292-48928292 on Build GRCh38
rs999576691 Chr.17: 48928449-48928449 on Build GRCh38
rs920894277 Chr.17: 48928096-48928096 on Build GRCh38
rs562587672 Chr.17: 48928122-48928122 on Build GRCh38
rs192998454 Chr.17: 48928536-48928536 on Build GRCh38
rs902576602 Chr.17: 48928252-48928253 on Build GRCh38
rs930246788 Chr.17: 48928474-48928474 on Build GRCh38
rs1028237220 Chr.17: 48928337-48928337 on Build GRCh38
rs866794801 Chr.17: 48928422-48928422 on Build GRCh38
rs888442786 Chr.17: 48928235-48928235 on Build GRCh38
rs1005680990 Chr.17: 48928241-48928241 on Build GRCh38
rs1057897 Chr.17: 48928147-48928147 on Build GRCh38
rs545134568 Chr.17: 48928206-48928206 on Build GRCh38
rs898687057 Chr.17: 48928200-48928200 on Build GRCh38
rs1057902 Chr.17: 48928226-48928226 on Build GRCh38
rs982370083 Chr.17: 48928344-48928344 on Build GRCh38
rs763695949 Chr.17: 48928245-48928245 on Build GRCh38
rs988008406 Chr.17: 48928370-48928370 on Build GRCh38
rs946386861 Chr.17: 48928399-48928399 on Build GRCh38
rs954134729 Chr.17: 48928340-48928340 on Build GRCh38
rs183523260 Chr.17: 48928322-48928322 on Build GRCh38
rs764843058 Chr.17: 48928089-48928089 on Build GRCh38

View Full Product Details