Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49848633-49849130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ45513
Entrez Gene ID
729220
Gene Name
uncharacterized LOC729220
Gene Aliases
-
UniGene
Hs.434138
Gene Chromosome Location
Chr.17: 49845910-49848837 on Build GRCh38
Transcripts
NM_001242791.1

SNP Detail

SNP ID Chromosome Location
rs775381860 Chr.17: 49848753-49848753 on Build GRCh38
rs778298724 Chr.17: 49848986-49848986 on Build GRCh38
rs141783307 Chr.17: 49848830-49848830 on Build GRCh38
rs938239909 Chr.17: 49848990-49848990 on Build GRCh38
rs1035177110 Chr.17: 49848734-49848734 on Build GRCh38
rs922426018 Chr.17: 49848859-49848859 on Build GRCh38
rs188746799 Chr.17: 49849002-49849002 on Build GRCh38
rs1027463352 Chr.17: 49848878-49848878 on Build GRCh38
rs1009750046 Chr.17: 49849066-49849066 on Build GRCh38
rs1050058364 Chr.17: 49848792-49848792 on Build GRCh38
rs896610667 Chr.17: 49849048-49849048 on Build GRCh38
rs575081265 Chr.17: 49848709-49848709 on Build GRCh38
rs546255739 Chr.17: 49848691-49848691 on Build GRCh38
rs755230781 Chr.17: 49849001-49849001 on Build GRCh38
rs559864400 Chr.17: 49848708-49848708 on Build GRCh38
rs978415360 Chr.17: 49848819-49848819 on Build GRCh38
rs564471142 Chr.17: 49848919-49848919 on Build GRCh38
rs1041835804 Chr.17: 49849083-49849083 on Build GRCh38
rs896689855 Chr.17: 49848769-49848769 on Build GRCh38
rs1008644224 Chr.17: 49848829-49848829 on Build GRCh38
rs754154955 Chr.17: 49848714-49848714 on Build GRCh38
rs1020037520 Chr.17: 49848790-49848790 on Build GRCh38
rs141673025 Chr.17: 49849098-49849098 on Build GRCh38
rs542016036 Chr.17: 49848727-49848727 on Build GRCh38
rs779394787 Chr.17: 49849036-49849036 on Build GRCh38
rs577326651 Chr.17: 49848663-49848663 on Build GRCh38
rs935514290 Chr.17: 49848717-49848717 on Build GRCh38
rs766739614 Chr.17: 49848710-49848710 on Build GRCh38
rs902913361 Chr.17: 49849085-49849085 on Build GRCh38
rs7213701 Chr.17: 49848789-49848789 on Build GRCh38
rs150575442 Chr.17: 49848844-49848844 on Build GRCh38
rs532888444 Chr.17: 49848918-49848918 on Build GRCh38
rs894034322 Chr.17: 49848782-49848782 on Build GRCh38
rs535158495 Chr.17: 49848835-49848835 on Build GRCh38
rs1032662334 Chr.17: 49848828-49848828 on Build GRCh38
rs908640242 Chr.17: 49848911-49848911 on Build GRCh38
rs546234716 Chr.17: 49848989-49848989 on Build GRCh38
rs138650911 Chr.17: 49848969-49848969 on Build GRCh38
rs916578535 Chr.17: 49848964-49848964 on Build GRCh38
rs949464429 Chr.17: 49848707-49848707 on Build GRCh38
rs994604035 Chr.17: 49848877-49848877 on Build GRCh38
rs975337262 Chr.17: 49848849-49848849 on Build GRCh38
rs983238185 Chr.17: 49848908-49848908 on Build GRCh38
rs967419272 Chr.17: 49848810-49848810 on Build GRCh38
rs1032633359 Chr.17: 49849101-49849101 on Build GRCh38
rs562311268 Chr.17: 49848760-49848760 on Build GRCh38
rs933842504 Chr.17: 49848872-49848872 on Build GRCh38

View Full Product Details