Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50354764-50355269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XYLT2
Entrez Gene ID
64132
Gene Name
xylosyltransferase 2
Gene Aliases
PXYLT2, SOS, XT-II, XT2, xylT-II
UniGene
Hs.463416
Gene Chromosome Location
Chr.17: 50346032-50361185 on Build GRCh38
Transcripts
NM_022167.3 NR_110010.1 XM_005257572.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291367 c.925C>A p.R309R Chr.17: 50354974-50354974 on Build GRCh38
1610454 c.863G>A p.R288Q Chr.17: 50354912-50354912 on Build GRCh38
6199101 c.811G>A p.D271N Chr.17: 50354860-50354860 on Build GRCh38
6824273 c.869C>T p.T290M Chr.17: 50354918-50354918 on Build GRCh38
4741796 c.885T>C p.V295V Chr.17: 50354934-50354934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770092664 Chr.17: 50355057-50355057 on Build GRCh38
rs748408591 Chr.17: 50354931-50354931 on Build GRCh38
rs968505968 Chr.17: 50355115-50355115 on Build GRCh38
rs749150640 Chr.17: 50354828-50354828 on Build GRCh38
rs753502919 Chr.17: 50354910-50354910 on Build GRCh38
rs200767788 Chr.17: 50354908-50354908 on Build GRCh38
rs768108110 Chr.17: 50355077-50355077 on Build GRCh38
rs750057430 Chr.17: 50355104-50355104 on Build GRCh38
rs372647172 Chr.17: 50354848-50354848 on Build GRCh38
rs576962870 Chr.17: 50355003-50355003 on Build GRCh38
rs774856254 Chr.17: 50355073-50355073 on Build GRCh38
rs976472835 Chr.17: 50355169-50355169 on Build GRCh38
rs534585281 Chr.17: 50354907-50354907 on Build GRCh38
rs779991481 Chr.17: 50354921-50354921 on Build GRCh38
rs755065411 Chr.17: 50355015-50355015 on Build GRCh38
rs560912771 Chr.17: 50355144-50355144 on Build GRCh38
rs199506938 Chr.17: 50354919-50354919 on Build GRCh38
rs756299118 Chr.17: 50355018-50355020 on Build GRCh38
rs934756929 Chr.17: 50354871-50354871 on Build GRCh38
rs142341132 Chr.17: 50354927-50354927 on Build GRCh38
rs777998528 Chr.17: 50355062-50355062 on Build GRCh38
rs112084668 Chr.17: 50354882-50354882 on Build GRCh38
rs530143415 Chr.17: 50355155-50355155 on Build GRCh38
rs1021471647 Chr.17: 50355092-50355092 on Build GRCh38
rs376174972 Chr.17: 50354975-50354975 on Build GRCh38
rs762208911 Chr.17: 50354872-50354872 on Build GRCh38
rs1045625596 Chr.17: 50355232-50355232 on Build GRCh38
rs200604247 Chr.17: 50354980-50354987 on Build GRCh38
rs990467752 Chr.17: 50354814-50354814 on Build GRCh38
rs777517215 Chr.17: 50354826-50354826 on Build GRCh38
rs763783567 Chr.17: 50354787-50354787 on Build GRCh38
rs767080751 Chr.17: 50354803-50354803 on Build GRCh38
rs752239898 Chr.17: 50354813-50354813 on Build GRCh38
rs369771174 Chr.17: 50354911-50354911 on Build GRCh38
rs755816240 Chr.17: 50354815-50354815 on Build GRCh38
rs183865553 Chr.17: 50355170-50355170 on Build GRCh38
rs761643084 Chr.17: 50354983-50354983 on Build GRCh38
rs753449527 Chr.17: 50354974-50354974 on Build GRCh38
rs982406140 Chr.17: 50355247-50355247 on Build GRCh38
rs138789012 Chr.17: 50354971-50354971 on Build GRCh38
rs923844804 Chr.17: 50355248-50355248 on Build GRCh38
rs186425233 Chr.17: 50354847-50354847 on Build GRCh38
rs199983537 Chr.17: 50354873-50354873 on Build GRCh38
rs770232369 Chr.17: 50354937-50354937 on Build GRCh38
rs767552605 Chr.17: 50355100-50355101 on Build GRCh38
rs774852996 Chr.17: 50354964-50354964 on Build GRCh38
rs748355606 Chr.17: 50355024-50355024 on Build GRCh38
rs550203224 Chr.17: 50355164-50355164 on Build GRCh38
rs916200857 Chr.17: 50355196-50355196 on Build GRCh38
rs764920884 Chr.17: 50354979-50354979 on Build GRCh38
rs536318185 Chr.17: 50354838-50354838 on Build GRCh38
rs138933160 Chr.17: 50354860-50354860 on Build GRCh38
rs929744879 Chr.17: 50355173-50355173 on Build GRCh38
rs985282415 Chr.17: 50355131-50355131 on Build GRCh38
rs930996431 Chr.17: 50354794-50354794 on Build GRCh38
rs776923746 Chr.17: 50354929-50354929 on Build GRCh38
rs545816817 Chr.17: 50355072-50355072 on Build GRCh38
rs948264765 Chr.17: 50355231-50355231 on Build GRCh38
rs764995882 Chr.17: 50355103-50355103 on Build GRCh38
rs766917700 Chr.17: 50354889-50354889 on Build GRCh38
rs916542461 Chr.17: 50354845-50354845 on Build GRCh38
rs990536947 Chr.17: 50355195-50355195 on Build GRCh38
rs547407791 Chr.17: 50355137-50355137 on Build GRCh38
rs766537621 Chr.17: 50355042-50355042 on Build GRCh38
rs201556386 Chr.17: 50355001-50355001 on Build GRCh38
rs527707924 Chr.17: 50355130-50355130 on Build GRCh38
rs879449843 Chr.17: 50355221-50355221 on Build GRCh38
rs548310973 Chr.17: 50355087-50355087 on Build GRCh38
rs1019798667 Chr.17: 50355154-50355154 on Build GRCh38
rs763535098 Chr.17: 50354884-50354884 on Build GRCh38
rs758253451 Chr.17: 50354984-50354984 on Build GRCh38
rs747171056 Chr.17: 50354926-50354926 on Build GRCh38
rs773672770 Chr.17: 50354946-50354946 on Build GRCh38
rs12451299 Chr.17: 50354963-50354963 on Build GRCh38
rs749723948 Chr.17: 50355070-50355070 on Build GRCh38
rs919606576 Chr.17: 50354785-50354785 on Build GRCh38
rs776168770 Chr.17: 50355080-50355080 on Build GRCh38
rs1048091478 Chr.17: 50355179-50355179 on Build GRCh38
rs1011570514 Chr.17: 50354936-50354936 on Build GRCh38
rs760124216 Chr.17: 50354970-50354970 on Build GRCh38
rs118049882 Chr.17: 50355076-50355076 on Build GRCh38
rs775662377 Chr.17: 50354855-50354855 on Build GRCh38
rs777120321 Chr.17: 50354861-50354861 on Build GRCh38
rs906778024 Chr.17: 50355004-50355004 on Build GRCh38
rs529723937 Chr.17: 50355202-50355202 on Build GRCh38
rs773549850 Chr.17: 50354878-50354878 on Build GRCh38
rs745745346 Chr.17: 50354918-50354918 on Build GRCh38
rs752293943 Chr.17: 50354897-50354897 on Build GRCh38
rs781440043 Chr.17: 50355018-50355018 on Build GRCh38
rs76588845 Chr.17: 50354859-50354859 on Build GRCh38

View Full Product Details