Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50550116-50550603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
UniGene
Hs.103147
Gene Chromosome Location
Chr.17: 50547089-50555852 on Build GRCh38
Transcripts
NM_001258372.1 NM_001258373.1 NM_022827.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5549680 c.1129G>A p.A377T Chr.17: 50550566-50550566 on Build GRCh38
4985951 c.1001A>C p.H334P Chr.17: 50550215-50550215 on Build GRCh38
1266576 c.1002C>A p.H334Q Chr.17: 50550216-50550216 on Build GRCh38
5755070 c.1050G>A p.M350I Chr.17: 50550264-50550264 on Build GRCh38
2699526 c.1156C>T p.R386W Chr.17: 50550593-50550593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368022155 Chr.17: 50550233-50550233 on Build GRCh38
rs551919616 Chr.17: 50550482-50550482 on Build GRCh38
rs10586811 Chr.17: 50550500-50550501 on Build GRCh38
rs376666552 Chr.17: 50550198-50550198 on Build GRCh38
rs796575155 Chr.17: 50550504-50550505 on Build GRCh38
rs1000060209 Chr.17: 50550508-50550508 on Build GRCh38
rs757327000 Chr.17: 50550504-50550504 on Build GRCh38
rs780944968 Chr.17: 50550266-50550266 on Build GRCh38
rs371179753 Chr.17: 50550165-50550165 on Build GRCh38
rs34990884 Chr.17: 50550155-50550156 on Build GRCh38
rs774794902 Chr.17: 50550218-50550218 on Build GRCh38
rs561570027 Chr.17: 50550492-50550492 on Build GRCh38
rs970134710 Chr.17: 50550546-50550546 on Build GRCh38
rs981157869 Chr.17: 50550563-50550563 on Build GRCh38
rs753245415 Chr.17: 50550143-50550143 on Build GRCh38
rs1032339557 Chr.17: 50550325-50550325 on Build GRCh38
rs777043959 Chr.17: 50550520-50550520 on Build GRCh38
rs761211061 Chr.17: 50550242-50550242 on Build GRCh38
rs143489120 Chr.17: 50550277-50550277 on Build GRCh38
rs147159001 Chr.17: 50550366-50550366 on Build GRCh38
rs765881123 Chr.17: 50550342-50550342 on Build GRCh38
rs150117341 Chr.17: 50550286-50550286 on Build GRCh38
rs751255651 Chr.17: 50550175-50550175 on Build GRCh38
rs1052663275 Chr.17: 50550427-50550427 on Build GRCh38
rs1028100807 Chr.17: 50550377-50550377 on Build GRCh38
rs751073977 Chr.17: 50550317-50550317 on Build GRCh38
rs200017654 Chr.17: 50550285-50550285 on Build GRCh38
rs778998861 Chr.17: 50550506-50550506 on Build GRCh38
rs748666868 Chr.17: 50550525-50550525 on Build GRCh38
rs529737167 Chr.17: 50550507-50550507 on Build GRCh38
rs767333045 Chr.17: 50550359-50550359 on Build GRCh38
rs138557972 Chr.17: 50550299-50550299 on Build GRCh38
rs769228040 Chr.17: 50550317-50550317 on Build GRCh38
rs751218668 Chr.17: 50550261-50550261 on Build GRCh38
rs148201021 Chr.17: 50550517-50550517 on Build GRCh38
rs779903001 Chr.17: 50550169-50550169 on Build GRCh38
rs754185263 Chr.17: 50550509-50550510 on Build GRCh38
rs993274031 Chr.17: 50550495-50550495 on Build GRCh38
rs143539531 Chr.17: 50550244-50550244 on Build GRCh38
rs765948946 Chr.17: 50550309-50550309 on Build GRCh38
rs975613832 Chr.17: 50550425-50550425 on Build GRCh38
rs184909241 Chr.17: 50550422-50550422 on Build GRCh38
rs756061843 Chr.17: 50550491-50550491 on Build GRCh38
rs1026133973 Chr.17: 50550497-50550497 on Build GRCh38
rs749834987 Chr.17: 50550247-50550247 on Build GRCh38
rs371604518 Chr.17: 50550252-50550252 on Build GRCh38
rs747519422 Chr.17: 50550300-50550300 on Build GRCh38
rs1019001428 Chr.17: 50550467-50550467 on Build GRCh38
rs750119291 Chr.17: 50550155-50550155 on Build GRCh38
rs770370460 Chr.17: 50550566-50550566 on Build GRCh38
rs375111043 Chr.17: 50550183-50550183 on Build GRCh38
rs761245796 Chr.17: 50550321-50550321 on Build GRCh38
rs558831067 Chr.17: 50550459-50550459 on Build GRCh38
rs375064933 Chr.17: 50550217-50550217 on Build GRCh38
rs762333564 Chr.17: 50550319-50550319 on Build GRCh38
rs201867412 Chr.17: 50550572-50550572 on Build GRCh38
rs1055411341 Chr.17: 50550451-50550451 on Build GRCh38
rs752653984 Chr.17: 50550267-50550267 on Build GRCh38
rs972382889 Chr.17: 50550446-50550446 on Build GRCh38
rs931534341 Chr.17: 50550450-50550450 on Build GRCh38
rs931586543 Chr.17: 50550241-50550241 on Build GRCh38
rs746134512 Chr.17: 50550265-50550265 on Build GRCh38
rs915596004 Chr.17: 50550363-50550363 on Build GRCh38
rs201592753 Chr.17: 50550500-50550500 on Build GRCh38
rs776185210 Chr.17: 50550240-50550240 on Build GRCh38
rs781172250 Chr.17: 50550196-50550196 on Build GRCh38
rs187716579 Chr.17: 50550447-50550447 on Build GRCh38
rs576048597 Chr.17: 50550431-50550431 on Build GRCh38
rs759878594 Chr.17: 50550232-50550232 on Build GRCh38
rs762322783 Chr.17: 50550164-50550165 on Build GRCh38
rs758624436 Chr.17: 50550509-50550509 on Build GRCh38
rs113990008 Chr.17: 50550501-50550502 on Build GRCh38
rs752423234 Chr.17: 50550489-50550489 on Build GRCh38
rs981600083 Chr.17: 50550376-50550376 on Build GRCh38
rs764816152 Chr.17: 50550154-50550154 on Build GRCh38
rs543530876 Chr.17: 50550510-50550510 on Build GRCh38
rs200597817 Chr.17: 50550269-50550269 on Build GRCh38
rs957285092 Chr.17: 50550365-50550365 on Build GRCh38
rs746078289 Chr.17: 50550298-50550298 on Build GRCh38
rs752895603 Chr.17: 50550264-50550265 on Build GRCh38
rs777781132 Chr.17: 50550202-50550202 on Build GRCh38
rs953916792 Chr.17: 50550419-50550419 on Build GRCh38
rs925475525 Chr.17: 50550445-50550445 on Build GRCh38
rs116451719 Chr.17: 50550465-50550465 on Build GRCh38
rs372412098 Chr.17: 50550346-50550346 on Build GRCh38
rs928082431 Chr.17: 50550385-50550385 on Build GRCh38
rs762435159 Chr.17: 50550336-50550336 on Build GRCh38
rs371268284 Chr.17: 50550201-50550201 on Build GRCh38
rs912317758 Chr.17: 50550423-50550423 on Build GRCh38
rs749559375 Chr.17: 50550215-50550215 on Build GRCh38
rs766003758 Chr.17: 50550260-50550260 on Build GRCh38
rs759139329 Chr.17: 50550352-50550352 on Build GRCh38
rs761446592 Chr.17: 50550151-50550151 on Build GRCh38

View Full Product Details