Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 55731590-55732096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM100
Entrez Gene ID
55273
Gene Name
transmembrane protein 100
Gene Aliases
-
UniGene
Hs.173233
Gene Chromosome Location
Chr.17: 55719627-55732121 on Build GRCh38
Transcripts
NM_001099640.1 XM_017024814.1

SNP Detail

SNP ID Chromosome Location
rs976429076 Chr.17: 55731626-55731626 on Build GRCh38
rs540818330 Chr.17: 55731934-55731934 on Build GRCh38
rs997435543 Chr.17: 55731824-55731824 on Build GRCh38
rs185454159 Chr.17: 55731616-55731616 on Build GRCh38
rs986557284 Chr.17: 55731840-55731840 on Build GRCh38
rs990515736 Chr.17: 55732047-55732047 on Build GRCh38
rs191269537 Chr.17: 55731830-55731830 on Build GRCh38
rs749020176 Chr.17: 55731658-55731658 on Build GRCh38
rs1012333811 Chr.17: 55731728-55731728 on Build GRCh38
rs1014747922 Chr.17: 55731778-55731778 on Build GRCh38
rs542619766 Chr.17: 55731877-55731877 on Build GRCh38
rs1006005010 Chr.17: 55731775-55731775 on Build GRCh38
rs181596756 Chr.17: 55731759-55731759 on Build GRCh38
rs182906726 Chr.17: 55731898-55731898 on Build GRCh38
rs551360844 Chr.17: 55731653-55731653 on Build GRCh38
rs190696069 Chr.17: 55731702-55731702 on Build GRCh38
rs1036147320 Chr.17: 55732054-55732054 on Build GRCh38
rs887466731 Chr.17: 55732026-55732026 on Build GRCh38
rs565511798 Chr.17: 55731962-55731962 on Build GRCh38
rs767507398 Chr.17: 55731977-55731977 on Build GRCh38
rs938880166 Chr.17: 55731936-55731936 on Build GRCh38
rs560157920 Chr.17: 55731686-55731686 on Build GRCh38
rs916818086 Chr.17: 55731626-55731626 on Build GRCh38
rs897628309 Chr.17: 55732059-55732059 on Build GRCh38
rs887543621 Chr.17: 55731760-55731760 on Build GRCh38
rs938916390 Chr.17: 55731718-55731718 on Build GRCh38
rs990875491 Chr.17: 55731789-55731789 on Build GRCh38
rs993060964 Chr.17: 55731947-55731947 on Build GRCh38
rs186323488 Chr.17: 55731768-55731768 on Build GRCh38
rs775523480 Chr.17: 55731990-55731990 on Build GRCh38
rs940787517 Chr.17: 55731897-55731897 on Build GRCh38
rs370803364 Chr.17: 55732048-55732048 on Build GRCh38
rs960433710 Chr.17: 55732035-55732035 on Build GRCh38
rs1050827461 Chr.17: 55731921-55731921 on Build GRCh38
rs540919870 Chr.17: 55731667-55731667 on Build GRCh38
rs923295376 Chr.17: 55731642-55731642 on Build GRCh38
rs896145639 Chr.17: 55731727-55731727 on Build GRCh38
rs879093573 Chr.17: 55731912-55731912 on Build GRCh38
rs947722465 Chr.17: 55731982-55731982 on Build GRCh38
rs1039309633 Chr.17: 55731842-55731842 on Build GRCh38
rs930862631 Chr.17: 55731841-55731841 on Build GRCh38
rs1034793397 Chr.17: 55731992-55731992 on Build GRCh38
rs961944209 Chr.17: 55731625-55731625 on Build GRCh38
rs986201202 Chr.17: 55731661-55731661 on Build GRCh38
rs376798178 Chr.17: 55731967-55731967 on Build GRCh38
rs947535423 Chr.17: 55731680-55731680 on Build GRCh38
rs575099354 Chr.17: 55731868-55731868 on Build GRCh38
rs997764762 Chr.17: 55732064-55732064 on Build GRCh38
rs563015424 Chr.17: 55732068-55732068 on Build GRCh38
rs1021637413 Chr.17: 55731856-55731856 on Build GRCh38
rs932011682 Chr.17: 55731913-55731913 on Build GRCh38
rs924809266 Chr.17: 55731704-55731704 on Build GRCh38
rs1023699954 Chr.17: 55732057-55732057 on Build GRCh38
rs534515174 Chr.17: 55731742-55731742 on Build GRCh38
rs779162941 Chr.17: 55731911-55731911 on Build GRCh38
rs748169486 Chr.17: 55731954-55731954 on Build GRCh38
rs567554928 Chr.17: 55731719-55731719 on Build GRCh38
rs968758876 Chr.17: 55731889-55731889 on Build GRCh38
rs901818703 Chr.17: 55731681-55731681 on Build GRCh38
rs550984373 Chr.17: 55731953-55731953 on Build GRCh38
rs902379937 Chr.17: 55731985-55731985 on Build GRCh38
rs118136140 Chr.17: 55731903-55731903 on Build GRCh38
rs375604020 Chr.17: 55732067-55732067 on Build GRCh38
rs1049684122 Chr.17: 55731633-55731633 on Build GRCh38
rs953368407 Chr.17: 55731836-55731836 on Build GRCh38
rs916150771 Chr.17: 55731951-55731951 on Build GRCh38
rs2960089 Chr.17: 55731804-55731804 on Build GRCh38

View Full Product Details