Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56594254-56594744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOG
Entrez Gene ID
9241
Gene Name
noggin
Gene Aliases
SYM1, SYNS1, SYNS1A
UniGene
Hs.248201
Gene Chromosome Location
Chr.17: 56593699-56595590 on Build GRCh38
Transcripts
NM_005450.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5079545 c.465delC p.V157fs*17 Chr.17: 56594688-56594688 on Build GRCh38
5028328 c.410T>A p.L137Q Chr.17: 56594633-56594633 on Build GRCh38
4944935 c.370G>T p.E124* Chr.17: 56594593-56594593 on Build GRCh38
4910058 c.49G>A p.V17I Chr.17: 56594272-56594272 on Build GRCh38
981625 c.414_415insAAGAAGCTG p.L141_R142insKKL Chr.17: 56594637-56594638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987861902 Chr.17: 56594502-56594502 on Build GRCh38
rs200203403 Chr.17: 56594671-56594671 on Build GRCh38
rs774298541 Chr.17: 56594528-56594528 on Build GRCh38
rs376069726 Chr.17: 56594490-56594490 on Build GRCh38
rs373305050 Chr.17: 56594531-56594531 on Build GRCh38
rs758063548 Chr.17: 56594393-56594393 on Build GRCh38
rs764354713 Chr.17: 56594632-56594632 on Build GRCh38
rs780467799 Chr.17: 56594679-56594679 on Build GRCh38
rs758969125 Chr.17: 56594675-56594675 on Build GRCh38
rs1033352311 Chr.17: 56594382-56594382 on Build GRCh38
rs770461784 Chr.17: 56594604-56594604 on Build GRCh38
rs748751127 Chr.17: 56594598-56594598 on Build GRCh38
rs772966707 Chr.17: 56594470-56594470 on Build GRCh38
rs373452045 Chr.17: 56594357-56594357 on Build GRCh38
rs772317007 Chr.17: 56594344-56594344 on Build GRCh38
rs121908949 Chr.17: 56594722-56594722 on Build GRCh38
rs769391519 Chr.17: 56594510-56594510 on Build GRCh38
rs777444621 Chr.17: 56594700-56594700 on Build GRCh38
rs748297297 Chr.17: 56594316-56594316 on Build GRCh38
rs773442175 Chr.17: 56594719-56594719 on Build GRCh38
rs375219346 Chr.17: 56594627-56594627 on Build GRCh38
rs199566527 Chr.17: 56594498-56594498 on Build GRCh38
rs759290193 Chr.17: 56594530-56594530 on Build GRCh38
rs751414234 Chr.17: 56594400-56594400 on Build GRCh38
rs1031172790 Chr.17: 56594405-56594405 on Build GRCh38
rs779513643 Chr.17: 56594566-56594566 on Build GRCh38
rs748158680 Chr.17: 56594409-56594409 on Build GRCh38
rs200253265 Chr.17: 56594293-56594293 on Build GRCh38
rs760935394 Chr.17: 56594545-56594545 on Build GRCh38
rs777064118 Chr.17: 56594503-56594503 on Build GRCh38
rs772774248 Chr.17: 56594516-56594516 on Build GRCh38
rs1054746726 Chr.17: 56594649-56594649 on Build GRCh38
rs781237616 Chr.17: 56594301-56594301 on Build GRCh38
rs1018505000 Chr.17: 56594348-56594348 on Build GRCh38
rs754029476 Chr.17: 56594644-56594644 on Build GRCh38
rs766177414 Chr.17: 56594396-56594396 on Build GRCh38
rs763894353 Chr.17: 56594351-56594351 on Build GRCh38
rs761177860 Chr.17: 56594452-56594452 on Build GRCh38
rs773820340 Chr.17: 56594319-56594319 on Build GRCh38
rs371150691 Chr.17: 56594561-56594561 on Build GRCh38
rs778105304 Chr.17: 56594424-56594424 on Build GRCh38
rs104894611 Chr.17: 56594327-56594327 on Build GRCh38
rs774210447 Chr.17: 56594605-56594605 on Build GRCh38
rs104894614 Chr.17: 56594551-56594551 on Build GRCh38
rs1054563506 Chr.17: 56594547-56594547 on Build GRCh38
rs749247710 Chr.17: 56594469-56594470 on Build GRCh38
rs756242702 Chr.17: 56594417-56594417 on Build GRCh38
rs777342652 Chr.17: 56594590-56594590 on Build GRCh38
rs1003152912 Chr.17: 56594636-56594636 on Build GRCh38
rs375026622 Chr.17: 56594495-56594495 on Build GRCh38
rs868405669 Chr.17: 56594552-56594552 on Build GRCh38
rs544032242 Chr.17: 56594576-56594576 on Build GRCh38
rs994939381 Chr.17: 56594425-56594425 on Build GRCh38
rs775347315 Chr.17: 56594616-56594616 on Build GRCh38
rs535456033 Chr.17: 56594406-56594406 on Build GRCh38
rs752998662 Chr.17: 56594486-56594486 on Build GRCh38
rs765062428 Chr.17: 56594388-56594388 on Build GRCh38
rs104894613 Chr.17: 56594609-56594609 on Build GRCh38
rs763181672 Chr.17: 56594324-56594324 on Build GRCh38
rs776106273 Chr.17: 56594500-56594500 on Build GRCh38
rs771958576 Chr.17: 56594613-56594613 on Build GRCh38
rs894878126 Chr.17: 56594707-56594707 on Build GRCh38
rs892664793 Chr.17: 56594665-56594665 on Build GRCh38
rs768664842 Chr.17: 56594298-56594298 on Build GRCh38
rs774839053 Chr.17: 56594438-56594438 on Build GRCh38
rs764285839 Chr.17: 56594548-56594548 on Build GRCh38
rs777919828 Chr.17: 56594488-56594488 on Build GRCh38
rs879793137 Chr.17: 56594403-56594403 on Build GRCh38
rs554180514 Chr.17: 56594448-56594448 on Build GRCh38
rs138776675 Chr.17: 56594291-56594291 on Build GRCh38
rs906481751 Chr.17: 56594625-56594625 on Build GRCh38
rs762937185 Chr.17: 56594471-56594471 on Build GRCh38
rs750395838 Chr.17: 56594391-56594391 on Build GRCh38
rs778974686 Chr.17: 56594281-56594281 on Build GRCh38
rs770174138 Chr.17: 56594317-56594317 on Build GRCh38
rs200290354 Chr.17: 56594431-56594431 on Build GRCh38
rs1024719173 Chr.17: 56594483-56594483 on Build GRCh38
rs539891840 Chr.17: 56594475-56594475 on Build GRCh38
rs776472075 Chr.17: 56594450-56594450 on Build GRCh38
rs991943783 Chr.17: 56594482-56594482 on Build GRCh38
rs780454130 Chr.17: 56594581-56594581 on Build GRCh38
rs879698029 Chr.17: 56594343-56594343 on Build GRCh38
rs747852410 Chr.17: 56594501-56594501 on Build GRCh38
rs770837708 Chr.17: 56594526-56594526 on Build GRCh38
rs749381932 Chr.17: 56594489-56594489 on Build GRCh38
rs138481449 Chr.17: 56594474-56594474 on Build GRCh38
rs747237879 Chr.17: 56594294-56594294 on Build GRCh38
rs750589581 Chr.17: 56594674-56594674 on Build GRCh38
rs764833488 Chr.17: 56594469-56594469 on Build GRCh38
rs771355844 Chr.17: 56594330-56594330 on Build GRCh38
rs1039686376 Chr.17: 56594606-56594606 on Build GRCh38
rs762410741 Chr.17: 56594524-56594524 on Build GRCh38
rs576666126 Chr.17: 56594540-56594540 on Build GRCh38
rs147499492 Chr.17: 56594437-56594437 on Build GRCh38
rs769458009 Chr.17: 56594586-56594586 on Build GRCh38
rs28937580 Chr.17: 56594326-56594326 on Build GRCh38
rs746297352 Chr.17: 56594442-56594442 on Build GRCh38
rs755324318 Chr.17: 56594683-56594683 on Build GRCh38
rs369585713 Chr.17: 56594648-56594648 on Build GRCh38
rs1001164330 Chr.17: 56594646-56594646 on Build GRCh38
rs117165670 Chr.17: 56594401-56594401 on Build GRCh38
rs1042267510 Chr.17: 56594589-56594589 on Build GRCh38
rs562364308 Chr.17: 56594607-56594607 on Build GRCh38
rs757461366 Chr.17: 56594562-56594562 on Build GRCh38
rs761426737 Chr.17: 56594380-56594380 on Build GRCh38
rs756078666 Chr.17: 56594487-56594487 on Build GRCh38
rs751932364 Chr.17: 56594682-56594682 on Build GRCh38
rs372535983 Chr.17: 56594350-56594350 on Build GRCh38
rs750997261 Chr.17: 56594573-56594573 on Build GRCh38
rs760706732 Chr.17: 56594479-56594479 on Build GRCh38
rs865912614 Chr.17: 56594612-56594612 on Build GRCh38

View Full Product Details