Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76002162-76002666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK3
Entrez Gene ID
1018
Gene Name
cyclin dependent kinase 3
Gene Aliases
-
UniGene
Hs.706766
Gene Chromosome Location
Chr.17: 76000906-76005999 on Build GRCh38
Transcripts
NM_001258.2 NR_037709.1

Gene Symbol
TEN1-CDK3
Entrez Gene ID
100529145
Gene Name
TEN1-CDK3 readthrough (NMD candidate)
Gene Aliases
C17orf106-CDK3
UniGene
Hs.706766
Gene Chromosome Location
Chr.17: 75979217-76005999 on Build GRCh38
Transcripts
NM_001258.2 NR_037709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069749 c.507C>T p.R169R Chr.17: 76002531-76002531 on Build GRCh38
4935092 c.482A>G p.H161R Chr.17: 76002414-76002414 on Build GRCh38
1303365 c.364C>T p.R122W Chr.17: 76002296-76002296 on Build GRCh38
4602066 c.338A>C p.Q113P Chr.17: 76002270-76002270 on Build GRCh38
6625511 c.340G>C p.G114R Chr.17: 76002272-76002272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376616847 Chr.17: 76002543-76002543 on Build GRCh38
rs773664890 Chr.17: 76002382-76002382 on Build GRCh38
rs201685548 Chr.17: 76002296-76002296 on Build GRCh38
rs945345613 Chr.17: 76002530-76002530 on Build GRCh38
rs61747000 Chr.17: 76002531-76002531 on Build GRCh38
rs201794034 Chr.17: 76002313-76002313 on Build GRCh38
rs540948291 Chr.17: 76002524-76002524 on Build GRCh38
rs757877562 Chr.17: 76002231-76002231 on Build GRCh38
rs773857999 Chr.17: 76002431-76002434 on Build GRCh38
rs761223956 Chr.17: 76002213-76002213 on Build GRCh38
rs780129446 Chr.17: 76002349-76002349 on Build GRCh38
rs573764568 Chr.17: 76002443-76002443 on Build GRCh38
rs771824554 Chr.17: 76002493-76002493 on Build GRCh38
rs764812585 Chr.17: 76002223-76002223 on Build GRCh38
rs1041369601 Chr.17: 76002314-76002314 on Build GRCh38
rs371481878 Chr.17: 76002212-76002212 on Build GRCh38
rs377464628 Chr.17: 76002376-76002376 on Build GRCh38
rs148982467 Chr.17: 76002395-76002395 on Build GRCh38
rs142329959 Chr.17: 76002344-76002344 on Build GRCh38
rs746058055 Chr.17: 76002254-76002254 on Build GRCh38
rs774617198 Chr.17: 76002197-76002197 on Build GRCh38
rs764108595 Chr.17: 76002529-76002529 on Build GRCh38
rs755912088 Chr.17: 76002238-76002238 on Build GRCh38
rs201655552 Chr.17: 76002405-76002405 on Build GRCh38
rs751010139 Chr.17: 76002233-76002233 on Build GRCh38
rs753726718 Chr.17: 76002411-76002411 on Build GRCh38
rs2069531 Chr.17: 76002196-76002196 on Build GRCh38
rs375401067 Chr.17: 76002219-76002219 on Build GRCh38
rs866091789 Chr.17: 76002308-76002308 on Build GRCh38
rs763937816 Chr.17: 76002406-76002406 on Build GRCh38
rs779004517 Chr.17: 76002252-76002252 on Build GRCh38
rs374525420 Chr.17: 76002403-76002403 on Build GRCh38
rs762715989 Chr.17: 76002297-76002297 on Build GRCh38
rs752878231 Chr.17: 76002547-76002547 on Build GRCh38
rs879871234 Chr.17: 76002442-76002442 on Build GRCh38
rs763612906 Chr.17: 76002431-76002432 on Build GRCh38
rs374666997 Chr.17: 76002187-76002187 on Build GRCh38
rs750026024 Chr.17: 76002229-76002229 on Build GRCh38
rs775227568 Chr.17: 76002607-76002607 on Build GRCh38
rs748605731 Chr.17: 76002371-76002371 on Build GRCh38
rs751742085 Chr.17: 76002426-76002426 on Build GRCh38
rs748144536 Chr.17: 76002447-76002447 on Build GRCh38
rs747107316 Chr.17: 76002352-76002352 on Build GRCh38
rs752033125 Chr.17: 76002615-76002616 on Build GRCh38
rs780226863 Chr.17: 76002273-76002273 on Build GRCh38
rs761700729 Chr.17: 76002532-76002532 on Build GRCh38
rs201007083 Chr.17: 76002222-76002222 on Build GRCh38
rs763495628 Chr.17: 76002490-76002491 on Build GRCh38
rs896420828 Chr.17: 76002245-76002245 on Build GRCh38
rs374751140 Chr.17: 76002537-76002537 on Build GRCh38
rs765915980 Chr.17: 76002298-76002298 on Build GRCh38
rs984520289 Chr.17: 76002453-76002453 on Build GRCh38
rs778345034 Chr.17: 76002558-76002558 on Build GRCh38
rs768013533 Chr.17: 76002623-76002623 on Build GRCh38
rs779466474 Chr.17: 76002590-76002590 on Build GRCh38
rs1039702806 Chr.17: 76002393-76002393 on Build GRCh38
rs951886209 Chr.17: 76002435-76002435 on Build GRCh38
rs558068161 Chr.17: 76002329-76002329 on Build GRCh38
rs746536767 Chr.17: 76002621-76002621 on Build GRCh38
rs777571429 Chr.17: 76002334-76002334 on Build GRCh38
rs377334389 Chr.17: 76002462-76002462 on Build GRCh38
rs777067823 Chr.17: 76002282-76002282 on Build GRCh38
rs951322395 Chr.17: 76002409-76002409 on Build GRCh38
rs778127690 Chr.17: 76002460-76002460 on Build GRCh38
rs1049810168 Chr.17: 76002548-76002548 on Build GRCh38
rs746233942 Chr.17: 76002592-76002592 on Build GRCh38
rs201961284 Chr.17: 76002201-76002201 on Build GRCh38
rs924308327 Chr.17: 76002596-76002596 on Build GRCh38
rs749715451 Chr.17: 76002567-76002567 on Build GRCh38
rs373239489 Chr.17: 76002323-76002323 on Build GRCh38
rs991972618 Chr.17: 76002509-76002509 on Build GRCh38
rs1009961179 Chr.17: 76002256-76002256 on Build GRCh38
rs776655545 Chr.17: 76002626-76002626 on Build GRCh38
rs144754110 Chr.17: 76002370-76002370 on Build GRCh38
rs772166842 Chr.17: 76002270-76002270 on Build GRCh38
rs749674994 Chr.17: 76002463-76002463 on Build GRCh38
rs766978077 Chr.17: 76002499-76002499 on Build GRCh38
rs755072658 Chr.17: 76002432-76002432 on Build GRCh38
rs760154229 Chr.17: 76002271-76002271 on Build GRCh38
rs752611765 Chr.17: 76002320-76002320 on Build GRCh38
rs760094955 Chr.17: 76002527-76002527 on Build GRCh38
rs767721527 Chr.17: 76002545-76002545 on Build GRCh38
rs369829842 Chr.17: 76002380-76002380 on Build GRCh38
rs756711142 Chr.17: 76002456-76002456 on Build GRCh38
rs752595166 Chr.17: 76002235-76002235 on Build GRCh38
rs762171036 Chr.17: 76002286-76002286 on Build GRCh38
rs768656670 Chr.17: 76002362-76002362 on Build GRCh38
rs763221782 Chr.17: 76002388-76002388 on Build GRCh38
rs949813280 Chr.17: 76002572-76002572 on Build GRCh38
rs1008380389 Chr.17: 76002591-76002591 on Build GRCh38
rs937087926 Chr.17: 76002291-76002291 on Build GRCh38
rs368861061 Chr.17: 76002389-76002389 on Build GRCh38
rs764558017 Chr.17: 76002214-76002214 on Build GRCh38
rs759118252 Chr.17: 76002394-76002394 on Build GRCh38
rs773685805 Chr.17: 76002469-76002469 on Build GRCh38
rs770156329 Chr.17: 76002288-76002288 on Build GRCh38
rs771688809 Chr.17: 76002201-76002202 on Build GRCh38
rs765907594 Chr.17: 76002578-76002578 on Build GRCh38
rs936675752 Chr.17: 76002538-76002538 on Build GRCh38
rs145748425 Chr.17: 76002392-76002392 on Build GRCh38
rs368962392 Chr.17: 76002202-76002202 on Build GRCh38
rs771130789 Chr.17: 76002464-76002464 on Build GRCh38
rs769212297 Chr.17: 76002275-76002275 on Build GRCh38
rs972125613 Chr.17: 76002381-76002381 on Build GRCh38
rs1054473159 Chr.17: 76002633-76002633 on Build GRCh38
rs775177528 Chr.17: 76002248-76002260 on Build GRCh38
rs756088442 Chr.17: 76002552-76002552 on Build GRCh38
rs747090635 Chr.17: 76002274-76002274 on Build GRCh38
rs34918446 Chr.17: 76002303-76002303 on Build GRCh38
rs766594851 Chr.17: 76002418-76002418 on Build GRCh38
rs369742590 Chr.17: 76002309-76002309 on Build GRCh38
rs542699103 Chr.17: 76002600-76002600 on Build GRCh38
rs745464671 Chr.17: 76002483-76002483 on Build GRCh38
rs761705395 Chr.17: 76002638-76002638 on Build GRCh38
rs755004179 Chr.17: 76002234-76002234 on Build GRCh38
rs370317479 Chr.17: 76002505-76002505 on Build GRCh38
rs765863589 Chr.17: 76002232-76002232 on Build GRCh38
rs775210941 Chr.17: 76002511-76002511 on Build GRCh38

View Full Product Details