Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76080151-76080649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZACN
Entrez Gene ID
353174
Gene Name
zinc activated ion channel
Gene Aliases
L2, LGICZ, LGICZ1, ZAC, ZAC1
UniGene
Hs.714919
Gene Chromosome Location
Chr.17: 76079182-76082804 on Build GRCh38
Transcripts
NM_180990.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6824646 c.458C>T p.T153M Chr.17: 76080338-76080338 on Build GRCh38
984598 c.486C>T p.L162L Chr.17: 76080366-76080366 on Build GRCh38
5803429 c.429C>G p.G143G Chr.17: 76080309-76080309 on Build GRCh38
6824647 c.471C>T p.C157C Chr.17: 76080351-76080351 on Build GRCh38
984600 c.519C>G p.L173L Chr.17: 76080399-76080399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759382585 Chr.17: 76080291-76080291 on Build GRCh38
rs573967794 Chr.17: 76080347-76080347 on Build GRCh38
rs556096032 Chr.17: 76080200-76080200 on Build GRCh38
rs753563747 Chr.17: 76080505-76080505 on Build GRCh38
rs372564433 Chr.17: 76080408-76080408 on Build GRCh38
rs985134243 Chr.17: 76080335-76080335 on Build GRCh38
rs139332239 Chr.17: 76080515-76080515 on Build GRCh38
rs143700160 Chr.17: 76080386-76080386 on Build GRCh38
rs771834004 Chr.17: 76080425-76080425 on Build GRCh38
rs780868665 Chr.17: 76080218-76080218 on Build GRCh38
rs149981522 Chr.17: 76080612-76080612 on Build GRCh38
rs954631029 Chr.17: 76080529-76080529 on Build GRCh38
rs769368528 Chr.17: 76080260-76080260 on Build GRCh38
rs1057398831 Chr.17: 76080620-76080621 on Build GRCh38
rs956118907 Chr.17: 76080172-76080172 on Build GRCh38
rs774347702 Chr.17: 76080472-76080472 on Build GRCh38
rs1057503857 Chr.17: 76080439-76080439 on Build GRCh38
rs752814887 Chr.17: 76080292-76080292 on Build GRCh38
rs755381949 Chr.17: 76080398-76080398 on Build GRCh38
rs976820666 Chr.17: 76080567-76080567 on Build GRCh38
rs546667827 Chr.17: 76080510-76080510 on Build GRCh38
rs940418739 Chr.17: 76080212-76080212 on Build GRCh38
rs777543307 Chr.17: 76080409-76080409 on Build GRCh38
rs760831332 Chr.17: 76080523-76080523 on Build GRCh38
rs146741550 Chr.17: 76080290-76080290 on Build GRCh38
rs746762631 Chr.17: 76080430-76080430 on Build GRCh38
rs762096571 Chr.17: 76080376-76080376 on Build GRCh38
rs538509029 Chr.17: 76080588-76080588 on Build GRCh38
rs542658193 Chr.17: 76080367-76080367 on Build GRCh38
rs746265187 Chr.17: 76080371-76080372 on Build GRCh38
rs140811505 Chr.17: 76080287-76080287 on Build GRCh38
rs779827735 Chr.17: 76080538-76080538 on Build GRCh38
rs372983560 Chr.17: 76080313-76080313 on Build GRCh38
rs764511927 Chr.17: 76080501-76080501 on Build GRCh38
rs747716836 Chr.17: 76080239-76080239 on Build GRCh38
rs774397792 Chr.17: 76080359-76080359 on Build GRCh38
rs781624422 Chr.17: 76080404-76080404 on Build GRCh38
rs369965962 Chr.17: 76080208-76080208 on Build GRCh38
rs554235617 Chr.17: 76080621-76080621 on Build GRCh38
rs1055739145 Chr.17: 76080388-76080388 on Build GRCh38
rs775416968 Chr.17: 76080426-76080426 on Build GRCh38
rs183586768 Chr.17: 76080551-76080551 on Build GRCh38
rs774699323 Chr.17: 76080272-76080274 on Build GRCh38
rs535148055 Chr.17: 76080189-76080189 on Build GRCh38
rs772540086 Chr.17: 76080264-76080264 on Build GRCh38
rs144907476 Chr.17: 76080289-76080289 on Build GRCh38
rs558515662 Chr.17: 76080593-76080593 on Build GRCh38
rs781667027 Chr.17: 76080576-76080576 on Build GRCh38
rs754119051 Chr.17: 76080293-76080293 on Build GRCh38
rs888369915 Chr.17: 76080379-76080379 on Build GRCh38
rs762992110 Chr.17: 76080378-76080378 on Build GRCh38
rs926266925 Chr.17: 76080331-76080331 on Build GRCh38
rs768109262 Chr.17: 76080573-76080573 on Build GRCh38
rs748846097 Chr.17: 76080417-76080417 on Build GRCh38
rs768463745 Chr.17: 76080547-76080547 on Build GRCh38
rs950120982 Chr.17: 76080619-76080619 on Build GRCh38
rs2257028 Chr.17: 76080561-76080561 on Build GRCh38
rs756400792 Chr.17: 76080494-76080494 on Build GRCh38
rs141226835 Chr.17: 76080325-76080325 on Build GRCh38
rs920331199 Chr.17: 76080414-76080414 on Build GRCh38
rs149383925 Chr.17: 76080374-76080374 on Build GRCh38
rs918376843 Chr.17: 76080575-76080575 on Build GRCh38
rs1022120340 Chr.17: 76080301-76080301 on Build GRCh38
rs1001523212 Chr.17: 76080226-76080226 on Build GRCh38
rs74826227 Chr.17: 76080580-76080580 on Build GRCh38
rs895716329 Chr.17: 76080615-76080615 on Build GRCh38
rs1014267713 Chr.17: 76080303-76080303 on Build GRCh38
rs994242895 Chr.17: 76080511-76080511 on Build GRCh38
rs766461944 Chr.17: 76080466-76080466 on Build GRCh38
rs755403154 Chr.17: 76080324-76080324 on Build GRCh38
rs776352378 Chr.17: 76080550-76080550 on Build GRCh38
rs747179970 Chr.17: 76080475-76080475 on Build GRCh38
rs372591093 Chr.17: 76080431-76080431 on Build GRCh38
rs116392333 Chr.17: 76080209-76080209 on Build GRCh38
rs1040769863 Chr.17: 76080368-76080368 on Build GRCh38
rs371628139 Chr.17: 76080521-76080521 on Build GRCh38
rs767896996 Chr.17: 76080479-76080479 on Build GRCh38
rs1049423525 Chr.17: 76080177-76080177 on Build GRCh38
rs201365116 Chr.17: 76080377-76080377 on Build GRCh38
rs772285615 Chr.17: 76080460-76080462 on Build GRCh38
rs202236062 Chr.17: 76080307-76080307 on Build GRCh38
rs138720210 Chr.17: 76080338-76080338 on Build GRCh38
rs986358788 Chr.17: 76080518-76080518 on Build GRCh38
rs759510614 Chr.17: 76080563-76080563 on Build GRCh38
rs898873151 Chr.17: 76080274-76080274 on Build GRCh38
rs753154328 Chr.17: 76080482-76080482 on Build GRCh38
rs771140184 Chr.17: 76080558-76080558 on Build GRCh38
rs914538877 Chr.17: 76080188-76080188 on Build GRCh38
rs766408355 Chr.17: 76080380-76080380 on Build GRCh38
rs8066946 Chr.17: 76080423-76080423 on Build GRCh38
rs867880373 Chr.17: 76080403-76080403 on Build GRCh38
rs150846523 Chr.17: 76080422-76080422 on Build GRCh38
rs533015634 Chr.17: 76080446-76080446 on Build GRCh38
rs778566420 Chr.17: 76080516-76080516 on Build GRCh38
rs374986088 Chr.17: 76080203-76080203 on Build GRCh38
rs7210718 Chr.17: 76080202-76080202 on Build GRCh38
rs770097104 Chr.17: 76080436-76080436 on Build GRCh38
rs375294185 Chr.17: 76080339-76080339 on Build GRCh38
rs947387334 Chr.17: 76080197-76080197 on Build GRCh38
rs1048330320 Chr.17: 76080210-76080210 on Build GRCh38
rs1044753103 Chr.17: 76080599-76080599 on Build GRCh38
rs763114983 Chr.17: 76080448-76080448 on Build GRCh38
rs761887504 Chr.17: 76080306-76080306 on Build GRCh38
rs2257020 Chr.17: 76080334-76080334 on Build GRCh38
rs148143793 Chr.17: 76080405-76080405 on Build GRCh38
rs775694503 Chr.17: 76080516-76080528 on Build GRCh38
rs758965476 Chr.17: 76080312-76080312 on Build GRCh38
rs760157411 Chr.17: 76080473-76080473 on Build GRCh38
rs112615474 Chr.17: 76080596-76080596 on Build GRCh38
rs771507326 Chr.17: 76080235-76080240 on Build GRCh38
rs759503538 Chr.17: 76080361-76080361 on Build GRCh38
rs745361753 Chr.17: 76080519-76080519 on Build GRCh38
rs369557345 Chr.17: 76080606-76080606 on Build GRCh38
rs1037778301 Chr.17: 76080241-76080241 on Build GRCh38
rs748794690 Chr.17: 76080340-76080340 on Build GRCh38
rs778914922 Chr.17: 76080275-76080275 on Build GRCh38
rs751964565 Chr.17: 76080318-76080318 on Build GRCh38
rs750500733 Chr.17: 76080310-76080310 on Build GRCh38
rs746929194 Chr.17: 76080544-76080544 on Build GRCh38
rs1021734460 Chr.17: 76080463-76080463 on Build GRCh38
rs751466313 Chr.17: 76080626-76080626 on Build GRCh38
rs749993050 Chr.17: 76080598-76080598 on Build GRCh38
rs1042105153 Chr.17: 76080620-76080620 on Build GRCh38

View Full Product Details