Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78132710-78133205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC8
Entrez Gene ID
147138
Gene Name
transmembrane channel like 8
Gene Aliases
EV2, EVER2, EVIN2
UniGene
Hs.592102
Gene Chromosome Location
Chr.17: 78129296-78142968 on Build GRCh38
Transcripts
NM_152468.4 XM_011524402.2 XM_011524403.2 XM_011524404.2 XM_011524406.2 XM_011524409.2 XM_011524410.2 XM_011524411.2 XM_017024238.1 XM_017024239.1 XM_017024240.1 XM_017024241.1 XM_017024243.1 XM_017024244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624361 c.479C>T p.P160L Chr.17: 78132818-78132818 on Build GRCh38
6195450 c.508C>A p.L170I Chr.17: 78132847-78132847 on Build GRCh38
3403305 c.456G>A p.Q152Q Chr.17: 78132795-78132795 on Build GRCh38
1710844 c.469C>T p.R157C Chr.17: 78132808-78132808 on Build GRCh38
6802878 c.530G>T p.R177M Chr.17: 78132869-78132869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944662590 Chr.17: 78132975-78132975 on Build GRCh38
rs373747548 Chr.17: 78133084-78133084 on Build GRCh38
rs112580325 Chr.17: 78133021-78133021 on Build GRCh38
rs556569953 Chr.17: 78133141-78133141 on Build GRCh38
rs1033268204 Chr.17: 78132923-78132923 on Build GRCh38
rs753915057 Chr.17: 78132826-78132826 on Build GRCh38
rs369252688 Chr.17: 78133061-78133061 on Build GRCh38
rs534681023 Chr.17: 78132991-78132991 on Build GRCh38
rs377532075 Chr.17: 78132893-78132893 on Build GRCh38
rs897565315 Chr.17: 78132734-78132734 on Build GRCh38
rs114426406 Chr.17: 78132976-78132976 on Build GRCh38
rs370056827 Chr.17: 78132900-78132900 on Build GRCh38
rs760616895 Chr.17: 78132914-78132915 on Build GRCh38
rs913271027 Chr.17: 78132820-78132820 on Build GRCh38
rs374134624 Chr.17: 78132763-78132763 on Build GRCh38
rs370037651 Chr.17: 78132849-78132849 on Build GRCh38
rs371997832 Chr.17: 78132789-78132789 on Build GRCh38
rs770636448 Chr.17: 78132916-78132916 on Build GRCh38
rs977024398 Chr.17: 78133018-78133018 on Build GRCh38
rs749611959 Chr.17: 78133105-78133105 on Build GRCh38
rs1036688033 Chr.17: 78133014-78133014 on Build GRCh38
rs1001692639 Chr.17: 78133132-78133132 on Build GRCh38
rs772256858 Chr.17: 78132809-78132809 on Build GRCh38
rs150261314 Chr.17: 78132819-78132819 on Build GRCh38
rs769482747 Chr.17: 78132791-78132791 on Build GRCh38
rs35222654 Chr.17: 78132769-78132769 on Build GRCh38
rs571071403 Chr.17: 78133019-78133019 on Build GRCh38
rs751448350 Chr.17: 78132885-78132885 on Build GRCh38
rs765373712 Chr.17: 78132842-78132842 on Build GRCh38
rs5822228 Chr.17: 78133131-78133131 on Build GRCh38
rs936687469 Chr.17: 78133115-78133115 on Build GRCh38
rs750545069 Chr.17: 78132847-78132847 on Build GRCh38
rs779947513 Chr.17: 78132872-78132872 on Build GRCh38
rs762677074 Chr.17: 78132808-78132808 on Build GRCh38
rs60591028 Chr.17: 78132953-78132953 on Build GRCh38
rs994922331 Chr.17: 78133058-78133058 on Build GRCh38
rs758071857 Chr.17: 78132750-78132750 on Build GRCh38
rs967787162 Chr.17: 78132873-78132873 on Build GRCh38
rs745665969 Chr.17: 78132747-78132747 on Build GRCh38
rs377668069 Chr.17: 78132787-78132787 on Build GRCh38
rs373321007 Chr.17: 78132890-78132890 on Build GRCh38
rs377434153 Chr.17: 78132818-78132818 on Build GRCh38
rs754932707 Chr.17: 78132889-78132889 on Build GRCh38
rs758680213 Chr.17: 78132856-78132856 on Build GRCh38
rs374369638 Chr.17: 78132902-78132902 on Build GRCh38
rs554721346 Chr.17: 78133027-78133027 on Build GRCh38
rs1050249976 Chr.17: 78132950-78132950 on Build GRCh38
rs376056138 Chr.17: 78132972-78132972 on Build GRCh38
rs190820706 Chr.17: 78133142-78133142 on Build GRCh38
rs141101059 Chr.17: 78132800-78132800 on Build GRCh38
rs865835205 Chr.17: 78132738-78132738 on Build GRCh38
rs1001723568 Chr.17: 78132788-78132788 on Build GRCh38
rs936875281 Chr.17: 78132770-78132770 on Build GRCh38
rs769125351 Chr.17: 78132896-78132896 on Build GRCh38
rs112142227 Chr.17: 78132777-78132777 on Build GRCh38
rs779905292 Chr.17: 78132762-78132762 on Build GRCh38
rs796325371 Chr.17: 78133134-78133134 on Build GRCh38
rs761860622 Chr.17: 78132834-78132834 on Build GRCh38
rs952394863 Chr.17: 78132946-78132946 on Build GRCh38
rs536857476 Chr.17: 78133128-78133128 on Build GRCh38

View Full Product Details