Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36087644-36088147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEKT2
Entrez Gene ID
27285
Gene Name
tektin 2
Gene Aliases
TEKTB1, TEKTIN-T, h-tektin-t
UniGene
Hs.127111
Gene Chromosome Location
Chr.1: 36084075-36088275 on Build GRCh38
Transcripts
NM_014466.2 XM_005270753.2 XM_011541258.2 XM_017001055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419169 c.1184G>A p.R395H Chr.1: 36088077-36088077 on Build GRCh38
7427300 c.1008C>T p.Y336Y Chr.1: 36087736-36087736 on Build GRCh38
5965585 c.1079+3A>G p.? Chr.1: 36087810-36087810 on Build GRCh38
7393448 c.1009G>A p.G337S Chr.1: 36087737-36087737 on Build GRCh38
6799368 c.1128C>T p.D376D Chr.1: 36088021-36088021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750876496 Chr.1: 36087764-36087764 on Build GRCh38
rs756758536 Chr.1: 36088039-36088039 on Build GRCh38
rs983387060 Chr.1: 36087772-36087772 on Build GRCh38
rs772426066 Chr.1: 36088077-36088077 on Build GRCh38
rs748353913 Chr.1: 36087986-36087986 on Build GRCh38
rs141119805 Chr.1: 36088008-36088008 on Build GRCh38
rs764272107 Chr.1: 36087754-36087754 on Build GRCh38
rs771915352 Chr.1: 36087827-36087827 on Build GRCh38
rs534486065 Chr.1: 36088034-36088034 on Build GRCh38
rs749521797 Chr.1: 36088073-36088073 on Build GRCh38
rs890719156 Chr.1: 36087922-36087922 on Build GRCh38
rs376282459 Chr.1: 36087679-36087679 on Build GRCh38
rs577850035 Chr.1: 36088101-36088101 on Build GRCh38
rs202132991 Chr.1: 36088071-36088071 on Build GRCh38
rs756781828 Chr.1: 36087749-36087749 on Build GRCh38
rs569596803 Chr.1: 36087844-36087844 on Build GRCh38
rs768447910 Chr.1: 36087718-36087718 on Build GRCh38
rs746775181 Chr.1: 36088076-36088076 on Build GRCh38
rs763779541 Chr.1: 36087907-36087907 on Build GRCh38
rs954915434 Chr.1: 36087706-36087706 on Build GRCh38
rs774931169 Chr.1: 36088017-36088017 on Build GRCh38
rs144739136 Chr.1: 36088075-36088075 on Build GRCh38
rs765304034 Chr.1: 36087763-36087763 on Build GRCh38
rs752006338 Chr.1: 36087794-36087794 on Build GRCh38
rs1023278900 Chr.1: 36087674-36087674 on Build GRCh38
rs547869588 Chr.1: 36087799-36087799 on Build GRCh38
rs745875770 Chr.1: 36087978-36087978 on Build GRCh38
rs769344061 Chr.1: 36087767-36087767 on Build GRCh38
rs763634723 Chr.1: 36088037-36088037 on Build GRCh38
rs539475196 Chr.1: 36088105-36088105 on Build GRCh38
rs145110449 Chr.1: 36087760-36087760 on Build GRCh38
rs399393 Chr.1: 36088072-36088072 on Build GRCh38
rs762800392 Chr.1: 36087744-36087744 on Build GRCh38
rs144497984 Chr.1: 36088007-36088007 on Build GRCh38
rs766801261 Chr.1: 36087776-36087776 on Build GRCh38
rs367655711 Chr.1: 36088052-36088052 on Build GRCh38
rs140017129 Chr.1: 36087737-36087737 on Build GRCh38
rs375278371 Chr.1: 36087976-36087976 on Build GRCh38
rs200994339 Chr.1: 36088085-36088085 on Build GRCh38
rs771792426 Chr.1: 36087981-36087981 on Build GRCh38
rs1034722368 Chr.1: 36088009-36088009 on Build GRCh38
rs755741308 Chr.1: 36087951-36087951 on Build GRCh38
rs755388512 Chr.1: 36087941-36087941 on Build GRCh38
rs767777861 Chr.1: 36087944-36087944 on Build GRCh38
rs150448306 Chr.1: 36088063-36088063 on Build GRCh38
rs765016377 Chr.1: 36087817-36087817 on Build GRCh38
rs770651880 Chr.1: 36087745-36087745 on Build GRCh38
rs755716323 Chr.1: 36087789-36087790 on Build GRCh38
rs755441412 Chr.1: 36087798-36087798 on Build GRCh38
rs143097095 Chr.1: 36088069-36088069 on Build GRCh38
rs1043485043 Chr.1: 36087870-36087870 on Build GRCh38
rs757967144 Chr.1: 36087995-36087995 on Build GRCh38
rs773525781 Chr.1: 36088005-36088005 on Build GRCh38
rs200433257 Chr.1: 36088088-36088088 on Build GRCh38
rs368430902 Chr.1: 36087961-36087961 on Build GRCh38
rs372096279 Chr.1: 36087834-36087834 on Build GRCh38
rs756811649 Chr.1: 36087964-36087964 on Build GRCh38
rs750302885 Chr.1: 36088050-36088050 on Build GRCh38
rs763516202 Chr.1: 36087848-36087848 on Build GRCh38
rs372268636 Chr.1: 36087746-36087746 on Build GRCh38
rs753031169 Chr.1: 36088025-36088025 on Build GRCh38
rs769942508 Chr.1: 36087987-36087987 on Build GRCh38
rs201792935 Chr.1: 36087758-36087758 on Build GRCh38
rs764682428 Chr.1: 36088102-36088102 on Build GRCh38
rs1055002428 Chr.1: 36087836-36087836 on Build GRCh38
rs770884687 Chr.1: 36087824-36087824 on Build GRCh38
rs747821045 Chr.1: 36087806-36087806 on Build GRCh38
rs374252973 Chr.1: 36087938-36087938 on Build GRCh38
rs761138704 Chr.1: 36088033-36088033 on Build GRCh38
rs760133545 Chr.1: 36088019-36088019 on Build GRCh38
rs778403264 Chr.1: 36088041-36088041 on Build GRCh38
rs775161619 Chr.1: 36087985-36087985 on Build GRCh38
rs752332474 Chr.1: 36087948-36087948 on Build GRCh38
rs146068790 Chr.1: 36087729-36087729 on Build GRCh38
rs765304178 Chr.1: 36087843-36087843 on Build GRCh38
rs59194887 Chr.1: 36087915-36087915 on Build GRCh38
rs369864150 Chr.1: 36088097-36088097 on Build GRCh38
rs781029540 Chr.1: 36087724-36087724 on Build GRCh38
rs761946409 Chr.1: 36087835-36087835 on Build GRCh38
rs1050707700 Chr.1: 36087916-36087916 on Build GRCh38
rs780571413 Chr.1: 36087703-36087703 on Build GRCh38
rs755584158 Chr.1: 36087816-36087816 on Build GRCh38
rs368178899 Chr.1: 36087714-36087714 on Build GRCh38
rs771072484 Chr.1: 36088074-36088074 on Build GRCh38
rs199939002 Chr.1: 36087748-36087748 on Build GRCh38
rs370708360 Chr.1: 36087685-36087685 on Build GRCh38
rs767870106 Chr.1: 36088023-36088023 on Build GRCh38
rs372321264 Chr.1: 36087826-36087826 on Build GRCh38
rs753549777 Chr.1: 36088038-36088038 on Build GRCh38
rs181577078 Chr.1: 36087959-36087959 on Build GRCh38
rs758354788 Chr.1: 36088062-36088062 on Build GRCh38

View Full Product Details