Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39412128-39412634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0754
Entrez Gene ID
643314
Gene Name
KIAA0754
Gene Aliases
-
UniGene
Hs.658760
Gene Chromosome Location
Chr.1: 39409504-39416482 on Build GRCh38
Transcripts
NM_015038.1 NM_012090.5 XM_005270694.1 XM_005270695.1 XM_005270696.1 XM_005270697.1 XM_005270698.1 XM_006710519.1 XM_006710520.2 XM_006710521.1 XM_006710522.1 XM_006710524.2 XM_006710526.3 XM_006710528.3 XM_006710529.3 XM_006710530.3 XM_006710531.3 XM_006710532.3 XM_006710533.2 XM_006710535.3 XM_006710536.2 XM_011541141.2 XM_011541142.2 XM_017000873.1 XM_017000874.1 XM_017000875.1 XM_017000876.1 XM_017000877.1 XM_017000878.1 XM_017000879.1 XM_017000880.1 XM_017000881.1 XM_017000882.1

Gene Symbol
MACF1
Entrez Gene ID
23499
Gene Name
microtubule-actin crosslinking factor 1
Gene Aliases
ABP620, ACF7, MACF, OFC4
UniGene
Hs.472475
Gene Chromosome Location
Chr.1: 39084167-39487138 on Build GRCh38
Transcripts
NM_015038.1 NM_012090.5 XM_005270694.1 XM_005270695.1 XM_005270696.1 XM_005270697.1 XM_005270698.1 XM_006710519.1 XM_006710520.2 XM_006710521.1 XM_006710522.1 XM_006710524.2 XM_006710526.3 XM_006710528.3 XM_006710529.3 XM_006710530.3 XM_006710531.3 XM_006710532.3 XM_006710533.2 XM_006710535.3 XM_006710536.2 XM_011541141.2 XM_011541142.2 XM_017000873.1 XM_017000874.1 XM_017000875.1 XM_017000876.1 XM_017000877.1 XM_017000878.1 XM_017000879.1 XM_017000880.1 XM_017000881.1 XM_017000882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260716 c.1695G>T p.M565I Chr.1: 39412368-39412368 on Build GRCh38
4599931 c.1787A>G p.E596G Chr.1: 39412460-39412460 on Build GRCh38
5814696 c.1689A>T p.T563T Chr.1: 39412362-39412362 on Build GRCh38
1645669 c.1517A>C p.N506T Chr.1: 39412190-39412190 on Build GRCh38
4532815 c.1513G>A p.G505R Chr.1: 39412186-39412186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766377950 Chr.1: 39412293-39412293 on Build GRCh38
rs779909505 Chr.1: 39412321-39412321 on Build GRCh38
rs752071678 Chr.1: 39412294-39412294 on Build GRCh38
rs764474956 Chr.1: 39412154-39412154 on Build GRCh38
rs201734236 Chr.1: 39412409-39412409 on Build GRCh38
rs374471195 Chr.1: 39412388-39412388 on Build GRCh38
rs775189974 Chr.1: 39412352-39412352 on Build GRCh38
rs577750358 Chr.1: 39412362-39412362 on Build GRCh38
rs775386603 Chr.1: 39412598-39412598 on Build GRCh38
rs747781874 Chr.1: 39412178-39412178 on Build GRCh38
rs776237759 Chr.1: 39412444-39412444 on Build GRCh38
rs372448047 Chr.1: 39412532-39412532 on Build GRCh38
rs181416924 Chr.1: 39412482-39412482 on Build GRCh38
rs757012584 Chr.1: 39412490-39412490 on Build GRCh38
rs755342342 Chr.1: 39412176-39412176 on Build GRCh38
rs761973203 Chr.1: 39412257-39412257 on Build GRCh38
rs771570352 Chr.1: 39412299-39412299 on Build GRCh38
rs373031092 Chr.1: 39412177-39412177 on Build GRCh38
rs780036070 Chr.1: 39412431-39412431 on Build GRCh38
rs773943650 Chr.1: 39412339-39412339 on Build GRCh38
rs370729605 Chr.1: 39412297-39412297 on Build GRCh38
rs780196349 Chr.1: 39412309-39412310 on Build GRCh38
rs759477008 Chr.1: 39412238-39412238 on Build GRCh38
rs1013637179 Chr.1: 39412206-39412206 on Build GRCh38
rs374338670 Chr.1: 39412453-39412453 on Build GRCh38
rs775375043 Chr.1: 39412249-39412249 on Build GRCh38
rs771195859 Chr.1: 39412557-39412557 on Build GRCh38
rs981231622 Chr.1: 39412283-39412283 on Build GRCh38
rs750526566 Chr.1: 39412278-39412278 on Build GRCh38
rs772096438 Chr.1: 39412348-39412348 on Build GRCh38
rs772642176 Chr.1: 39412190-39412190 on Build GRCh38
rs781077468 Chr.1: 39412529-39412529 on Build GRCh38
rs750313513 Chr.1: 39412161-39412161 on Build GRCh38
rs765468153 Chr.1: 39412276-39412276 on Build GRCh38
rs12141765 Chr.1: 39412500-39412500 on Build GRCh38
rs755767809 Chr.1: 39412319-39412319 on Build GRCh38
rs370747704 Chr.1: 39412383-39412383 on Build GRCh38
rs761277600 Chr.1: 39412603-39412603 on Build GRCh38
rs754113562 Chr.1: 39412477-39412477 on Build GRCh38
rs367871192 Chr.1: 39412471-39412471 on Build GRCh38
rs780764574 Chr.1: 39412504-39412504 on Build GRCh38
rs989079598 Chr.1: 39412585-39412585 on Build GRCh38
rs750011342 Chr.1: 39412501-39412501 on Build GRCh38
rs113273786 Chr.1: 39412163-39412163 on Build GRCh38
rs969811554 Chr.1: 39412279-39412279 on Build GRCh38
rs376262532 Chr.1: 39412264-39412264 on Build GRCh38
rs746854578 Chr.1: 39412432-39412432 on Build GRCh38
rs959105680 Chr.1: 39412427-39412427 on Build GRCh38
rs758499824 Chr.1: 39412282-39412282 on Build GRCh38
rs199527234 Chr.1: 39412417-39412417 on Build GRCh38
rs988741695 Chr.1: 39412340-39412340 on Build GRCh38
rs999392567 Chr.1: 39412479-39412479 on Build GRCh38
rs758490422 Chr.1: 39412276-39412277 on Build GRCh38
rs779373109 Chr.1: 39412509-39412509 on Build GRCh38
rs760073001 Chr.1: 39412366-39412366 on Build GRCh38
rs747185933 Chr.1: 39412375-39412377 on Build GRCh38
rs372397870 Chr.1: 39412439-39412439 on Build GRCh38
rs748820728 Chr.1: 39412328-39412328 on Build GRCh38
rs778880071 Chr.1: 39412158-39412158 on Build GRCh38
rs778599052 Chr.1: 39412394-39412394 on Build GRCh38
rs560189598 Chr.1: 39412452-39412452 on Build GRCh38
rs374253856 Chr.1: 39412359-39412359 on Build GRCh38
rs759594851 Chr.1: 39412457-39412457 on Build GRCh38
rs774459908 Chr.1: 39412456-39412456 on Build GRCh38
rs1032262079 Chr.1: 39412497-39412497 on Build GRCh38
rs759691112 Chr.1: 39412572-39412572 on Build GRCh38
rs746949070 Chr.1: 39412339-39412342 on Build GRCh38
rs781565672 Chr.1: 39412304-39412304 on Build GRCh38
rs774555569 Chr.1: 39412567-39412567 on Build GRCh38
rs767985607 Chr.1: 39412370-39412370 on Build GRCh38
rs748511136 Chr.1: 39412306-39412306 on Build GRCh38
rs754820383 Chr.1: 39412425-39412425 on Build GRCh38
rs764544738 Chr.1: 39412476-39412476 on Build GRCh38
rs202020434 Chr.1: 39412496-39412496 on Build GRCh38
rs531621497 Chr.1: 39412367-39412367 on Build GRCh38
rs777473132 Chr.1: 39412325-39412325 on Build GRCh38
rs957902470 Chr.1: 39412332-39412332 on Build GRCh38
rs113364091 Chr.1: 39412157-39412157 on Build GRCh38
rs764478188 Chr.1: 39412613-39412613 on Build GRCh38
rs541803925 Chr.1: 39412513-39412513 on Build GRCh38
rs369303537 Chr.1: 39412229-39412229 on Build GRCh38
rs371977018 Chr.1: 39412472-39412472 on Build GRCh38
rs376854788 Chr.1: 39412503-39412503 on Build GRCh38
rs147147341 Chr.1: 39412343-39412343 on Build GRCh38
rs767431600 Chr.1: 39412242-39412242 on Build GRCh38
rs751596537 Chr.1: 39412365-39412365 on Build GRCh38
rs753065767 Chr.1: 39412378-39412378 on Build GRCh38
rs376127172 Chr.1: 39412357-39412357 on Build GRCh38
rs1005503643 Chr.1: 39412462-39412462 on Build GRCh38
rs756500345 Chr.1: 39412381-39412381 on Build GRCh38
rs777937033 Chr.1: 39412382-39412382 on Build GRCh38
rs772210286 Chr.1: 39412584-39412584 on Build GRCh38
rs200761968 Chr.1: 39412523-39412523 on Build GRCh38
rs748805543 Chr.1: 39412204-39412204 on Build GRCh38
rs768083567 Chr.1: 39412459-39412459 on Build GRCh38
rs562082053 Chr.1: 39412582-39412582 on Build GRCh38
rs575429508 Chr.1: 39412207-39412207 on Build GRCh38
rs768991964 Chr.1: 39412602-39412603 on Build GRCh38
rs969760677 Chr.1: 39412512-39412512 on Build GRCh38
rs774422328 Chr.1: 39412219-39412219 on Build GRCh38

View Full Product Details