Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80213713-80214222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGSH
Entrez Gene ID
6448
Gene Name
N-sulfoglucosamine sulfohydrolase
Gene Aliases
HSS, MPS3A, SFMD
UniGene
Hs.31074
Gene Chromosome Location
Chr.17: 80200668-80220400 on Build GRCh38
Transcripts
NM_000199.3 XM_005257582.3 XM_005257583.4 XM_011525126.1 XM_011525127.2 XM_017024951.1 XM_017024952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5621812 c.718T>C p.Y240H Chr.17: 80213831-80213831 on Build GRCh38
2803356 c.625G>C p.D209H Chr.17: 80214210-80214210 on Build GRCh38
6505942 c.705C>T p.D235D Chr.17: 80213844-80213844 on Build GRCh38
5713853 c.703G>A p.D235N Chr.17: 80213846-80213846 on Build GRCh38
1130125 c.659T>C p.V220A Chr.17: 80214176-80214176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374255942 Chr.17: 80213739-80213739 on Build GRCh38
rs538131971 Chr.17: 80214089-80214089 on Build GRCh38
rs748102171 Chr.17: 80214027-80214027 on Build GRCh38
rs778580350 Chr.17: 80213787-80213787 on Build GRCh38
rs975256776 Chr.17: 80213966-80213966 on Build GRCh38
rs752502301 Chr.17: 80213767-80213767 on Build GRCh38
rs757720392 Chr.17: 80213922-80213923 on Build GRCh38
rs758960452 Chr.17: 80214063-80214063 on Build GRCh38
rs767054077 Chr.17: 80214067-80214067 on Build GRCh38
rs912449935 Chr.17: 80213953-80213953 on Build GRCh38
rs386799751 Chr.17: 80213921-80213924 on Build GRCh38
rs753472891 Chr.17: 80213846-80213846 on Build GRCh38
rs751436563 Chr.17: 80213762-80213762 on Build GRCh38
rs766771816 Chr.17: 80214168-80214168 on Build GRCh38
rs368107414 Chr.17: 80213898-80213898 on Build GRCh38
rs763800418 Chr.17: 80213845-80213845 on Build GRCh38
rs770822589 Chr.17: 80214147-80214147 on Build GRCh38
rs150508741 Chr.17: 80214177-80214177 on Build GRCh38
rs532471989 Chr.17: 80213955-80213955 on Build GRCh38
rs1008935649 Chr.17: 80214032-80214032 on Build GRCh38
rs372662337 Chr.17: 80214165-80214165 on Build GRCh38
rs769885949 Chr.17: 80214139-80214139 on Build GRCh38
rs746167494 Chr.17: 80213975-80213975 on Build GRCh38
rs765527249 Chr.17: 80214020-80214020 on Build GRCh38
rs754406482 Chr.17: 80214001-80214001 on Build GRCh38
rs772131004 Chr.17: 80213927-80213927 on Build GRCh38
rs938422354 Chr.17: 80214106-80214106 on Build GRCh38
rs367654527 Chr.17: 80213829-80213829 on Build GRCh38
rs767253428 Chr.17: 80213766-80213766 on Build GRCh38
rs938321209 Chr.17: 80214000-80214000 on Build GRCh38
rs181508522 Chr.17: 80214012-80214012 on Build GRCh38
rs773480617 Chr.17: 80213819-80213819 on Build GRCh38
rs928284256 Chr.17: 80213977-80213977 on Build GRCh38
rs763900162 Chr.17: 80213893-80213893 on Build GRCh38
rs761216892 Chr.17: 80213851-80213858 on Build GRCh38
rs758578322 Chr.17: 80214007-80214007 on Build GRCh38
rs767947361 Chr.17: 80214108-80214110 on Build GRCh38
rs533943166 Chr.17: 80213740-80213740 on Build GRCh38
rs750755458 Chr.17: 80214095-80214095 on Build GRCh38
rs751677255 Chr.17: 80214174-80214174 on Build GRCh38
rs765710634 Chr.17: 80214163-80214163 on Build GRCh38
rs760542684 Chr.17: 80213881-80213881 on Build GRCh38
rs374621913 Chr.17: 80213852-80213852 on Build GRCh38
rs767279396 Chr.17: 80214055-80214055 on Build GRCh38
rs755909391 Chr.17: 80213770-80213770 on Build GRCh38
rs566859520 Chr.17: 80214022-80214022 on Build GRCh38
rs752632460 Chr.17: 80213837-80213837 on Build GRCh38
rs769316353 Chr.17: 80213791-80213791 on Build GRCh38
rs772935365 Chr.17: 80213936-80213936 on Build GRCh38
rs761794760 Chr.17: 80214016-80214016 on Build GRCh38
rs756437160 Chr.17: 80213861-80213861 on Build GRCh38
rs923212033 Chr.17: 80213864-80213864 on Build GRCh38
rs768838972 Chr.17: 80214017-80214017 on Build GRCh38
rs111663478 Chr.17: 80213981-80213981 on Build GRCh38
rs104894635 Chr.17: 80213815-80213815 on Build GRCh38
rs748525651 Chr.17: 80213803-80213803 on Build GRCh38
rs747468287 Chr.17: 80213788-80213788 on Build GRCh38
rs756719496 Chr.17: 80213782-80213782 on Build GRCh38
rs151227799 Chr.17: 80214190-80214190 on Build GRCh38
rs536492725 Chr.17: 80214006-80214006 on Build GRCh38
rs748671633 Chr.17: 80213860-80213860 on Build GRCh38
rs868111441 Chr.17: 80214105-80214105 on Build GRCh38
rs559165082 Chr.17: 80213795-80213795 on Build GRCh38
rs546291124 Chr.17: 80213932-80213932 on Build GRCh38
rs6565647 Chr.17: 80214155-80214155 on Build GRCh38
rs372194251 Chr.17: 80214142-80214142 on Build GRCh38
rs1021812109 Chr.17: 80213939-80213939 on Build GRCh38
rs117217614 Chr.17: 80213935-80213935 on Build GRCh38
rs760216365 Chr.17: 80214159-80214159 on Build GRCh38
rs777465002 Chr.17: 80213774-80213774 on Build GRCh38
rs762711093 Chr.17: 80213976-80213976 on Build GRCh38
rs945701545 Chr.17: 80213772-80213772 on Build GRCh38
rs752995788 Chr.17: 80214130-80214131 on Build GRCh38
rs528044353 Chr.17: 80213928-80213928 on Build GRCh38
rs771177142 Chr.17: 80213866-80213866 on Build GRCh38
rs756939944 Chr.17: 80214192-80214192 on Build GRCh38
rs200644359 Chr.17: 80214198-80214198 on Build GRCh38
rs766980170 Chr.17: 80213879-80213879 on Build GRCh38
rs977576958 Chr.17: 80214103-80214103 on Build GRCh38
rs746093033 Chr.17: 80213926-80213926 on Build GRCh38
rs771259698 Chr.17: 80213923-80213923 on Build GRCh38
rs371802886 Chr.17: 80213912-80213912 on Build GRCh38
rs141802955 Chr.17: 80213996-80213996 on Build GRCh38
rs34029730 Chr.17: 80213923-80213924 on Build GRCh38
rs973240120 Chr.17: 80214156-80214156 on Build GRCh38
rs975288528 Chr.17: 80214083-80214083 on Build GRCh38
rs915561976 Chr.17: 80214124-80214124 on Build GRCh38
rs781715169 Chr.17: 80213859-80213859 on Build GRCh38
rs774602372 Chr.17: 80213869-80213869 on Build GRCh38
rs765812346 Chr.17: 80213902-80213902 on Build GRCh38
rs754282279 Chr.17: 80214068-80214068 on Build GRCh38
rs977066227 Chr.17: 80213959-80213959 on Build GRCh38
rs113641837 Chr.17: 80213848-80213848 on Build GRCh38
rs866795427 Chr.17: 80213825-80213825 on Build GRCh38
rs773166549 Chr.17: 80214034-80214034 on Build GRCh38
rs145967352 Chr.17: 80213873-80213873 on Build GRCh38
rs886053560 Chr.17: 80213831-80213831 on Build GRCh38
rs778163282 Chr.17: 80213918-80213918 on Build GRCh38
rs769656022 Chr.17: 80214031-80214031 on Build GRCh38
rs762657621 Chr.17: 80214035-80214035 on Build GRCh38
rs766515536 Chr.17: 80213759-80213759 on Build GRCh38
rs751066044 Chr.17: 80213988-80213988 on Build GRCh38
rs558702724 Chr.17: 80214057-80214057 on Build GRCh38
rs74000629 Chr.17: 80214194-80214194 on Build GRCh38
rs777843748 Chr.17: 80214189-80214189 on Build GRCh38
rs750158313 Chr.17: 80213897-80213897 on Build GRCh38
rs760487031 Chr.17: 80214109-80214109 on Build GRCh38
rs17854435 Chr.17: 80213820-80213820 on Build GRCh38
rs773765512 Chr.17: 80214164-80214164 on Build GRCh38
rs375949842 Chr.17: 80214143-80214143 on Build GRCh38
rs776319554 Chr.17: 80213895-80213895 on Build GRCh38
rs377691452 Chr.17: 80213763-80213763 on Build GRCh38
rs776255192 Chr.17: 80213780-80213781 on Build GRCh38
rs756315850 Chr.17: 80214178-80214178 on Build GRCh38
rs34520362 Chr.17: 80213874-80213874 on Build GRCh38
rs773805076 Chr.17: 80214152-80214152 on Build GRCh38
rs774439487 Chr.17: 80213827-80213827 on Build GRCh38
rs370291256 Chr.17: 80213851-80213851 on Build GRCh38
rs771680187 Chr.17: 80214199-80214199 on Build GRCh38
rs113192021 Chr.17: 80213924-80213924 on Build GRCh38
rs759238037 Chr.17: 80214154-80214154 on Build GRCh38
rs753497239 Chr.17: 80213777-80213777 on Build GRCh38
rs1035779152 Chr.17: 80214125-80214125 on Build GRCh38
rs767416947 Chr.17: 80213836-80213836 on Build GRCh38
rs776680729 Chr.17: 80213930-80213930 on Build GRCh38
rs764492244 Chr.17: 80213997-80213997 on Build GRCh38
rs534162119 Chr.17: 80214047-80214047 on Build GRCh38
rs941825478 Chr.17: 80213998-80213998 on Build GRCh38
rs761740352 Chr.17: 80213896-80213896 on Build GRCh38
rs754948363 Chr.17: 80214137-80214137 on Build GRCh38
rs35087113 Chr.17: 80213921-80213921 on Build GRCh38
rs760510972 Chr.17: 80213839-80213839 on Build GRCh38
rs759996536 Chr.17: 80214020-80214023 on Build GRCh38
rs781271038 Chr.17: 80214024-80214024 on Build GRCh38
rs770944322 Chr.17: 80213822-80213822 on Build GRCh38
rs781436148 Chr.17: 80214138-80214138 on Build GRCh38
rs778017231 Chr.17: 80213862-80213862 on Build GRCh38
rs368461024 Chr.17: 80214129-80214129 on Build GRCh38
rs756603224 Chr.17: 80213915-80213915 on Build GRCh38
rs74841027 Chr.17: 80214085-80214085 on Build GRCh38
rs562717448 Chr.17: 80213865-80213865 on Build GRCh38
rs763076202 Chr.17: 80213755-80213755 on Build GRCh38
rs774199911 Chr.17: 80214015-80214015 on Build GRCh38
rs569059660 Chr.17: 80214004-80214004 on Build GRCh38
rs951397766 Chr.17: 80213785-80213785 on Build GRCh38

View Full Product Details