Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81907112-81907616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYT2
Entrez Gene ID
5833
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
Gene Aliases
ET
UniGene
Hs.569843
Gene Chromosome Location
Chr.17: 81900965-81911477 on Build GRCh38
Transcripts
NM_001184917.2 NM_001256433.2 NM_001256434.2 NM_001256435.2 NM_001282203.1 NM_001282204.1 NM_002861.4 XM_005256386.3 XM_005256387.3 XM_006722287.3 XM_017024910.1 XM_017024911.1 XM_017024912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6282531 c.591+1G>A p.? Chr.17: 81907203-81907203 on Build GRCh38
3737425 c.525C>T p.D175D Chr.17: 81907566-81907566 on Build GRCh38
6386093 c.587C>A p.S196Y Chr.17: 81907208-81907208 on Build GRCh38
1219901 c.512G>T p.R171L Chr.17: 81907579-81907579 on Build GRCh38
5790174 c.501C>T p.S167S Chr.17: 81907590-81907590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958486360 Chr.17: 81907139-81907139 on Build GRCh38
rs1032502287 Chr.17: 81907138-81907138 on Build GRCh38
rs769632882 Chr.17: 81907542-81907542 on Build GRCh38
rs554700919 Chr.17: 81907327-81907327 on Build GRCh38
rs936381545 Chr.17: 81907545-81907545 on Build GRCh38
rs988333923 Chr.17: 81907170-81907170 on Build GRCh38
rs767941149 Chr.17: 81907578-81907578 on Build GRCh38
rs1005963386 Chr.17: 81907482-81907482 on Build GRCh38
rs923645897 Chr.17: 81907326-81907326 on Build GRCh38
rs984210731 Chr.17: 81907509-81907509 on Build GRCh38
rs762155415 Chr.17: 81907574-81907574 on Build GRCh38
rs906351387 Chr.17: 81907435-81907435 on Build GRCh38
rs982043345 Chr.17: 81907499-81907499 on Build GRCh38
rs565465086 Chr.17: 81907407-81907407 on Build GRCh38
rs772007355 Chr.17: 81907461-81907461 on Build GRCh38
rs371049023 Chr.17: 81907519-81907519 on Build GRCh38
rs914181922 Chr.17: 81907175-81907175 on Build GRCh38
rs1025616826 Chr.17: 81907132-81907132 on Build GRCh38
rs372489827 Chr.17: 81907248-81907248 on Build GRCh38
rs959663204 Chr.17: 81907503-81907503 on Build GRCh38
rs775662262 Chr.17: 81907496-81907496 on Build GRCh38
rs958218449 Chr.17: 81907450-81907450 on Build GRCh38
rs984230449 Chr.17: 81907171-81907171 on Build GRCh38
rs752236699 Chr.17: 81907590-81907590 on Build GRCh38
rs994057912 Chr.17: 81907436-81907436 on Build GRCh38
rs778424850 Chr.17: 81907198-81907198 on Build GRCh38
rs897764161 Chr.17: 81907347-81907347 on Build GRCh38
rs942921418 Chr.17: 81907236-81907236 on Build GRCh38
rs770751893 Chr.17: 81907521-81907521 on Build GRCh38
rs544541474 Chr.17: 81907580-81907580 on Build GRCh38
rs1012108934 Chr.17: 81907368-81907368 on Build GRCh38
rs926643147 Chr.17: 81907500-81907500 on Build GRCh38
rs575306151 Chr.17: 81907177-81907177 on Build GRCh38
rs922786063 Chr.17: 81907510-81907510 on Build GRCh38
rs376970397 Chr.17: 81907263-81907263 on Build GRCh38
rs745627865 Chr.17: 81907274-81907274 on Build GRCh38
rs764367108 Chr.17: 81907560-81907560 on Build GRCh38
rs909029330 Chr.17: 81907306-81907306 on Build GRCh38
rs111226512 Chr.17: 81907568-81907568 on Build GRCh38
rs575751107 Chr.17: 81907242-81907243 on Build GRCh38
rs572915653 Chr.17: 81907342-81907342 on Build GRCh38
rs766862276 Chr.17: 81907581-81907581 on Build GRCh38
rs374956302 Chr.17: 81907495-81907495 on Build GRCh38
rs577486641 Chr.17: 81907413-81907413 on Build GRCh38
rs370462105 Chr.17: 81907440-81907440 on Build GRCh38
rs775576674 Chr.17: 81907546-81907546 on Build GRCh38
rs1007306992 Chr.17: 81907363-81907363 on Build GRCh38
rs777459855 Chr.17: 81907518-81907518 on Build GRCh38
rs774765526 Chr.17: 81907564-81907564 on Build GRCh38
rs982094307 Chr.17: 81907239-81907239 on Build GRCh38
rs888812964 Chr.17: 81907356-81907356 on Build GRCh38
rs997145515 Chr.17: 81907348-81907348 on Build GRCh38
rs1051437940 Chr.17: 81907349-81907349 on Build GRCh38
rs759469369 Chr.17: 81907538-81907538 on Build GRCh38
rs973204584 Chr.17: 81907152-81907152 on Build GRCh38
rs926192541 Chr.17: 81907532-81907532 on Build GRCh38
rs540217730 Chr.17: 81907365-81907365 on Build GRCh38
rs747612763 Chr.17: 81907507-81907507 on Build GRCh38
rs542747177 Chr.17: 81907181-81907181 on Build GRCh38
rs771019207 Chr.17: 81907137-81907137 on Build GRCh38
rs941874612 Chr.17: 81907341-81907341 on Build GRCh38
rs1009701349 Chr.17: 81907451-81907451 on Build GRCh38
rs1048997892 Chr.17: 81907297-81907297 on Build GRCh38
rs771750726 Chr.17: 81907508-81907508 on Build GRCh38
rs368923480 Chr.17: 81907559-81907559 on Build GRCh38
rs933218162 Chr.17: 81907557-81907557 on Build GRCh38
rs894877902 Chr.17: 81907355-81907355 on Build GRCh38
rs750826803 Chr.17: 81907579-81907579 on Build GRCh38
rs778813819 Chr.17: 81907143-81907143 on Build GRCh38
rs756598756 Chr.17: 81907161-81907161 on Build GRCh38
rs976943653 Chr.17: 81907501-81907501 on Build GRCh38
rs776444401 Chr.17: 81907522-81907522 on Build GRCh38
rs1032530578 Chr.17: 81907369-81907369 on Build GRCh38
rs936293869 Chr.17: 81907337-81907337 on Build GRCh38
rs952745980 Chr.17: 81907467-81907467 on Build GRCh38
rs1021137229 Chr.17: 81907470-81907470 on Build GRCh38
rs970671534 Chr.17: 81907213-81907213 on Build GRCh38

View Full Product Details