Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82027921-82028420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC45
Entrez Gene ID
201255
Gene Name
leucine rich repeat containing 45
Gene Aliases
-
UniGene
Hs.143774
Gene Chromosome Location
Chr.17: 82023252-82031151 on Build GRCh38
Transcripts
NM_144999.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
437811 c.1094C>T p.A365V Chr.17: 82028280-82028280 on Build GRCh38
1610991 c.1047+4G>T p.? Chr.17: 82028150-82028150 on Build GRCh38
6556752 c.971A>T p.E324V Chr.17: 82028070-82028070 on Build GRCh38
6556751 c.970G>T p.E324* Chr.17: 82028069-82028069 on Build GRCh38
3523754 c.994G>A p.E332K Chr.17: 82028093-82028093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754953981 Chr.17: 82028095-82028095 on Build GRCh38
rs765105152 Chr.17: 82028213-82028213 on Build GRCh38
rs781686785 Chr.17: 82028252-82028252 on Build GRCh38
rs866506995 Chr.17: 82028297-82028297 on Build GRCh38
rs985617023 Chr.17: 82028253-82028253 on Build GRCh38
rs376004370 Chr.17: 82028212-82028212 on Build GRCh38
rs115959429 Chr.17: 82027996-82027996 on Build GRCh38
rs147028558 Chr.17: 82028044-82028044 on Build GRCh38
rs775069825 Chr.17: 82028066-82028106 on Build GRCh38
rs532734054 Chr.17: 82028272-82028272 on Build GRCh38
rs908727349 Chr.17: 82028163-82028163 on Build GRCh38
rs1056288871 Chr.17: 82028342-82028342 on Build GRCh38
rs763675286 Chr.17: 82028041-82028041 on Build GRCh38
rs577629174 Chr.17: 82028283-82028283 on Build GRCh38
rs762505245 Chr.17: 82027980-82027980 on Build GRCh38
rs746167982 Chr.17: 82028254-82028254 on Build GRCh38
rs981472812 Chr.17: 82028311-82028311 on Build GRCh38
rs757611384 Chr.17: 82028238-82028238 on Build GRCh38
rs749733717 Chr.17: 82028354-82028354 on Build GRCh38
rs562880374 Chr.17: 82027947-82027947 on Build GRCh38
rs772697300 Chr.17: 82028192-82028192 on Build GRCh38
rs940450862 Chr.17: 82027952-82027952 on Build GRCh38
rs369464190 Chr.17: 82028371-82028371 on Build GRCh38
rs746467921 Chr.17: 82028360-82028360 on Build GRCh38
rs747281696 Chr.17: 82028024-82028024 on Build GRCh38
rs1022382770 Chr.17: 82028255-82028255 on Build GRCh38
rs535827164 Chr.17: 82028151-82028151 on Build GRCh38
rs746364656 Chr.17: 82028169-82028169 on Build GRCh38
rs370424586 Chr.17: 82028388-82028388 on Build GRCh38
rs377633173 Chr.17: 82028352-82028352 on Build GRCh38
rs770489710 Chr.17: 82028183-82028183 on Build GRCh38
rs767074566 Chr.17: 82028190-82028190 on Build GRCh38
rs751709972 Chr.17: 82028237-82028237 on Build GRCh38
rs780504920 Chr.17: 82028353-82028353 on Build GRCh38
rs369260696 Chr.17: 82028221-82028221 on Build GRCh38
rs764068458 Chr.17: 82028333-82028333 on Build GRCh38
rs777238783 Chr.17: 82028030-82028030 on Build GRCh38
rs1002573889 Chr.17: 82028274-82028274 on Build GRCh38
rs749423772 Chr.17: 82028198-82028198 on Build GRCh38
rs955189834 Chr.17: 82028218-82028218 on Build GRCh38
rs959349689 Chr.17: 82027948-82027948 on Build GRCh38
rs763730335 Chr.17: 82027981-82027981 on Build GRCh38
rs559110270 Chr.17: 82028298-82028298 on Build GRCh38
rs750638760 Chr.17: 82028396-82028396 on Build GRCh38
rs781507380 Chr.17: 82028349-82028349 on Build GRCh38
rs936229114 Chr.17: 82028025-82028025 on Build GRCh38
rs748376279 Chr.17: 82028001-82028001 on Build GRCh38
rs201324260 Chr.17: 82027961-82027961 on Build GRCh38
rs960988930 Chr.17: 82028147-82028147 on Build GRCh38
rs756909158 Chr.17: 82027985-82027985 on Build GRCh38
rs773490993 Chr.17: 82028011-82028011 on Build GRCh38
rs913511936 Chr.17: 82028284-82028284 on Build GRCh38
rs373503054 Chr.17: 82028376-82028376 on Build GRCh38
rs749830148 Chr.17: 82028265-82028265 on Build GRCh38
rs777812169 Chr.17: 82028140-82028140 on Build GRCh38
rs567147404 Chr.17: 82028269-82028269 on Build GRCh38
rs369152792 Chr.17: 82028193-82028193 on Build GRCh38
rs766732573 Chr.17: 82027975-82027975 on Build GRCh38
rs867269892 Chr.17: 82028258-82028258 on Build GRCh38
rs770293879 Chr.17: 82028260-82028260 on Build GRCh38
rs760111002 Chr.17: 82028236-82028237 on Build GRCh38
rs757416425 Chr.17: 82028343-82028343 on Build GRCh38
rs752546807 Chr.17: 82028326-82028326 on Build GRCh38
rs778963352 Chr.17: 82028125-82028125 on Build GRCh38
rs992387064 Chr.17: 82028312-82028312 on Build GRCh38
rs78172878 Chr.17: 82028086-82028086 on Build GRCh38
rs538358837 Chr.17: 82028338-82028338 on Build GRCh38
rs765114302 Chr.17: 82028314-82028314 on Build GRCh38
rs752153420 Chr.17: 82028191-82028191 on Build GRCh38
rs753081478 Chr.17: 82028363-82028387 on Build GRCh38
rs61360272 Chr.17: 82028186-82028186 on Build GRCh38
rs776228927 Chr.17: 82028370-82028370 on Build GRCh38
rs770455444 Chr.17: 82028361-82028361 on Build GRCh38
rs753739520 Chr.17: 82028093-82028093 on Build GRCh38
rs766189850 Chr.17: 82028089-82028089 on Build GRCh38
rs761167948 Chr.17: 82028187-82028187 on Build GRCh38
rs750101555 Chr.17: 82028062-82028062 on Build GRCh38
rs561144243 Chr.17: 82027984-82027984 on Build GRCh38
rs770071516 Chr.17: 82027966-82027966 on Build GRCh38
rs774061951 Chr.17: 82028043-82028043 on Build GRCh38
rs542965854 Chr.17: 82027979-82027979 on Build GRCh38
rs577327035 Chr.17: 82028328-82028328 on Build GRCh38
rs764024752 Chr.17: 82028380-82028380 on Build GRCh38
rs780981618 Chr.17: 82027950-82027950 on Build GRCh38
rs72861736 Chr.17: 82028280-82028280 on Build GRCh38
rs1053291770 Chr.17: 82027990-82027990 on Build GRCh38
rs200171991 Chr.17: 82028026-82028026 on Build GRCh38
rs867550009 Chr.17: 82028241-82028241 on Build GRCh38
rs374025066 Chr.17: 82028351-82028351 on Build GRCh38
rs747287630 Chr.17: 82028144-82028144 on Build GRCh38
rs867072350 Chr.17: 82028134-82028134 on Build GRCh38
rs768043487 Chr.17: 82028263-82028264 on Build GRCh38
rs748113860 Chr.17: 82028128-82028128 on Build GRCh38
rs761932947 Chr.17: 82028387-82028387 on Build GRCh38
rs1040209087 Chr.17: 82028247-82028247 on Build GRCh38
rs1003403856 Chr.17: 82027946-82027946 on Build GRCh38
rs749244445 Chr.17: 82027999-82027999 on Build GRCh38
rs772254764 Chr.17: 82028002-82028002 on Build GRCh38
rs767330027 Chr.17: 82027994-82027994 on Build GRCh38
rs546762726 Chr.17: 82028000-82028000 on Build GRCh38
rs1008833545 Chr.17: 82028224-82028224 on Build GRCh38
rs140038062 Chr.17: 82028397-82028397 on Build GRCh38
rs913209465 Chr.17: 82028029-82028029 on Build GRCh38
rs776283974 Chr.17: 82028293-82028293 on Build GRCh38
rs550219000 Chr.17: 82028053-82028053 on Build GRCh38
rs755941531 Chr.17: 82027997-82027997 on Build GRCh38
rs376877514 Chr.17: 82028383-82028383 on Build GRCh38
rs371786698 Chr.17: 82027998-82027998 on Build GRCh38
rs760161973 Chr.17: 82028039-82028039 on Build GRCh38
rs372185907 Chr.17: 82028135-82028135 on Build GRCh38
rs769259959 Chr.17: 82028355-82028355 on Build GRCh38
rs764588195 Chr.17: 82027984-82027985 on Build GRCh38
rs371537453 Chr.17: 82028197-82028197 on Build GRCh38
rs752706446 Chr.17: 82028220-82028220 on Build GRCh38
rs780375635 Chr.17: 82028263-82028263 on Build GRCh38
rs761243118 Chr.17: 82027968-82027968 on Build GRCh38
rs574914327 Chr.17: 82028378-82028378 on Build GRCh38
rs993817297 Chr.17: 82028207-82028207 on Build GRCh38
rs377589998 Chr.17: 82028143-82028143 on Build GRCh38
rs375239179 Chr.17: 82028358-82028358 on Build GRCh38
rs761402427 Chr.17: 82027968-82027968 on Build GRCh38

View Full Product Details