Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82234814-82235310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A3
Entrez Gene ID
9123
Gene Name
solute carrier family 16 member 3
Gene Aliases
MCT 3, MCT 4, MCT-3, MCT-4, MCT3, MCT4
UniGene
Hs.500761
Gene Chromosome Location
Chr.17: 82228406-82239499 on Build GRCh38
Transcripts
NM_001042422.2 NM_001042423.2 NM_001206950.1 NM_001206951.1 NM_004207.3 XM_011523607.1

SNP Detail

SNP ID Chromosome Location
rs4077541 Chr.17: 82234933-82234933 on Build GRCh38
rs1018920961 Chr.17: 82235285-82235285 on Build GRCh38
rs905523930 Chr.17: 82234918-82234918 on Build GRCh38
rs1011262002 Chr.17: 82235079-82235079 on Build GRCh38
rs973431976 Chr.17: 82234868-82234868 on Build GRCh38
rs772117756 Chr.17: 82235083-82235083 on Build GRCh38
rs868805192 Chr.17: 82235202-82235202 on Build GRCh38
rs777167361 Chr.17: 82235237-82235237 on Build GRCh38
rs778912251 Chr.17: 82235033-82235033 on Build GRCh38
rs919291729 Chr.17: 82235264-82235264 on Build GRCh38
rs746179766 Chr.17: 82234996-82234996 on Build GRCh38
rs953485993 Chr.17: 82234979-82234979 on Build GRCh38
rs1001849204 Chr.17: 82234847-82234847 on Build GRCh38
rs541689739 Chr.17: 82235182-82235182 on Build GRCh38
rs1006627543 Chr.17: 82234878-82234878 on Build GRCh38
rs1012230939 Chr.17: 82234893-82234893 on Build GRCh38
rs972095796 Chr.17: 82235262-82235262 on Build GRCh38
rs386799958 Chr.17: 82235237-82235240 on Build GRCh38
rs1035527042 Chr.17: 82234854-82234854 on Build GRCh38
rs890996671 Chr.17: 82235017-82235017 on Build GRCh38
rs956901049 Chr.17: 82235040-82235040 on Build GRCh38
rs777771180 Chr.17: 82234901-82234901 on Build GRCh38
rs895740880 Chr.17: 82235134-82235134 on Build GRCh38
rs12947333 Chr.17: 82235128-82235128 on Build GRCh38
rs549219674 Chr.17: 82234869-82234869 on Build GRCh38
rs538030726 Chr.17: 82234959-82234959 on Build GRCh38
rs572859065 Chr.17: 82235239-82235239 on Build GRCh38
rs181212912 Chr.17: 82235125-82235125 on Build GRCh38
rs775846476 Chr.17: 82235133-82235133 on Build GRCh38
rs772783901 Chr.17: 82235239-82235239 on Build GRCh38
rs909191841 Chr.17: 82234937-82234937 on Build GRCh38
rs570498492 Chr.17: 82235061-82235061 on Build GRCh38
rs912091641 Chr.17: 82234881-82234881 on Build GRCh38
rs1038318531 Chr.17: 82234841-82234841 on Build GRCh38
rs983529341 Chr.17: 82234858-82234858 on Build GRCh38
rs1020915748 Chr.17: 82235082-82235082 on Build GRCh38
rs963669097 Chr.17: 82235227-82235227 on Build GRCh38
rs530448308 Chr.17: 82234940-82234940 on Build GRCh38
rs12937198 Chr.17: 82235129-82235129 on Build GRCh38
rs748431156 Chr.17: 82234863-82234863 on Build GRCh38
rs555974912 Chr.17: 82235132-82235132 on Build GRCh38
rs912165153 Chr.17: 82235025-82235025 on Build GRCh38
rs375363159 Chr.17: 82234879-82234879 on Build GRCh38
rs892767935 Chr.17: 82235073-82235073 on Build GRCh38
rs896951314 Chr.17: 82234855-82234855 on Build GRCh38
rs532690440 Chr.17: 82234912-82234912 on Build GRCh38
rs767299240 Chr.17: 82235273-82235273 on Build GRCh38
rs1052581397 Chr.17: 82234897-82234897 on Build GRCh38
rs988313941 Chr.17: 82235056-82235056 on Build GRCh38
rs1036234968 Chr.17: 82235186-82235186 on Build GRCh38
rs898992412 Chr.17: 82235230-82235231 on Build GRCh38
rs112021316 Chr.17: 82235237-82235237 on Build GRCh38
rs557616754 Chr.17: 82235008-82235008 on Build GRCh38
rs4077540 Chr.17: 82234902-82234902 on Build GRCh38
rs553663402 Chr.17: 82235238-82235238 on Build GRCh38
rs73366211 Chr.17: 82235154-82235154 on Build GRCh38
rs1050759891 Chr.17: 82234866-82234866 on Build GRCh38
rs905788022 Chr.17: 82234846-82234846 on Build GRCh38
rs540269532 Chr.17: 82235240-82235240 on Build GRCh38
rs745958242 Chr.17: 82235058-82235058 on Build GRCh38
rs1039486999 Chr.17: 82235069-82235069 on Build GRCh38
rs116317447 Chr.17: 82234961-82234961 on Build GRCh38
rs144274216 Chr.17: 82234934-82234934 on Build GRCh38
rs146003680 Chr.17: 82234958-82234958 on Build GRCh38
rs1001430082 Chr.17: 82234919-82234919 on Build GRCh38
rs942054986 Chr.17: 82234947-82234947 on Build GRCh38
rs565372695 Chr.17: 82235272-82235272 on Build GRCh38
rs981610324 Chr.17: 82234907-82234907 on Build GRCh38
rs1035744356 Chr.17: 82234932-82234932 on Build GRCh38
rs546082016 Chr.17: 82235106-82235106 on Build GRCh38
rs942427617 Chr.17: 82235062-82235062 on Build GRCh38
rs548921332 Chr.17: 82234951-82234951 on Build GRCh38
rs540685099 Chr.17: 82234891-82234891 on Build GRCh38
rs1035858595 Chr.17: 82235138-82235138 on Build GRCh38
rs772471901 Chr.17: 82235021-82235021 on Build GRCh38
rs751278061 Chr.17: 82235240-82235240 on Build GRCh38

View Full Product Details